Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38129100-38129602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGM2
Entrez Gene ID
7052
Gene Name
transglutaminase 2
Gene Aliases
TG(C), TGC
UniGene
Hs.517033
Gene Chromosome Location
Chr.20: 38127387-38166578 on Build GRCh38
Transcripts
NM_001323316.1 NM_001323317.1 NM_001323318.1 NM_004613.3 XM_011529028.1

SNP Detail

SNP ID Chromosome Location
rs550561232 Chr.20: 38129463-38129463 on Build GRCh38
rs750016676 Chr.20: 38129146-38129146 on Build GRCh38
rs779705951 Chr.20: 38129353-38129353 on Build GRCh38
rs944061571 Chr.20: 38129294-38129294 on Build GRCh38
rs45455096 Chr.20: 38129157-38129157 on Build GRCh38
rs551145461 Chr.20: 38129577-38129577 on Build GRCh38
rs924035947 Chr.20: 38129514-38129514 on Build GRCh38
rs889694459 Chr.20: 38129158-38129158 on Build GRCh38
rs1025352092 Chr.20: 38129277-38129277 on Build GRCh38
rs149496265 Chr.20: 38129260-38129260 on Build GRCh38
rs796763526 Chr.20: 38129323-38129323 on Build GRCh38
rs187164909 Chr.20: 38129285-38129285 on Build GRCh38
rs7270628 Chr.20: 38129373-38129373 on Build GRCh38
rs539872926 Chr.20: 38129135-38129135 on Build GRCh38
rs1010640507 Chr.20: 38129559-38129559 on Build GRCh38
rs989756593 Chr.20: 38129200-38129200 on Build GRCh38
rs568804875 Chr.20: 38129468-38129468 on Build GRCh38
rs989464162 Chr.20: 38129548-38129548 on Build GRCh38
rs1041368872 Chr.20: 38129578-38129578 on Build GRCh38
rs867743991 Chr.20: 38129190-38129190 on Build GRCh38
rs1040882722 Chr.20: 38129234-38129234 on Build GRCh38
rs953252914 Chr.20: 38129562-38129564 on Build GRCh38
rs1049078032 Chr.20: 38129328-38129328 on Build GRCh38
rs1042872367 Chr.20: 38129139-38129139 on Build GRCh38
rs45482996 Chr.20: 38129560-38129560 on Build GRCh38
rs755755235 Chr.20: 38129239-38129239 on Build GRCh38
rs911266522 Chr.20: 38129558-38129558 on Build GRCh38
rs529245045 Chr.20: 38129441-38129441 on Build GRCh38
rs991512817 Chr.20: 38129385-38129385 on Build GRCh38
rs936675056 Chr.20: 38129148-38129148 on Build GRCh38
rs537426659 Chr.20: 38129504-38129504 on Build GRCh38
rs936768249 Chr.20: 38129535-38129535 on Build GRCh38
rs1804502 Chr.20: 38129325-38129325 on Build GRCh38
rs1046693636 Chr.20: 38129407-38129407 on Build GRCh38
rs888082957 Chr.20: 38129312-38129312 on Build GRCh38
rs906759032 Chr.20: 38129435-38129435 on Build GRCh38
rs1033998479 Chr.20: 38129371-38129371 on Build GRCh38
rs1804503 Chr.20: 38129333-38129333 on Build GRCh38
rs754486308 Chr.20: 38129425-38129425 on Build GRCh38
rs568409183 Chr.20: 38129290-38129290 on Build GRCh38
rs748170105 Chr.20: 38129492-38129492 on Build GRCh38
rs1005160640 Chr.20: 38129327-38129327 on Build GRCh38
rs921788514 Chr.20: 38129246-38129246 on Build GRCh38
rs895295658 Chr.20: 38129353-38129354 on Build GRCh38
rs182470097 Chr.20: 38129196-38129196 on Build GRCh38
rs918677636 Chr.20: 38129315-38129315 on Build GRCh38
rs563927379 Chr.20: 38129151-38129151 on Build GRCh38
rs931406988 Chr.20: 38129319-38129319 on Build GRCh38
rs143869572 Chr.20: 38129268-38129268 on Build GRCh38
rs1062737 Chr.20: 38129484-38129484 on Build GRCh38

View Full Product Details