Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44206796-44207303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSER1
Entrez Gene ID
51526
Gene Name
oxidative stress responsive serine rich 1
Gene Aliases
C20orf111, HSPC207, Osr1, Perit1, dJ1183I21.1
UniGene
Hs.75798
Gene Chromosome Location
Chr.20: 44195939-44217229 on Build GRCh38
Transcripts
NM_016470.7 XM_011528851.1 XM_011528853.2 XM_011528854.2 XM_017027873.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5916810 c.25G>A p.E9K Chr.20: 44206933-44206933 on Build GRCh38
5560337 c.3G>A p.M1I Chr.20: 44206955-44206955 on Build GRCh38
443862 c.3G>C p.M1I Chr.20: 44206955-44206955 on Build GRCh38
5741892 c.10G>A p.E4K Chr.20: 44206948-44206948 on Build GRCh38
4620393 c.75A>C p.S25S Chr.20: 44206883-44206883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565901704 Chr.20: 44207135-44207135 on Build GRCh38
rs775939935 Chr.20: 44206925-44206925 on Build GRCh38
rs1008274818 Chr.20: 44207171-44207171 on Build GRCh38
rs2232286 Chr.20: 44207117-44207117 on Build GRCh38
rs752656757 Chr.20: 44206971-44206971 on Build GRCh38
rs574637437 Chr.20: 44207204-44207204 on Build GRCh38
rs967256735 Chr.20: 44207276-44207276 on Build GRCh38
rs530163803 Chr.20: 44206940-44206940 on Build GRCh38
rs554896025 Chr.20: 44207199-44207199 on Build GRCh38
rs113650683 Chr.20: 44207267-44207267 on Build GRCh38
rs928384694 Chr.20: 44207016-44207016 on Build GRCh38
rs1057036867 Chr.20: 44207063-44207063 on Build GRCh38
rs754545132 Chr.20: 44206843-44206843 on Build GRCh38
rs772148035 Chr.20: 44206922-44206922 on Build GRCh38
rs746502641 Chr.20: 44207123-44207123 on Build GRCh38
rs2232284 Chr.20: 44207275-44207275 on Build GRCh38
rs1041368240 Chr.20: 44207012-44207012 on Build GRCh38
rs186587148 Chr.20: 44206824-44206824 on Build GRCh38
rs1012265661 Chr.20: 44207069-44207069 on Build GRCh38
rs576099599 Chr.20: 44207058-44207058 on Build GRCh38
rs745891778 Chr.20: 44206918-44206918 on Build GRCh38
rs781207817 Chr.20: 44206848-44206848 on Build GRCh38
rs987053669 Chr.20: 44206870-44206870 on Build GRCh38
rs371334414 Chr.20: 44206982-44206982 on Build GRCh38
rs762498187 Chr.20: 44206943-44206943 on Build GRCh38
rs570172263 Chr.20: 44206999-44206999 on Build GRCh38
rs377611248 Chr.20: 44206965-44206965 on Build GRCh38
rs1015517115 Chr.20: 44207118-44207118 on Build GRCh38
rs747539051 Chr.20: 44206938-44206938 on Build GRCh38
rs1018533653 Chr.20: 44207188-44207188 on Build GRCh38
rs756532916 Chr.20: 44207020-44207020 on Build GRCh38
rs763794108 Chr.20: 44206990-44206990 on Build GRCh38
rs2232285 Chr.20: 44207161-44207161 on Build GRCh38
rs938421016 Chr.20: 44207019-44207019 on Build GRCh38
rs774147753 Chr.20: 44206949-44206949 on Build GRCh38
rs1012360618 Chr.20: 44207091-44207091 on Build GRCh38
rs976350980 Chr.20: 44207156-44207160 on Build GRCh38
rs576554978 Chr.20: 44207278-44207278 on Build GRCh38
rs539087266 Chr.20: 44207072-44207072 on Build GRCh38
rs189688626 Chr.20: 44206913-44206913 on Build GRCh38
rs755814981 Chr.20: 44206883-44206883 on Build GRCh38
rs775469456 Chr.20: 44207001-44207003 on Build GRCh38
rs989962464 Chr.20: 44207274-44207274 on Build GRCh38
rs192810455 Chr.20: 44206833-44206833 on Build GRCh38
rs750113098 Chr.20: 44207119-44207119 on Build GRCh38
rs989890648 Chr.20: 44207242-44207242 on Build GRCh38
rs771253796 Chr.20: 44206915-44206915 on Build GRCh38
rs953052823 Chr.20: 44207197-44207197 on Build GRCh38
rs779041131 Chr.20: 44207000-44207000 on Build GRCh38
rs550187705 Chr.20: 44206995-44206995 on Build GRCh38
rs576418441 Chr.20: 44207160-44207160 on Build GRCh38
rs898734511 Chr.20: 44207112-44207112 on Build GRCh38
rs184717997 Chr.20: 44206873-44206873 on Build GRCh38
rs966292809 Chr.20: 44207147-44207147 on Build GRCh38
rs1038630428 Chr.20: 44207097-44207097 on Build GRCh38
rs765002959 Chr.20: 44206896-44206897 on Build GRCh38
rs200554027 Chr.20: 44207071-44207072 on Build GRCh38
rs567839198 Chr.20: 44206916-44206916 on Build GRCh38
rs759075405 Chr.20: 44206950-44206950 on Build GRCh38
rs765994337 Chr.20: 44206948-44206948 on Build GRCh38

View Full Product Details