Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45098334-45098829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNS1
Entrez Gene ID
3787
Gene Name
potassium voltage-gated channel modifier subfamily S member 1
Gene Aliases
Kv9.1
UniGene
Hs.117780
Gene Chromosome Location
Chr.20: 45091214-45101112 on Build GRCh38
Transcripts
NM_001322799.1 NM_002251.4 XM_017027846.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6691809 c.166G>A p.V56M Chr.20: 45098606-45098606 on Build GRCh38
4624258 c.384G>A p.E128E Chr.20: 45098388-45098388 on Build GRCh38
3758591 c.258G>A p.E86E Chr.20: 45098514-45098514 on Build GRCh38
4653290 c.373G>A p.V125M Chr.20: 45098399-45098399 on Build GRCh38
3740149 c.299G>A p.R100H Chr.20: 45098473-45098473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924554573 Chr.20: 45098456-45098456 on Build GRCh38
rs968994291 Chr.20: 45098408-45098408 on Build GRCh38
rs909631248 Chr.20: 45098597-45098597 on Build GRCh38
rs866211337 Chr.20: 45098449-45098449 on Build GRCh38
rs183256639 Chr.20: 45098730-45098730 on Build GRCh38
rs6104005 Chr.20: 45098501-45098501 on Build GRCh38
rs371968715 Chr.20: 45098661-45098661 on Build GRCh38
rs576341455 Chr.20: 45098664-45098664 on Build GRCh38
rs760160451 Chr.20: 45098635-45098635 on Build GRCh38
rs770087624 Chr.20: 45098479-45098479 on Build GRCh38
rs776877543 Chr.20: 45098362-45098362 on Build GRCh38
rs781566611 Chr.20: 45098587-45098587 on Build GRCh38
rs752197004 Chr.20: 45098504-45098504 on Build GRCh38
rs755017938 Chr.20: 45098702-45098702 on Build GRCh38
rs753719398 Chr.20: 45098657-45098657 on Build GRCh38
rs751203756 Chr.20: 45098376-45098376 on Build GRCh38
rs1040376123 Chr.20: 45098358-45098358 on Build GRCh38
rs376610403 Chr.20: 45098717-45098717 on Build GRCh38
rs777743004 Chr.20: 45098410-45098410 on Build GRCh38
rs932021768 Chr.20: 45098638-45098638 on Build GRCh38
rs750286854 Chr.20: 45098668-45098668 on Build GRCh38
rs758267249 Chr.20: 45098669-45098669 on Build GRCh38
rs968820357 Chr.20: 45098694-45098694 on Build GRCh38
rs771749697 Chr.20: 45098621-45098621 on Build GRCh38
rs763600774 Chr.20: 45098644-45098644 on Build GRCh38
rs752664327 Chr.20: 45098402-45098402 on Build GRCh38
rs370688992 Chr.20: 45098789-45098789 on Build GRCh38
rs780395600 Chr.20: 45098556-45098556 on Build GRCh38
rs764005105 Chr.20: 45098507-45098507 on Build GRCh38
rs759151235 Chr.20: 45098498-45098498 on Build GRCh38
rs770175779 Chr.20: 45098598-45098598 on Build GRCh38
rs772280462 Chr.20: 45098442-45098442 on Build GRCh38
rs750703786 Chr.20: 45098524-45098524 on Build GRCh38
rs375528670 Chr.20: 45098715-45098715 on Build GRCh38
rs1017677972 Chr.20: 45098404-45098404 on Build GRCh38
rs757013104 Chr.20: 45098521-45098521 on Build GRCh38
rs747093109 Chr.20: 45098457-45098457 on Build GRCh38
rs766016752 Chr.20: 45098365-45098365 on Build GRCh38
rs749718334 Chr.20: 45098733-45098733 on Build GRCh38
rs898100347 Chr.20: 45098627-45098627 on Build GRCh38
rs754498637 Chr.20: 45098379-45098379 on Build GRCh38
rs779334922 Chr.20: 45098436-45098436 on Build GRCh38
rs536644815 Chr.20: 45098566-45098566 on Build GRCh38
rs760198636 Chr.20: 45098506-45098506 on Build GRCh38
rs749076353 Chr.20: 45098415-45098415 on Build GRCh38
rs554416667 Chr.20: 45098474-45098474 on Build GRCh38
rs768771653 Chr.20: 45098586-45098586 on Build GRCh38
rs762209311 Chr.20: 45098361-45098361 on Build GRCh38
rs35456522 Chr.20: 45098802-45098802 on Build GRCh38
rs748623492 Chr.20: 45098593-45098593 on Build GRCh38
rs775018492 Chr.20: 45098734-45098734 on Build GRCh38
rs748626741 Chr.20: 45098716-45098716 on Build GRCh38
rs751752353 Chr.20: 45098670-45098670 on Build GRCh38
rs755958406 Chr.20: 45098406-45098406 on Build GRCh38
rs778779068 Chr.20: 45098523-45098523 on Build GRCh38
rs978599031 Chr.20: 45098430-45098430 on Build GRCh38
rs758641099 Chr.20: 45098548-45098548 on Build GRCh38
rs34643859 Chr.20: 45098726-45098726 on Build GRCh38
rs201526054 Chr.20: 45098655-45098655 on Build GRCh38
rs201080873 Chr.20: 45098394-45098394 on Build GRCh38
rs769086547 Chr.20: 45098465-45098465 on Build GRCh38
rs762145037 Chr.20: 45098478-45098478 on Build GRCh38
rs948459284 Chr.20: 45098540-45098540 on Build GRCh38
rs773997776 Chr.20: 45098484-45098484 on Build GRCh38
rs534791107 Chr.20: 45098441-45098441 on Build GRCh38
rs773518281 Chr.20: 45098613-45098613 on Build GRCh38
rs757070162 Chr.20: 45098416-45098416 on Build GRCh38
rs368678884 Chr.20: 45098633-45098633 on Build GRCh38
rs13043825 Chr.20: 45098514-45098514 on Build GRCh38
rs767038455 Chr.20: 45098499-45098499 on Build GRCh38
rs556885291 Chr.20: 45098615-45098615 on Build GRCh38
rs1050587783 Chr.20: 45098648-45098648 on Build GRCh38

View Full Product Details