Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45815199-45815724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2C
Entrez Gene ID
11065
Gene Name
ubiquitin conjugating enzyme E2 C
Gene Aliases
UBCH10, dJ447F3.2
UniGene
Hs.93002
Gene Chromosome Location
Chr.20: 45812576-45816957 on Build GRCh38
Transcripts
NM_001281741.1 NM_001281742.1 NM_007019.3 NM_181799.2 NM_181800.2 NM_181801.3 NR_104036.1 NR_104037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
724441 c.387G>T p.R129S Chr.20: 45815711-45815711 on Build GRCh38
3547092 c.352C>T p.L118L Chr.20: 45815676-45815676 on Build GRCh38
6899731 c.384C>T p.V128V Chr.20: 45815708-45815708 on Build GRCh38
5692055 c.294C>T p.P98P Chr.20: 45815467-45815467 on Build GRCh38
6815055 c.217-5G>A p.? Chr.20: 45815536-45815536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533696759 Chr.20: 45815657-45815657 on Build GRCh38
rs760491592 Chr.20: 45815380-45815380 on Build GRCh38
rs758135325 Chr.20: 45815599-45815599 on Build GRCh38
rs755388093 Chr.20: 45815424-45815424 on Build GRCh38
rs775502728 Chr.20: 45815379-45815379 on Build GRCh38
rs754226233 Chr.20: 45815591-45815591 on Build GRCh38
rs1044464325 Chr.20: 45815500-45815500 on Build GRCh38
rs747579357 Chr.20: 45815415-45815415 on Build GRCh38
rs375815317 Chr.20: 45815506-45815506 on Build GRCh38
rs751722028 Chr.20: 45815576-45815576 on Build GRCh38
rs780038846 Chr.20: 45815404-45815404 on Build GRCh38
rs745539672 Chr.20: 45815537-45815537 on Build GRCh38
rs193140601 Chr.20: 45815354-45815354 on Build GRCh38
rs34158793 Chr.20: 45815527-45815527 on Build GRCh38
rs774074252 Chr.20: 45815373-45815373 on Build GRCh38
rs761307294 Chr.20: 45815668-45815668 on Build GRCh38
rs566343358 Chr.20: 45815603-45815603 on Build GRCh38
rs753193301 Chr.20: 45815508-45815508 on Build GRCh38
rs773543281 Chr.20: 45815627-45815629 on Build GRCh38
rs368265822 Chr.20: 45815570-45815570 on Build GRCh38
rs748102960 Chr.20: 45815612-45815612 on Build GRCh38
rs748461028 Chr.20: 45815432-45815432 on Build GRCh38
rs771830619 Chr.20: 45815538-45815538 on Build GRCh38
rs759764860 Chr.20: 45815643-45815643 on Build GRCh38
rs374439898 Chr.20: 45815592-45815592 on Build GRCh38
rs776555374 Chr.20: 45815455-45815455 on Build GRCh38
rs747412622 Chr.20: 45815340-45815340 on Build GRCh38
rs568818326 Chr.20: 45815501-45815501 on Build GRCh38
rs78668952 Chr.20: 45815460-45815460 on Build GRCh38
rs1050715422 Chr.20: 45815246-45815246 on Build GRCh38
rs367706641 Chr.20: 45815607-45815607 on Build GRCh38
rs759107820 Chr.20: 45815375-45815375 on Build GRCh38
rs370295230 Chr.20: 45815363-45815363 on Build GRCh38
rs950123867 Chr.20: 45815498-45815498 on Build GRCh38
rs753552447 Chr.20: 45815384-45815384 on Build GRCh38
rs558429475 Chr.20: 45815693-45815693 on Build GRCh38
rs547850953 Chr.20: 45815558-45815558 on Build GRCh38
rs978337546 Chr.20: 45815343-45815343 on Build GRCh38
rs145980514 Chr.20: 45815435-45815435 on Build GRCh38
rs750559851 Chr.20: 45815398-45815398 on Build GRCh38
rs769604009 Chr.20: 45815562-45815562 on Build GRCh38
rs371491283 Chr.20: 45815258-45815258 on Build GRCh38
rs369963837 Chr.20: 45815527-45815527 on Build GRCh38
rs759441937 Chr.20: 45815658-45815658 on Build GRCh38
rs778333049 Chr.20: 45815590-45815590 on Build GRCh38
rs768695868 Chr.20: 45815549-45815549 on Build GRCh38
rs756640882 Chr.20: 45815585-45815585 on Build GRCh38
rs989240851 Chr.20: 45815473-45815473 on Build GRCh38
rs756556863 Chr.20: 45815524-45815524 on Build GRCh38
rs764954619 Chr.20: 45815478-45815478 on Build GRCh38
rs997900527 Chr.20: 45815627-45815627 on Build GRCh38
rs577176266 Chr.20: 45815325-45815325 on Build GRCh38
rs757509893 Chr.20: 45815389-45815389 on Build GRCh38
rs761667583 Chr.20: 45815560-45815560 on Build GRCh38
rs746495033 Chr.20: 45815546-45815546 on Build GRCh38
rs781652276 Chr.20: 45815431-45815431 on Build GRCh38
rs779653538 Chr.20: 45815541-45815541 on Build GRCh38
rs780709357 Chr.20: 45815610-45815610 on Build GRCh38
rs535990120 Chr.20: 45815525-45815525 on Build GRCh38
rs77269727 Chr.20: 45815434-45815434 on Build GRCh38
rs768631028 Chr.20: 45815404-45815405 on Build GRCh38
rs763209065 Chr.20: 45815569-45815569 on Build GRCh38
rs769065843 Chr.20: 45815348-45815348 on Build GRCh38
rs1015248369 Chr.20: 45815331-45815331 on Build GRCh38
rs574879843 Chr.20: 45815365-45815365 on Build GRCh38
rs976266522 Chr.20: 45815614-45815614 on Build GRCh38
rs770704295 Chr.20: 45815641-45815641 on Build GRCh38
rs763119054 Chr.20: 45815483-45815483 on Build GRCh38
rs764691584 Chr.20: 45815686-45815686 on Build GRCh38
rs759551578 Chr.20: 45815580-45815580 on Build GRCh38
rs775693708 Chr.20: 45815667-45815667 on Build GRCh38
rs767082146 Chr.20: 45815378-45815378 on Build GRCh38
rs766500004 Chr.20: 45815485-45815485 on Build GRCh38
rs1027992765 Chr.20: 45815636-45815636 on Build GRCh38
rs780492032 Chr.20: 45815361-45815365 on Build GRCh38
rs572679134 Chr.20: 45815251-45815251 on Build GRCh38
rs749787205 Chr.20: 45815371-45815371 on Build GRCh38
rs377514203 Chr.20: 45815442-45815442 on Build GRCh38
rs772939191 Chr.20: 45815564-45815564 on Build GRCh38
rs750643445 Chr.20: 45815481-45815481 on Build GRCh38
rs751202605 Chr.20: 45815699-45815699 on Build GRCh38
rs778984580 Chr.20: 45815415-45815415 on Build GRCh38
rs200696786 Chr.20: 45815393-45815393 on Build GRCh38
rs200049340 Chr.20: 45815403-45815403 on Build GRCh38
rs1033161095 Chr.20: 45815688-45815688 on Build GRCh38
rs552432611 Chr.20: 45815383-45815383 on Build GRCh38
rs141555061 Chr.20: 45815600-45815600 on Build GRCh38
rs772368153 Chr.20: 45815625-45815637 on Build GRCh38
rs533855132 Chr.20: 45815315-45815315 on Build GRCh38
rs769693911 Chr.20: 45815615-45815615 on Build GRCh38
rs748408670 Chr.20: 45815471-45815472 on Build GRCh38
rs11697338 Chr.20: 45815382-45815382 on Build GRCh38
rs146143353 Chr.20: 45815642-45815642 on Build GRCh38
rs753288094 Chr.20: 45815583-45815583 on Build GRCh38
rs774815934 Chr.20: 45815438-45815438 on Build GRCh38
rs768444997 Chr.20: 45815452-45815452 on Build GRCh38
rs932190109 Chr.20: 45815632-45815632 on Build GRCh38
rs1004318524 Chr.20: 45815277-45815277 on Build GRCh38
rs767520518 Chr.20: 45815656-45815656 on Build GRCh38
rs1028463933 Chr.20: 45815427-45815427 on Build GRCh38
rs757572822 Chr.20: 45815698-45815698 on Build GRCh38
rs371616231 Chr.20: 45815581-45815581 on Build GRCh38
rs201934862 Chr.20: 45815489-45815489 on Build GRCh38
rs558394302 Chr.20: 45815324-45815324 on Build GRCh38
rs749725543 Chr.20: 45815494-45815494 on Build GRCh38
rs749071967 Chr.20: 45815639-45815639 on Build GRCh38
rs773029038 Chr.20: 45815623-45815623 on Build GRCh38

View Full Product Details