Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45943006-45943506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCIF1
Entrez Gene ID
63935
Gene Name
PDX1 C-terminal inhibiting factor 1
Gene Aliases
C20orf67, PPP1R121
UniGene
Hs.472856
Gene Chromosome Location
Chr.20: 45934674-45948023 on Build GRCh38
Transcripts
NM_022104.3 XM_011528980.2 XM_011528981.2 XM_017028013.1 XM_017028014.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6870434 c.840C>T p.F280F Chr.20: 45943358-45943358 on Build GRCh38
262830 c.678T>C p.I226I Chr.20: 45943101-45943101 on Build GRCh38
3693539 c.882C>T p.A294A Chr.20: 45943400-45943400 on Build GRCh38
5392212 c.881C>T p.A294V Chr.20: 45943399-45943399 on Build GRCh38
4712702 c.856C>T p.R286C Chr.20: 45943374-45943374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906040948 Chr.20: 45943376-45943376 on Build GRCh38
rs761766199 Chr.20: 45943438-45943438 on Build GRCh38
rs763325965 Chr.20: 45943079-45943079 on Build GRCh38
rs148464921 Chr.20: 45943230-45943230 on Build GRCh38
rs918989854 Chr.20: 45943161-45943161 on Build GRCh38
rs35751664 Chr.20: 45943209-45943210 on Build GRCh38
rs751214275 Chr.20: 45943451-45943451 on Build GRCh38
rs201864490 Chr.20: 45943078-45943078 on Build GRCh38
rs776327636 Chr.20: 45943429-45943429 on Build GRCh38
rs779190135 Chr.20: 45943400-45943400 on Build GRCh38
rs565847900 Chr.20: 45943191-45943191 on Build GRCh38
rs561638877 Chr.20: 45943068-45943068 on Build GRCh38
rs771209657 Chr.20: 45943394-45943394 on Build GRCh38
rs772655871 Chr.20: 45943443-45943443 on Build GRCh38
rs768473040 Chr.20: 45943418-45943418 on Build GRCh38
rs746955388 Chr.20: 45943287-45943287 on Build GRCh38
rs1002669502 Chr.20: 45943037-45943037 on Build GRCh38
rs756785423 Chr.20: 45943200-45943200 on Build GRCh38
rs769086558 Chr.20: 45943070-45943071 on Build GRCh38
rs749402768 Chr.20: 45943393-45943393 on Build GRCh38
rs1044868645 Chr.20: 45943349-45943349 on Build GRCh38
rs760479692 Chr.20: 45943173-45943173 on Build GRCh38
rs961632704 Chr.20: 45943445-45943445 on Build GRCh38
rs75941048 Chr.20: 45943255-45943255 on Build GRCh38
rs774920023 Chr.20: 45943076-45943077 on Build GRCh38
rs759162574 Chr.20: 45943469-45943469 on Build GRCh38
rs754471454 Chr.20: 45943385-45943385 on Build GRCh38
rs369816948 Chr.20: 45943055-45943055 on Build GRCh38
rs148089002 Chr.20: 45943388-45943388 on Build GRCh38
rs1056962005 Chr.20: 45943478-45943478 on Build GRCh38
rs751808254 Chr.20: 45943364-45943364 on Build GRCh38
rs368133509 Chr.20: 45943067-45943067 on Build GRCh38
rs372823087 Chr.20: 45943318-45943318 on Build GRCh38
rs868647350 Chr.20: 45943238-45943238 on Build GRCh38
rs750052866 Chr.20: 45943216-45943216 on Build GRCh38
rs144543839 Chr.20: 45943215-45943215 on Build GRCh38
rs762540613 Chr.20: 45943350-45943350 on Build GRCh38
rs531254927 Chr.20: 45943032-45943032 on Build GRCh38
rs775466956 Chr.20: 45943135-45943135 on Build GRCh38
rs945333123 Chr.20: 45943336-45943336 on Build GRCh38
rs758265911 Chr.20: 45943248-45943248 on Build GRCh38
rs761703253 Chr.20: 45943335-45943335 on Build GRCh38
rs747426641 Chr.20: 45943253-45943253 on Build GRCh38
rs781470289 Chr.20: 45943382-45943382 on Build GRCh38
rs1036021789 Chr.20: 45943039-45943039 on Build GRCh38
rs778049244 Chr.20: 45943277-45943277 on Build GRCh38
rs748276366 Chr.20: 45943075-45943075 on Build GRCh38
rs748671998 Chr.20: 45943256-45943256 on Build GRCh38
rs769982215 Chr.20: 45943271-45943271 on Build GRCh38
rs766038014 Chr.20: 45943361-45943361 on Build GRCh38
rs765943938 Chr.20: 45943448-45943448 on Build GRCh38
rs762458890 Chr.20: 45943447-45943447 on Build GRCh38
rs764692730 Chr.20: 45943345-45943345 on Build GRCh38
rs755385891 Chr.20: 45943254-45943254 on Build GRCh38
rs769740499 Chr.20: 45943439-45943439 on Build GRCh38
rs56007236 Chr.20: 45943184-45943184 on Build GRCh38
rs375365216 Chr.20: 45943093-45943093 on Build GRCh38
rs1015863296 Chr.20: 45943073-45943073 on Build GRCh38
rs1025975003 Chr.20: 45943471-45943471 on Build GRCh38
rs994358405 Chr.20: 45943465-45943465 on Build GRCh38
rs755368530 Chr.20: 45943375-45943375 on Build GRCh38
rs375505997 Chr.20: 45943323-45943323 on Build GRCh38
rs943600890 Chr.20: 45943312-45943312 on Build GRCh38
rs771309690 Chr.20: 45943289-45943289 on Build GRCh38
rs746131827 Chr.20: 45943407-45943407 on Build GRCh38
rs376609010 Chr.20: 45943348-45943348 on Build GRCh38
rs764104695 Chr.20: 45943197-45943197 on Build GRCh38
rs776616651 Chr.20: 45943326-45943326 on Build GRCh38
rs777960495 Chr.20: 45943391-45943391 on Build GRCh38
rs780442240 Chr.20: 45943049-45943049 on Build GRCh38
rs761852756 Chr.20: 45943253-45943253 on Build GRCh38

View Full Product Details