Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 49505827-49506366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTGIS
Entrez Gene ID
5740
Gene Name
prostaglandin I2 (prostacyclin) synthase
Gene Aliases
CYP8, CYP8A1, PGIS, PTGI
UniGene
Hs.302085
Gene Chromosome Location
Chr.20: 49503874-49568170 on Build GRCh38
Transcripts
NM_000961.3

SNP Detail

SNP ID Chromosome Location
rs5598 Chr.20: 49506317-49506317 on Build GRCh38
rs6095552 Chr.20: 49505991-49505991 on Build GRCh38
rs144433871 Chr.20: 49506181-49506181 on Build GRCh38
rs761432064 Chr.20: 49506208-49506208 on Build GRCh38
rs141655556 Chr.20: 49505877-49505877 on Build GRCh38
rs970398750 Chr.20: 49505870-49505870 on Build GRCh38
rs1024651108 Chr.20: 49505865-49505865 on Build GRCh38
rs780602636 Chr.20: 49506109-49506109 on Build GRCh38
rs373895388 Chr.20: 49506087-49506087 on Build GRCh38
rs1055963130 Chr.20: 49506153-49506153 on Build GRCh38
rs934509653 Chr.20: 49505851-49505851 on Build GRCh38
rs571620614 Chr.20: 49506041-49506041 on Build GRCh38
rs866676593 Chr.20: 49505972-49505972 on Build GRCh38
rs973207520 Chr.20: 49506001-49506001 on Build GRCh38
rs147989372 Chr.20: 49506229-49506229 on Build GRCh38
rs924513689 Chr.20: 49506168-49506168 on Build GRCh38
rs938310659 Chr.20: 49506264-49506264 on Build GRCh38
rs1012023278 Chr.20: 49506219-49506223 on Build GRCh38
rs758390054 Chr.20: 49506289-49506289 on Build GRCh38
rs1047974229 Chr.20: 49506265-49506265 on Build GRCh38
rs774873402 Chr.20: 49505940-49505940 on Build GRCh38
rs1037898759 Chr.20: 49506299-49506299 on Build GRCh38
rs1030367317 Chr.20: 49506130-49506130 on Build GRCh38
rs886664854 Chr.20: 49506273-49506273 on Build GRCh38
rs569876620 Chr.20: 49506128-49506128 on Build GRCh38
rs904754116 Chr.20: 49506068-49506068 on Build GRCh38
rs911898232 Chr.20: 49506039-49506039 on Build GRCh38
rs922055176 Chr.20: 49506013-49506013 on Build GRCh38
rs5599 Chr.20: 49506301-49506301 on Build GRCh38
rs6095553 Chr.20: 49506309-49506309 on Build GRCh38
rs886606978 Chr.20: 49506045-49506045 on Build GRCh38
rs986757487 Chr.20: 49506163-49506163 on Build GRCh38
rs1006353847 Chr.20: 49506102-49506102 on Build GRCh38
rs763683858 Chr.20: 49506119-49506119 on Build GRCh38
rs551595389 Chr.20: 49506113-49506113 on Build GRCh38
rs568330035 Chr.20: 49506231-49506231 on Build GRCh38
rs5600 Chr.20: 49506225-49506225 on Build GRCh38
rs765056205 Chr.20: 49506214-49506214 on Build GRCh38
rs565960167 Chr.20: 49506035-49506035 on Build GRCh38
rs1044062942 Chr.20: 49505987-49505987 on Build GRCh38
rs753479496 Chr.20: 49506186-49506186 on Build GRCh38
rs1054616719 Chr.20: 49505931-49505931 on Build GRCh38
rs956075539 Chr.20: 49506180-49506180 on Build GRCh38
rs954688164 Chr.20: 49506155-49506155 on Build GRCh38
rs932075400 Chr.20: 49506016-49506016 on Build GRCh38
rs983788293 Chr.20: 49505920-49505920 on Build GRCh38
rs1000402932 Chr.20: 49506096-49506096 on Build GRCh38
rs907772994 Chr.20: 49505933-49505933 on Build GRCh38

View Full Product Details