Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 54475722-54476239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOK5
Entrez Gene ID
55816
Gene Name
docking protein 5
Gene Aliases
C20orf180, IRS-6, IRS6
UniGene
Hs.473133
Gene Chromosome Location
Chr.20: 54475472-54651171 on Build GRCh38
Transcripts
NM_001294161.1 NM_018431.4 NM_177959.2 XM_011528904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4634549 c.4G>A p.A2T Chr.20: 54475950-54475950 on Build GRCh38
5530137 c.36G>A p.G12G Chr.20: 54475982-54475982 on Build GRCh38
4998469 c.64G>C p.G22R Chr.20: 54476010-54476010 on Build GRCh38
6206924 c.50G>A p.R17Q Chr.20: 54475996-54475996 on Build GRCh38
3740188 c.58C>T p.R20C Chr.20: 54476004-54476004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019316988 Chr.20: 54476166-54476166 on Build GRCh38
rs751370091 Chr.20: 54476046-54476046 on Build GRCh38
rs1049449845 Chr.20: 54475741-54475741 on Build GRCh38
rs966830454 Chr.20: 54475772-54475772 on Build GRCh38
rs1040729769 Chr.20: 54476197-54476197 on Build GRCh38
rs965183462 Chr.20: 54475996-54475996 on Build GRCh38
rs1046889130 Chr.20: 54475787-54475787 on Build GRCh38
rs767064202 Chr.20: 54476023-54476023 on Build GRCh38
rs943930409 Chr.20: 54476188-54476188 on Build GRCh38
rs966992103 Chr.20: 54475930-54475930 on Build GRCh38
rs762898036 Chr.20: 54476038-54476038 on Build GRCh38
rs755738005 Chr.20: 54476062-54476062 on Build GRCh38
rs780952483 Chr.20: 54476051-54476051 on Build GRCh38
rs1011045775 Chr.20: 54476125-54476125 on Build GRCh38
rs957632921 Chr.20: 54475823-54475823 on Build GRCh38
rs539917701 Chr.20: 54476074-54476074 on Build GRCh38
rs999654497 Chr.20: 54476073-54476073 on Build GRCh38
rs765217379 Chr.20: 54475920-54475920 on Build GRCh38
rs1028079277 Chr.20: 54476214-54476214 on Build GRCh38
rs1008435348 Chr.20: 54475766-54475766 on Build GRCh38
rs998910267 Chr.20: 54476160-54476160 on Build GRCh38
rs753099485 Chr.20: 54476057-54476058 on Build GRCh38
rs190769826 Chr.20: 54476127-54476127 on Build GRCh38
rs142360201 Chr.20: 54475955-54475955 on Build GRCh38
rs531171846 Chr.20: 54475816-54475816 on Build GRCh38
rs765127484 Chr.20: 54476031-54476031 on Build GRCh38
rs145031079 Chr.20: 54475976-54475976 on Build GRCh38
rs760811282 Chr.20: 54476019-54476019 on Build GRCh38
rs574669896 Chr.20: 54476205-54476205 on Build GRCh38
rs1033890051 Chr.20: 54475909-54475909 on Build GRCh38
rs1036070075 Chr.20: 54476130-54476130 on Build GRCh38
rs1046416341 Chr.20: 54476096-54476096 on Build GRCh38
rs778347873 Chr.20: 54475790-54475803 on Build GRCh38
rs758504848 Chr.20: 54476147-54476147 on Build GRCh38
rs778964928 Chr.20: 54476119-54476119 on Build GRCh38
rs970674505 Chr.20: 54476109-54476109 on Build GRCh38
rs1036877826 Chr.20: 54475927-54475927 on Build GRCh38
rs759132965 Chr.20: 54475924-54475925 on Build GRCh38
rs752731986 Chr.20: 54476120-54476120 on Build GRCh38
rs753494617 Chr.20: 54475924-54475924 on Build GRCh38
rs1031778241 Chr.20: 54475802-54475802 on Build GRCh38
rs902279549 Chr.20: 54476072-54476072 on Build GRCh38
rs754688929 Chr.20: 54475943-54475943 on Build GRCh38
rs766120081 Chr.20: 54476043-54476043 on Build GRCh38
rs180784893 Chr.20: 54476142-54476142 on Build GRCh38
rs887073693 Chr.20: 54475770-54475770 on Build GRCh38
rs1049667645 Chr.20: 54476050-54476050 on Build GRCh38
rs761599246 Chr.20: 54475914-54475914 on Build GRCh38
rs200877470 Chr.20: 54475968-54475968 on Build GRCh38
rs770408047 Chr.20: 54475969-54475969 on Build GRCh38
rs958274172 Chr.20: 54475911-54475911 on Build GRCh38
rs750409167 Chr.20: 54476034-54476034 on Build GRCh38
rs778468129 Chr.20: 54475975-54475975 on Build GRCh38
rs750215059 Chr.20: 54475924-54475924 on Build GRCh38
rs775419478 Chr.20: 54476015-54476015 on Build GRCh38
rs555486668 Chr.20: 54476195-54476195 on Build GRCh38
rs752140141 Chr.20: 54476053-54476053 on Build GRCh38
rs983999592 Chr.20: 54476061-54476061 on Build GRCh38
rs754526397 Chr.20: 54476047-54476047 on Build GRCh38
rs200424144 Chr.20: 54475925-54475925 on Build GRCh38
rs965495206 Chr.20: 54476179-54476179 on Build GRCh38
rs547896444 Chr.20: 54475856-54475856 on Build GRCh38
rs772353772 Chr.20: 54476001-54476001 on Build GRCh38
rs73274916 Chr.20: 54475749-54475749 on Build GRCh38
rs113393523 Chr.20: 54476069-54476069 on Build GRCh38
rs938142817 Chr.20: 54476113-54476113 on Build GRCh38
rs759610972 Chr.20: 54476000-54476000 on Build GRCh38
rs751121183 Chr.20: 54475932-54475932 on Build GRCh38
rs765618763 Chr.20: 54475925-54475926 on Build GRCh38
rs1021163404 Chr.20: 54475928-54475928 on Build GRCh38
rs556661066 Chr.20: 54476098-54476098 on Build GRCh38
rs535795564 Chr.20: 54476149-54476149 on Build GRCh38
rs780498456 Chr.20: 54475952-54475952 on Build GRCh38
rs770546102 Chr.20: 54475844-54475844 on Build GRCh38
rs73141289 Chr.20: 54476056-54476056 on Build GRCh38
rs1011817463 Chr.20: 54476102-54476102 on Build GRCh38
rs911146753 Chr.20: 54476180-54476180 on Build GRCh38
rs940003318 Chr.20: 54476093-54476093 on Build GRCh38
rs398035965 Chr.20: 54475930-54475931 on Build GRCh38
rs71196453 Chr.20: 54475923-54475924 on Build GRCh38
rs376779165 Chr.20: 54475904-54475904 on Build GRCh38
rs1023180618 Chr.20: 54476105-54476105 on Build GRCh38
rs138338080 Chr.20: 54475997-54475997 on Build GRCh38
rs1004219271 Chr.20: 54475778-54475778 on Build GRCh38

View Full Product Details