Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62214879-62215379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HRH3
Entrez Gene ID
11255
Gene Name
histamine receptor H3
Gene Aliases
GPCR97, HH3R
UniGene
Hs.251399
Gene Chromosome Location
Chr.20: 62213845-62220267 on Build GRCh38
Transcripts
NM_007232.2 XM_005260266.3 XM_017027623.1

SNP Detail

SNP ID Chromosome Location
rs974774733 Chr.20: 62215013-62215013 on Build GRCh38
rs1045003725 Chr.20: 62214971-62214971 on Build GRCh38
rs200479618 Chr.20: 62215287-62215287 on Build GRCh38
rs978861769 Chr.20: 62215045-62215045 on Build GRCh38
rs1013702127 Chr.20: 62214965-62214965 on Build GRCh38
rs1056374857 Chr.20: 62215146-62215146 on Build GRCh38
rs200421002 Chr.20: 62215270-62215270 on Build GRCh38
rs907712608 Chr.20: 62215065-62215065 on Build GRCh38
rs980104970 Chr.20: 62214999-62214999 on Build GRCh38
rs201127894 Chr.20: 62215130-62215130 on Build GRCh38
rs1033015957 Chr.20: 62215253-62215253 on Build GRCh38
rs572686226 Chr.20: 62215116-62215116 on Build GRCh38
rs1020297840 Chr.20: 62215023-62215023 on Build GRCh38
rs201986523 Chr.20: 62214979-62214979 on Build GRCh38
rs923347082 Chr.20: 62215035-62215035 on Build GRCh38
rs199654871 Chr.20: 62215296-62215296 on Build GRCh38
rs911576104 Chr.20: 62215057-62215057 on Build GRCh38
rs200855618 Chr.20: 62215218-62215218 on Build GRCh38
rs3746654 Chr.20: 62215106-62215106 on Build GRCh38
rs201798277 Chr.20: 62215236-62215236 on Build GRCh38
rs538055280 Chr.20: 62214942-62214942 on Build GRCh38
rs1010767902 Chr.20: 62215018-62215018 on Build GRCh38
rs200034469 Chr.20: 62215114-62215114 on Build GRCh38
rs201903975 Chr.20: 62215131-62215131 on Build GRCh38
rs968876537 Chr.20: 62215043-62215043 on Build GRCh38
rs756649793 Chr.20: 62215342-62215342 on Build GRCh38
rs1009971929 Chr.20: 62215177-62215177 on Build GRCh38
rs933414549 Chr.20: 62215048-62215048 on Build GRCh38
rs200366085 Chr.20: 62215241-62215241 on Build GRCh38
rs930469345 Chr.20: 62215102-62215102 on Build GRCh38
rs539882207 Chr.20: 62215055-62215055 on Build GRCh38
rs200973313 Chr.20: 62214945-62214945 on Build GRCh38
rs199682301 Chr.20: 62215034-62215034 on Build GRCh38
rs199788934 Chr.20: 62215285-62215285 on Build GRCh38
rs200273161 Chr.20: 62214984-62214984 on Build GRCh38
rs201783541 Chr.20: 62215077-62215077 on Build GRCh38
rs200806823 Chr.20: 62215040-62215040 on Build GRCh38
rs200508884 Chr.20: 62214964-62214964 on Build GRCh38
rs967299829 Chr.20: 62215223-62215223 on Build GRCh38
rs959813889 Chr.20: 62214949-62214949 on Build GRCh38
rs970488418 Chr.20: 62215198-62215198 on Build GRCh38
rs573965346 Chr.20: 62215337-62215337 on Build GRCh38
rs201909183 Chr.20: 62214925-62214925 on Build GRCh38
rs544403090 Chr.20: 62215339-62215339 on Build GRCh38
rs200092835 Chr.20: 62215080-62215080 on Build GRCh38
rs1037343724 Chr.20: 62214921-62214921 on Build GRCh38
rs202063736 Chr.20: 62215316-62215316 on Build GRCh38
rs202205028 Chr.20: 62214953-62214953 on Build GRCh38
rs966973623 Chr.20: 62214998-62214998 on Build GRCh38
rs1056765929 Chr.20: 62215135-62215135 on Build GRCh38
rs200208833 Chr.20: 62215014-62215014 on Build GRCh38
rs968740744 Chr.20: 62215222-62215222 on Build GRCh38
rs1010170221 Chr.20: 62215167-62215167 on Build GRCh38
rs201371445 Chr.20: 62214971-62214971 on Build GRCh38
rs201989508 Chr.20: 62215269-62215269 on Build GRCh38
rs895820429 Chr.20: 62215161-62215161 on Build GRCh38
rs201772541 Chr.20: 62215265-62215265 on Build GRCh38
rs905074339 Chr.20: 62214982-62214982 on Build GRCh38
rs974722398 Chr.20: 62215262-62215262 on Build GRCh38
rs866735441 Chr.20: 62215331-62215331 on Build GRCh38
rs1021094465 Chr.20: 62215195-62215195 on Build GRCh38
rs201643814 Chr.20: 62215089-62215089 on Build GRCh38
rs199828759 Chr.20: 62215001-62215001 on Build GRCh38
rs754825748 Chr.20: 62215344-62215344 on Build GRCh38
rs905500542 Chr.20: 62215090-62215090 on Build GRCh38
rs200809187 Chr.20: 62215308-62215308 on Build GRCh38
rs199553034 Chr.20: 62215047-62215047 on Build GRCh38
rs182141144 Chr.20: 62215049-62215049 on Build GRCh38
rs201316619 Chr.20: 62215295-62215295 on Build GRCh38
rs964997650 Chr.20: 62215258-62215258 on Build GRCh38
rs202091322 Chr.20: 62215042-62215042 on Build GRCh38
rs923802269 Chr.20: 62215291-62215291 on Build GRCh38
rs199903482 Chr.20: 62215140-62215140 on Build GRCh38
rs199862798 Chr.20: 62214944-62214944 on Build GRCh38
rs941932498 Chr.20: 62215073-62215073 on Build GRCh38
rs201468811 Chr.20: 62215281-62215281 on Build GRCh38
rs1739585 Chr.20: 62215000-62215000 on Build GRCh38
rs201388416 Chr.20: 62215027-62215027 on Build GRCh38
rs201042804 Chr.20: 62215088-62215088 on Build GRCh38
rs1001202932 Chr.20: 62214926-62214926 on Build GRCh38

View Full Product Details