Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62390118-62390613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABLES2
Entrez Gene ID
81928
Gene Name
Cdk5 and Abl enzyme substrate 2
Gene Aliases
C20orf150, dJ908M14.2, ik3-2
UniGene
Hs.301040
Gene Chromosome Location
Chr.20: 62388630-62407283 on Build GRCh38
Transcripts
NM_031215.2

SNP Detail

SNP ID Chromosome Location
rs915597902 Chr.20: 62390322-62390322 on Build GRCh38
rs550853606 Chr.20: 62390320-62390320 on Build GRCh38
rs987980703 Chr.20: 62390256-62390256 on Build GRCh38
rs936490030 Chr.20: 62390246-62390246 on Build GRCh38
rs544988596 Chr.20: 62390585-62390585 on Build GRCh38
rs762096190 Chr.20: 62390481-62390481 on Build GRCh38
rs1031108209 Chr.20: 62390184-62390184 on Build GRCh38
rs137987461 Chr.20: 62390583-62390583 on Build GRCh38
rs920423237 Chr.20: 62390308-62390308 on Build GRCh38
rs970593704 Chr.20: 62390169-62390169 on Build GRCh38
rs878915183 Chr.20: 62390216-62390216 on Build GRCh38
rs569219729 Chr.20: 62390344-62390344 on Build GRCh38
rs1052583709 Chr.20: 62390457-62390457 on Build GRCh38
rs770071625 Chr.20: 62390474-62390474 on Build GRCh38
rs537683335 Chr.20: 62390551-62390551 on Build GRCh38
rs542394948 Chr.20: 62390179-62390179 on Build GRCh38
rs562287008 Chr.20: 62390235-62390235 on Build GRCh38
rs879581393 Chr.20: 62390255-62390255 on Build GRCh38
rs370483514 Chr.20: 62390250-62390251 on Build GRCh38
rs943503560 Chr.20: 62390275-62390275 on Build GRCh38
rs951834373 Chr.20: 62390593-62390593 on Build GRCh38
rs902954305 Chr.20: 62390442-62390442 on Build GRCh38
rs1006796815 Chr.20: 62390419-62390419 on Build GRCh38
rs1011104429 Chr.20: 62390199-62390199 on Build GRCh38
rs138649352 Chr.20: 62390395-62390395 on Build GRCh38
rs911954348 Chr.20: 62390276-62390276 on Build GRCh38
rs11697527 Chr.20: 62390530-62390530 on Build GRCh38
rs1045797060 Chr.20: 62390417-62390417 on Build GRCh38
rs971914969 Chr.20: 62390286-62390286 on Build GRCh38
rs970394190 Chr.20: 62390523-62390523 on Build GRCh38
rs972135970 Chr.20: 62390589-62390589 on Build GRCh38
rs150310978 Chr.20: 62390570-62390570 on Build GRCh38
rs369099076 Chr.20: 62390588-62390588 on Build GRCh38
rs1024654178 Chr.20: 62390144-62390144 on Build GRCh38
rs1027498098 Chr.20: 62390592-62390592 on Build GRCh38
rs141174640 Chr.20: 62390456-62390456 on Build GRCh38
rs374972236 Chr.20: 62390408-62390408 on Build GRCh38
rs953368294 Chr.20: 62390243-62390243 on Build GRCh38
rs930582249 Chr.20: 62390309-62390309 on Build GRCh38
rs1008930634 Chr.20: 62390574-62390574 on Build GRCh38
rs879089364 Chr.20: 62390563-62390565 on Build GRCh38
rs923741460 Chr.20: 62390202-62390202 on Build GRCh38
rs529510851 Chr.20: 62390236-62390236 on Build GRCh38
rs368992172 Chr.20: 62390470-62390470 on Build GRCh38
rs376869600 Chr.20: 62390277-62390277 on Build GRCh38
rs1019345843 Chr.20: 62390575-62390575 on Build GRCh38
rs3752318 Chr.20: 62390510-62390510 on Build GRCh38
rs1024621665 Chr.20: 62390485-62390485 on Build GRCh38
rs534285022 Chr.20: 62390485-62390485 on Build GRCh38
rs551383096 Chr.20: 62390449-62390449 on Build GRCh38
rs958319664 Chr.20: 62390193-62390193 on Build GRCh38
rs886715658 Chr.20: 62390356-62390357 on Build GRCh38
rs1028956566 Chr.20: 62390233-62390233 on Build GRCh38
rs978020476 Chr.20: 62390191-62390191 on Build GRCh38

View Full Product Details