Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62473320-62473825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA5
Entrez Gene ID
140628
Gene Name
GATA binding protein 5
Gene Aliases
GATAS, bB379O24.1
UniGene
Hs.352250
Gene Chromosome Location
Chr.20: 62463497-62475970 on Build GRCh38
Transcripts
NM_080473.4 XM_006723699.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4100127 c.629G>A p.C210Y Chr.20: 62473473-62473473 on Build GRCh38
5748198 c.600G>A p.W200* Chr.20: 62473502-62473502 on Build GRCh38
4646643 c.548C>T p.P183L Chr.20: 62473554-62473554 on Build GRCh38
5774328 c.638G>A p.C213Y Chr.20: 62473464-62473464 on Build GRCh38
252516 c.602G>A p.R201H Chr.20: 62473500-62473500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782068460 Chr.20: 62473378-62473378 on Build GRCh38
rs1017442862 Chr.20: 62473432-62473432 on Build GRCh38
rs560458272 Chr.20: 62473561-62473561 on Build GRCh38
rs542925110 Chr.20: 62473509-62473509 on Build GRCh38
rs782462807 Chr.20: 62473554-62473554 on Build GRCh38
rs782748816 Chr.20: 62473395-62473395 on Build GRCh38
rs369626023 Chr.20: 62473370-62473370 on Build GRCh38
rs782554904 Chr.20: 62473574-62473574 on Build GRCh38
rs782776140 Chr.20: 62473394-62473394 on Build GRCh38
rs782339380 Chr.20: 62473775-62473775 on Build GRCh38
rs782281730 Chr.20: 62473758-62473758 on Build GRCh38
rs139428354 Chr.20: 62473424-62473424 on Build GRCh38
rs373453165 Chr.20: 62473371-62473371 on Build GRCh38
rs533015904 Chr.20: 62473532-62473532 on Build GRCh38
rs375617512 Chr.20: 62473446-62473446 on Build GRCh38
rs6061554 Chr.20: 62473420-62473420 on Build GRCh38
rs114309782 Chr.20: 62473408-62473408 on Build GRCh38
rs1039514030 Chr.20: 62473640-62473640 on Build GRCh38
rs147905576 Chr.20: 62473615-62473626 on Build GRCh38
rs781952577 Chr.20: 62473377-62473377 on Build GRCh38
rs375857054 Chr.20: 62473553-62473553 on Build GRCh38
rs781885402 Chr.20: 62473538-62473538 on Build GRCh38
rs782656299 Chr.20: 62473489-62473489 on Build GRCh38
rs199934543 Chr.20: 62473423-62473423 on Build GRCh38
rs377441586 Chr.20: 62473387-62473387 on Build GRCh38
rs781894505 Chr.20: 62473454-62473454 on Build GRCh38
rs567273738 Chr.20: 62473635-62473635 on Build GRCh38
rs539618042 Chr.20: 62473687-62473687 on Build GRCh38
rs782595656 Chr.20: 62473484-62473484 on Build GRCh38
rs782112827 Chr.20: 62473428-62473428 on Build GRCh38
rs144579982 Chr.20: 62473419-62473419 on Build GRCh38
rs782033443 Chr.20: 62473389-62473389 on Build GRCh38
rs782511940 Chr.20: 62473680-62473680 on Build GRCh38
rs116164480 Chr.20: 62473404-62473404 on Build GRCh38
rs782140584 Chr.20: 62473459-62473459 on Build GRCh38
rs901706981 Chr.20: 62473645-62473645 on Build GRCh38
rs782773387 Chr.20: 62473552-62473552 on Build GRCh38
rs886361966 Chr.20: 62473700-62473700 on Build GRCh38
rs782260539 Chr.20: 62473492-62473492 on Build GRCh38
rs527597541 Chr.20: 62473590-62473590 on Build GRCh38
rs566244361 Chr.20: 62473605-62473605 on Build GRCh38
rs997414695 Chr.20: 62473474-62473474 on Build GRCh38
rs782051102 Chr.20: 62473533-62473533 on Build GRCh38
rs980231005 Chr.20: 62473500-62473500 on Build GRCh38
rs782105775 Chr.20: 62473525-62473525 on Build GRCh38
rs782648093 Chr.20: 62473577-62473577 on Build GRCh38
rs144082730 Chr.20: 62473427-62473427 on Build GRCh38
rs41305803 Chr.20: 62473493-62473493 on Build GRCh38
rs753656900 Chr.20: 62473407-62473407 on Build GRCh38
rs116503780 Chr.20: 62473438-62473438 on Build GRCh38
rs782230478 Chr.20: 62473709-62473709 on Build GRCh38
rs782315967 Chr.20: 62473505-62473505 on Build GRCh38
rs782614097 Chr.20: 62473497-62473497 on Build GRCh38
rs370091622 Chr.20: 62473448-62473448 on Build GRCh38
rs757063452 Chr.20: 62473494-62473494 on Build GRCh38
rs945397123 Chr.20: 62473608-62473608 on Build GRCh38
rs566323003 Chr.20: 62473761-62473761 on Build GRCh38
rs782766788 Chr.20: 62473462-62473462 on Build GRCh38
rs537553037 Chr.20: 62473356-62473356 on Build GRCh38
rs943477497 Chr.20: 62473639-62473639 on Build GRCh38
rs782474774 Chr.20: 62473403-62473403 on Build GRCh38
rs1043747827 Chr.20: 62473622-62473622 on Build GRCh38
rs1026176736 Chr.20: 62473477-62473477 on Build GRCh38
rs914148035 Chr.20: 62473583-62473583 on Build GRCh38
rs782578916 Chr.20: 62473556-62473556 on Build GRCh38
rs782037364 Chr.20: 62473621-62473621 on Build GRCh38
rs782509793 Chr.20: 62473545-62473545 on Build GRCh38
rs781938227 Chr.20: 62473784-62473784 on Build GRCh38
rs569416669 Chr.20: 62473684-62473684 on Build GRCh38
rs537943787 Chr.20: 62473661-62473661 on Build GRCh38
rs782208424 Chr.20: 62473501-62473501 on Build GRCh38
rs549649551 Chr.20: 62473671-62473671 on Build GRCh38
rs782475918 Chr.20: 62473476-62473476 on Build GRCh38
rs782802751 Chr.20: 62473537-62473537 on Build GRCh38
rs782412130 Chr.20: 62473369-62473369 on Build GRCh38
rs370137374 Chr.20: 62473433-62473433 on Build GRCh38
rs201106575 Chr.20: 62473526-62473526 on Build GRCh38
rs782249004 Chr.20: 62473581-62473581 on Build GRCh38
rs781959959 Chr.20: 62473600-62473600 on Build GRCh38
rs781843254 Chr.20: 62473463-62473463 on Build GRCh38
rs200354101 Chr.20: 62473606-62473606 on Build GRCh38
rs765274094 Chr.20: 62473673-62473673 on Build GRCh38
rs782322254 Chr.20: 62473421-62473421 on Build GRCh38
rs547292393 Chr.20: 62473747-62473747 on Build GRCh38
rs374060174 Chr.20: 62473431-62473431 on Build GRCh38
rs781910978 Chr.20: 62473508-62473508 on Build GRCh38
rs782577538 Chr.20: 62473352-62473352 on Build GRCh38
rs368830497 Chr.20: 62473516-62473516 on Build GRCh38
rs181577536 Chr.20: 62473688-62473688 on Build GRCh38
rs781876016 Chr.20: 62473418-62473418 on Build GRCh38
rs782358488 Chr.20: 62473593-62473593 on Build GRCh38
rs373764622 Chr.20: 62473515-62473515 on Build GRCh38
rs113609354 Chr.20: 62473392-62473392 on Build GRCh38
rs970105024 Chr.20: 62473791-62473791 on Build GRCh38

View Full Product Details