Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63558004-63558508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HELZ2
Entrez Gene ID
85441
Gene Name
helicase with zinc finger 2
Gene Aliases
PDIP-1, PRIC285
UniGene
Hs.517180
Gene Chromosome Location
Chr.20: 63558086-63574239 on Build GRCh38
Transcripts
NM_001037335.2 NM_033405.3

SNP Detail

SNP ID Chromosome Location
rs1050352504 Chr.20: 63558173-63558173 on Build GRCh38
rs576940726 Chr.20: 63558377-63558377 on Build GRCh38
rs1009809349 Chr.20: 63558428-63558428 on Build GRCh38
rs191107024 Chr.20: 63558106-63558106 on Build GRCh38
rs961007219 Chr.20: 63558307-63558307 on Build GRCh38
rs564747763 Chr.20: 63558047-63558047 on Build GRCh38
rs866777662 Chr.20: 63558270-63558270 on Build GRCh38
rs764953593 Chr.20: 63558383-63558383 on Build GRCh38
rs538633737 Chr.20: 63558228-63558228 on Build GRCh38
rs541892735 Chr.20: 63558368-63558369 on Build GRCh38
rs1054229594 Chr.20: 63558441-63558441 on Build GRCh38
rs552135820 Chr.20: 63558076-63558076 on Build GRCh38
rs1046470420 Chr.20: 63558044-63558044 on Build GRCh38
rs796519714 Chr.20: 63558451-63558451 on Build GRCh38
rs532108257 Chr.20: 63558056-63558056 on Build GRCh38
rs549300358 Chr.20: 63558145-63558145 on Build GRCh38
rs980974229 Chr.20: 63558031-63558031 on Build GRCh38
rs919691595 Chr.20: 63558219-63558219 on Build GRCh38
rs949574986 Chr.20: 63558237-63558237 on Build GRCh38
rs991426568 Chr.20: 63558229-63558229 on Build GRCh38
rs959703587 Chr.20: 63558035-63558035 on Build GRCh38
rs34009189 Chr.20: 63558301-63558302 on Build GRCh38
rs970896412 Chr.20: 63558389-63558389 on Build GRCh38
rs537956523 Chr.20: 63558200-63558200 on Build GRCh38
rs961036796 Chr.20: 63558217-63558217 on Build GRCh38
rs548106818 Chr.20: 63558201-63558201 on Build GRCh38
rs949354355 Chr.20: 63558043-63558043 on Build GRCh38
rs1055361567 Chr.20: 63558027-63558027 on Build GRCh38
rs892946792 Chr.20: 63558140-63558140 on Build GRCh38
rs78778752 Chr.20: 63558251-63558251 on Build GRCh38
rs1026723283 Chr.20: 63558317-63558317 on Build GRCh38
rs568000121 Chr.20: 63558240-63558240 on Build GRCh38
rs144228742 Chr.20: 63558088-63558088 on Build GRCh38
rs536242110 Chr.20: 63558296-63558296 on Build GRCh38
rs567807582 Chr.20: 63558185-63558185 on Build GRCh38
rs990585157 Chr.20: 63558322-63558322 on Build GRCh38
rs1008065553 Chr.20: 63558297-63558297 on Build GRCh38
rs889588762 Chr.20: 63558483-63558483 on Build GRCh38
rs982335124 Chr.20: 63558393-63558393 on Build GRCh38
rs1049139537 Chr.20: 63558371-63558371 on Build GRCh38
rs576012706 Chr.20: 63558318-63558318 on Build GRCh38
rs149545125 Chr.20: 63558026-63558026 on Build GRCh38
rs919249553 Chr.20: 63558346-63558346 on Build GRCh38
rs953996366 Chr.20: 63558163-63558163 on Build GRCh38
rs538323045 Chr.20: 63558399-63558399 on Build GRCh38
rs932021022 Chr.20: 63558455-63558455 on Build GRCh38
rs916714375 Chr.20: 63558329-63558329 on Build GRCh38
rs944711226 Chr.20: 63558189-63558189 on Build GRCh38
rs935333809 Chr.20: 63558432-63558432 on Build GRCh38
rs1056556907 Chr.20: 63558398-63558398 on Build GRCh38
rs1028227261 Chr.20: 63558149-63558149 on Build GRCh38
rs994264748 Chr.20: 63558310-63558310 on Build GRCh38
rs1000230439 Chr.20: 63558447-63558447 on Build GRCh38
rs999574991 Chr.20: 63558061-63558061 on Build GRCh38
rs1011399555 Chr.20: 63558274-63558274 on Build GRCh38
rs1028615359 Chr.20: 63558287-63558287 on Build GRCh38
rs1019931172 Chr.20: 63558266-63558266 on Build GRCh38
rs747060324 Chr.20: 63558256-63558256 on Build GRCh38
rs140760582 Chr.20: 63558301-63558301 on Build GRCh38
rs942922845 Chr.20: 63558397-63558397 on Build GRCh38
rs916829014 Chr.20: 63558236-63558236 on Build GRCh38
rs895413340 Chr.20: 63558034-63558034 on Build GRCh38
rs1050479375 Chr.20: 63558467-63558467 on Build GRCh38
rs372125400 Chr.20: 63558255-63558255 on Build GRCh38
rs143002392 Chr.20: 63558446-63558446 on Build GRCh38
rs182202251 Chr.20: 63558269-63558269 on Build GRCh38
rs979027294 Chr.20: 63558077-63558077 on Build GRCh38
rs925669142 Chr.20: 63558284-63558284 on Build GRCh38
rs896767309 Chr.20: 63558419-63558419 on Build GRCh38
rs186806664 Chr.20: 63558392-63558392 on Build GRCh38
rs571567334 Chr.20: 63558218-63558218 on Build GRCh38
rs376043380 Chr.20: 63558369-63558373 on Build GRCh38
rs930609005 Chr.20: 63558369-63558378 on Build GRCh38
rs534409315 Chr.20: 63558041-63558041 on Build GRCh38

View Full Product Details