Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63657654-63658165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RTEL1
Entrez Gene ID
51750
Gene Name
regulator of telomere elongation helicase 1
Gene Aliases
C20orf41, DKCA4, DKCB5, NHL, PFBMFT3, RTEL
UniGene
Hs.745057
Gene Chromosome Location
Chr.20: 63657810-63696253 on Build GRCh38
Transcripts
NM_001283009.1 NM_001283010.1 NM_016434.3 NM_032957.4 NR_037882.1

Gene Symbol
RTEL1-TNFRSF6B
Entrez Gene ID
100533107
Gene Name
RTEL1-TNFRSF6B readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.745057
Gene Chromosome Location
Chr.20: 63657810-63698698 on Build GRCh38
Transcripts
NM_001283009.1 NM_001283010.1 NM_016434.3 NM_032957.4 NR_037882.1

SNP Detail

SNP ID Chromosome Location
rs971450656 Chr.20: 63657977-63657977 on Build GRCh38
rs375737208 Chr.20: 63657824-63657824 on Build GRCh38
rs978444895 Chr.20: 63657727-63657727 on Build GRCh38
rs967276278 Chr.20: 63657945-63657945 on Build GRCh38
rs760362968 Chr.20: 63657999-63657999 on Build GRCh38
rs774771738 Chr.20: 63657676-63657676 on Build GRCh38
rs545619025 Chr.20: 63657758-63657758 on Build GRCh38
rs750201076 Chr.20: 63657744-63657748 on Build GRCh38
rs545660391 Chr.20: 63657744-63657758 on Build GRCh38
rs942447652 Chr.20: 63657760-63657760 on Build GRCh38
rs779251089 Chr.20: 63657752-63657753 on Build GRCh38
rs941392873 Chr.20: 63657809-63657809 on Build GRCh38
rs1039433824 Chr.20: 63657790-63657790 on Build GRCh38
rs908178731 Chr.20: 63658132-63658132 on Build GRCh38
rs905167571 Chr.20: 63657842-63657842 on Build GRCh38
rs1032697732 Chr.20: 63658005-63658005 on Build GRCh38
rs990395292 Chr.20: 63658037-63658037 on Build GRCh38
rs576892822 Chr.20: 63657756-63657757 on Build GRCh38
rs71325460 Chr.20: 63658027-63658027 on Build GRCh38
rs748491781 Chr.20: 63657744-63657745 on Build GRCh38
rs1031660050 Chr.20: 63658027-63658027 on Build GRCh38
rs964840392 Chr.20: 63658066-63658066 on Build GRCh38
rs561745358 Chr.20: 63657791-63657791 on Build GRCh38
rs941025744 Chr.20: 63658138-63658138 on Build GRCh38
rs369602575 Chr.20: 63657705-63657714 on Build GRCh38
rs913116318 Chr.20: 63657689-63657689 on Build GRCh38
rs547215972 Chr.20: 63657858-63657858 on Build GRCh38
rs923473122 Chr.20: 63658100-63658100 on Build GRCh38
rs895656170 Chr.20: 63657900-63657900 on Build GRCh38
rs576937350 Chr.20: 63657752-63657752 on Build GRCh38
rs999194620 Chr.20: 63657779-63657780 on Build GRCh38
rs201287782 Chr.20: 63657743-63657744 on Build GRCh38
rs564437490 Chr.20: 63657827-63657827 on Build GRCh38
rs910797687 Chr.20: 63658039-63658039 on Build GRCh38
rs530838324 Chr.20: 63657802-63657802 on Build GRCh38
rs977383630 Chr.20: 63657692-63657692 on Build GRCh38
rs982899343 Chr.20: 63658141-63658141 on Build GRCh38
rs1014095527 Chr.20: 63657920-63657920 on Build GRCh38
rs976241369 Chr.20: 63658070-63658070 on Build GRCh38
rs578042579 Chr.20: 63657816-63657816 on Build GRCh38
rs925723956 Chr.20: 63657745-63657745 on Build GRCh38
rs1024344484 Chr.20: 63657678-63657678 on Build GRCh38
rs533540884 Chr.20: 63657846-63657846 on Build GRCh38
rs572811871 Chr.20: 63657782-63657782 on Build GRCh38
rs773162378 Chr.20: 63657921-63657921 on Build GRCh38
rs146700551 Chr.20: 63657776-63657776 on Build GRCh38
rs922353452 Chr.20: 63658106-63658106 on Build GRCh38
rs934193838 Chr.20: 63658120-63658120 on Build GRCh38
rs1002303332 Chr.20: 63657857-63657857 on Build GRCh38
rs922781575 Chr.20: 63657779-63657779 on Build GRCh38
rs760392520 Chr.20: 63657714-63657715 on Build GRCh38
rs556816546 Chr.20: 63657713-63657713 on Build GRCh38
rs529601155 Chr.20: 63658043-63658043 on Build GRCh38
rs1035483978 Chr.20: 63657888-63657888 on Build GRCh38
rs978858999 Chr.20: 63657954-63657954 on Build GRCh38
rs75094645 Chr.20: 63657784-63657784 on Build GRCh38

View Full Product Details