Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 14372463-14372976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA13
Entrez Gene ID
6782
Gene Name
heat shock protein family A (Hsp70) member 13
Gene Aliases
STCH
UniGene
Hs.352341
Gene Chromosome Location
Chr.21: 14371115-14383188 on Build GRCh38
Transcripts
NM_006948.4

SNP Detail

SNP ID Chromosome Location
rs773095260 Chr.21: 14372663-14372663 on Build GRCh38
rs976406438 Chr.21: 14372905-14372905 on Build GRCh38
rs1028864404 Chr.21: 14372669-14372669 on Build GRCh38
rs943805789 Chr.21: 14372542-14372542 on Build GRCh38
rs868448306 Chr.21: 14372664-14372664 on Build GRCh38
rs1002993100 Chr.21: 14372511-14372511 on Build GRCh38
rs75289380 Chr.21: 14372748-14372748 on Build GRCh38
rs896769802 Chr.21: 14372551-14372551 on Build GRCh38
rs939961365 Chr.21: 14372755-14372755 on Build GRCh38
rs561083596 Chr.21: 14372854-14372854 on Build GRCh38
rs969056251 Chr.21: 14372800-14372800 on Build GRCh38
rs62229676 Chr.21: 14372875-14372875 on Build GRCh38
rs1040824619 Chr.21: 14372548-14372548 on Build GRCh38
rs574286271 Chr.21: 14372657-14372657 on Build GRCh38
rs1001474415 Chr.21: 14372940-14372940 on Build GRCh38
rs536901868 Chr.21: 14372690-14372690 on Build GRCh38
rs929670774 Chr.21: 14372934-14372934 on Build GRCh38
rs572646609 Chr.21: 14372569-14372569 on Build GRCh38
rs1035771413 Chr.21: 14372725-14372725 on Build GRCh38
rs896710795 Chr.21: 14372871-14372871 on Build GRCh38
rs888030074 Chr.21: 14372643-14372643 on Build GRCh38
rs968503393 Chr.21: 14372597-14372597 on Build GRCh38
rs1006440080 Chr.21: 14372656-14372656 on Build GRCh38
rs751081569 Chr.21: 14372766-14372766 on Build GRCh38
rs2822638 Chr.21: 14372785-14372785 on Build GRCh38
rs12479 Chr.21: 14372948-14372948 on Build GRCh38
rs572392619 Chr.21: 14372802-14372802 on Build GRCh38
rs970060018 Chr.21: 14372675-14372675 on Build GRCh38
rs1021093588 Chr.21: 14372920-14372920 on Build GRCh38
rs988607777 Chr.21: 14372771-14372771 on Build GRCh38
rs912459295 Chr.21: 14372754-14372754 on Build GRCh38
rs972514392 Chr.21: 14372784-14372784 on Build GRCh38
rs931019118 Chr.21: 14372793-14372793 on Build GRCh38
rs541404732 Chr.21: 14372805-14372805 on Build GRCh38
rs2822639 Chr.21: 14372855-14372855 on Build GRCh38
rs986789935 Chr.21: 14372753-14372753 on Build GRCh38
rs187422579 Chr.21: 14372491-14372491 on Build GRCh38
rs7275514 Chr.21: 14372801-14372801 on Build GRCh38
rs896893636 Chr.21: 14372556-14372556 on Build GRCh38
rs1047920304 Chr.21: 14372632-14372632 on Build GRCh38
rs564095747 Chr.21: 14372930-14372930 on Build GRCh38
rs9978693 Chr.21: 14372555-14372555 on Build GRCh38
rs535069843 Chr.21: 14372694-14372694 on Build GRCh38
rs1009831502 Chr.21: 14372903-14372903 on Build GRCh38
rs938347548 Chr.21: 14372835-14372835 on Build GRCh38
rs905531373 Chr.21: 14372508-14372508 on Build GRCh38
rs78283728 Chr.21: 14372538-14372538 on Build GRCh38
rs1047930187 Chr.21: 14372938-14372938 on Build GRCh38

View Full Product Details