Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 21291977-21292507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAM2
Entrez Gene ID
4685
Gene Name
neural cell adhesion molecule 2
Gene Aliases
NCAM21
UniGene
Hs.473450
Gene Chromosome Location
Chr.21: 20998315-21543324 on Build GRCh38
Transcripts
NM_004540.3 XM_011529575.2 XM_011529576.2 XM_011529579.2 XM_011529580.2 XM_011529581.2 XM_011529582.2 XM_011529585.2 XM_017028353.1 XM_017028354.1 XM_017028355.1 XM_017028356.1 XM_017028357.1 XM_017028358.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5770054 c.579C>G p.N193K Chr.21: 21292126-21292126 on Build GRCh38
1029562 c.514C>A p.Q172K Chr.21: 21292136-21292136 on Build GRCh38
5765288 c.694+6C>T p.? Chr.21: 21292247-21292247 on Build GRCh38
478456 c.485G>C p.R162P Chr.21: 21292107-21292107 on Build GRCh38
7097804 c.625T>C p.Y209H Chr.21: 21292172-21292172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990255529 Chr.21: 21292229-21292229 on Build GRCh38
rs762470702 Chr.21: 21292217-21292217 on Build GRCh38
rs772294530 Chr.21: 21292065-21292065 on Build GRCh38
rs8134502 Chr.21: 21292075-21292075 on Build GRCh38
rs138101658 Chr.21: 21292298-21292298 on Build GRCh38
rs750933321 Chr.21: 21292264-21292264 on Build GRCh38
rs769816442 Chr.21: 21292206-21292206 on Build GRCh38
rs181341727 Chr.21: 21292314-21292314 on Build GRCh38
rs386817043 Chr.21: 21292436-21292438 on Build GRCh38
rs751151354 Chr.21: 21292251-21292251 on Build GRCh38
rs781597346 Chr.21: 21292288-21292288 on Build GRCh38
rs759230294 Chr.21: 21292210-21292210 on Build GRCh38
rs759008835 Chr.21: 21292265-21292265 on Build GRCh38
rs746895721 Chr.21: 21292376-21292376 on Build GRCh38
rs8126533 Chr.21: 21292438-21292438 on Build GRCh38
rs558913062 Chr.21: 21292415-21292415 on Build GRCh38
rs566983233 Chr.21: 21292342-21292342 on Build GRCh38
rs898441273 Chr.21: 21292112-21292112 on Build GRCh38
rs768730916 Chr.21: 21292286-21292286 on Build GRCh38
rs751921520 Chr.21: 21292290-21292291 on Build GRCh38
rs745983097 Chr.21: 21292063-21292063 on Build GRCh38
rs7275226 Chr.21: 21292412-21292412 on Build GRCh38
rs942075780 Chr.21: 21292349-21292349 on Build GRCh38
rs761740603 Chr.21: 21292196-21292196 on Build GRCh38
rs765315095 Chr.21: 21292100-21292100 on Build GRCh38
rs779423340 Chr.21: 21292147-21292147 on Build GRCh38
rs749923007 Chr.21: 21292103-21292103 on Build GRCh38
rs766008654 Chr.21: 21292106-21292106 on Build GRCh38
rs145537614 Chr.21: 21292126-21292126 on Build GRCh38
rs907935933 Chr.21: 21292340-21292340 on Build GRCh38
rs898394470 Chr.21: 21292362-21292362 on Build GRCh38
rs747160968 Chr.21: 21292270-21292270 on Build GRCh38
rs767193911 Chr.21: 21292253-21292253 on Build GRCh38
rs60892739 Chr.21: 21292299-21292299 on Build GRCh38
rs760459830 Chr.21: 21292085-21292085 on Build GRCh38
rs966364787 Chr.21: 21292215-21292215 on Build GRCh38
rs1047847690 Chr.21: 21292305-21292305 on Build GRCh38
rs886459366 Chr.21: 21292042-21292042 on Build GRCh38
rs757639076 Chr.21: 21292260-21292260 on Build GRCh38
rs185885193 Chr.21: 21292169-21292169 on Build GRCh38
rs765814157 Chr.21: 21292218-21292218 on Build GRCh38
rs1011221087 Chr.21: 21292436-21292436 on Build GRCh38
rs1037737885 Chr.21: 21292356-21292356 on Build GRCh38
rs757974090 Chr.21: 21292115-21292115 on Build GRCh38
rs371793861 Chr.21: 21292160-21292160 on Build GRCh38
rs76939234 Chr.21: 21292446-21292446 on Build GRCh38
rs757840167 Chr.21: 21292001-21292001 on Build GRCh38
rs376209656 Chr.21: 21292168-21292168 on Build GRCh38
rs1038377851 Chr.21: 21292027-21292027 on Build GRCh38
rs28693888 Chr.21: 21292271-21292271 on Build GRCh38
rs928751187 Chr.21: 21292471-21292471 on Build GRCh38
rs370739586 Chr.21: 21292086-21292086 on Build GRCh38
rs553981033 Chr.21: 21292003-21292003 on Build GRCh38
rs776633904 Chr.21: 21292093-21292093 on Build GRCh38
rs771080079 Chr.21: 21292417-21292417 on Build GRCh38
rs754157429 Chr.21: 21292255-21292255 on Build GRCh38
rs562609911 Chr.21: 21292336-21292336 on Build GRCh38
rs7275731 Chr.21: 21292453-21292453 on Build GRCh38
rs374401078 Chr.21: 21292121-21292121 on Build GRCh38
rs762586053 Chr.21: 21292104-21292104 on Build GRCh38
rs549856274 Chr.21: 21292290-21292290 on Build GRCh38
rs773306072 Chr.21: 21292208-21292208 on Build GRCh38
rs1021739795 Chr.21: 21292437-21292437 on Build GRCh38
rs751188340 Chr.21: 21292107-21292107 on Build GRCh38
rs779401813 Chr.21: 21292123-21292123 on Build GRCh38
rs9978043 Chr.21: 21292433-21292433 on Build GRCh38
rs780979777 Chr.21: 21292301-21292301 on Build GRCh38
rs757727097 Chr.21: 21292132-21292132 on Build GRCh38
rs1007896957 Chr.21: 21292425-21292425 on Build GRCh38
rs745896216 Chr.21: 21292152-21292152 on Build GRCh38
rs750950880 Chr.21: 21292087-21292087 on Build GRCh38
rs776545833 Chr.21: 21292187-21292187 on Build GRCh38
rs997180355 Chr.21: 21292407-21292407 on Build GRCh38
rs191736031 Chr.21: 21292266-21292266 on Build GRCh38

View Full Product Details