Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 26919623-26920157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS5
Entrez Gene ID
11096
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 5
Gene Aliases
ADAM-TS 11, ADAM-TS 5, ADAM-TS5, ADAMTS-11, ADAMTS-5, ADAMTS11, ADMP-2
UniGene
Hs.58324
Gene Chromosome Location
Chr.21: 26917912-26967120 on Build GRCh38
Transcripts
NM_007038.4 XM_011529448.2

SNP Detail

SNP ID Chromosome Location
rs186978268 Chr.21: 26919780-26919780 on Build GRCh38
rs958024849 Chr.21: 26919992-26919992 on Build GRCh38
rs184257829 Chr.21: 26920081-26920081 on Build GRCh38
rs1046251223 Chr.21: 26919822-26919822 on Build GRCh38
rs954218492 Chr.21: 26920108-26920108 on Build GRCh38
rs932114104 Chr.21: 26919712-26919712 on Build GRCh38
rs966318588 Chr.21: 26919942-26919942 on Build GRCh38
rs530920930 Chr.21: 26919863-26919863 on Build GRCh38
rs569783610 Chr.21: 26919852-26919852 on Build GRCh38
rs551306582 Chr.21: 26919778-26919778 on Build GRCh38
rs999194167 Chr.21: 26919932-26919932 on Build GRCh38
rs910183354 Chr.21: 26919747-26919747 on Build GRCh38
rs533227756 Chr.21: 26919735-26919735 on Build GRCh38
rs562760609 Chr.21: 26919884-26919884 on Build GRCh38
rs11910638 Chr.21: 26919717-26919717 on Build GRCh38
rs372509258 Chr.21: 26919766-26919766 on Build GRCh38
rs945529130 Chr.21: 26919703-26919703 on Build GRCh38
rs940708868 Chr.21: 26919733-26919733 on Build GRCh38
rs1029185777 Chr.21: 26920003-26920003 on Build GRCh38
rs985803670 Chr.21: 26920110-26920110 on Build GRCh38
rs1011976602 Chr.21: 26919767-26919767 on Build GRCh38
rs143398098 Chr.21: 26920021-26920024 on Build GRCh38
rs189513706 Chr.21: 26920121-26920121 on Build GRCh38
rs755931519 Chr.21: 26920052-26920052 on Build GRCh38
rs142385271 Chr.21: 26919896-26919896 on Build GRCh38
rs549499170 Chr.21: 26919924-26919924 on Build GRCh38
rs781607766 Chr.21: 26919832-26919832 on Build GRCh38
rs906375612 Chr.21: 26919830-26919830 on Build GRCh38
rs914200618 Chr.21: 26919652-26919652 on Build GRCh38
rs993720950 Chr.21: 26919771-26919771 on Build GRCh38
rs372994329 Chr.21: 26920022-26920025 on Build GRCh38
rs976643936 Chr.21: 26919991-26919991 on Build GRCh38
rs1031255619 Chr.21: 26919937-26919937 on Build GRCh38
rs918494230 Chr.21: 26920117-26920117 on Build GRCh38
rs762044694 Chr.21: 26919710-26919710 on Build GRCh38
rs1010948525 Chr.21: 26920019-26920019 on Build GRCh38
rs1036874792 Chr.21: 26919739-26919739 on Build GRCh38
rs530132278 Chr.21: 26919986-26919986 on Build GRCh38
rs1002257010 Chr.21: 26919879-26919879 on Build GRCh38
rs977428222 Chr.21: 26920070-26920070 on Build GRCh38
rs191871015 Chr.21: 26919984-26919984 on Build GRCh38
rs553853916 Chr.21: 26919748-26919748 on Build GRCh38
rs1052961 Chr.21: 26920031-26920031 on Build GRCh38
rs1021613159 Chr.21: 26920042-26920042 on Build GRCh38
rs542156640 Chr.21: 26919914-26919914 on Build GRCh38
rs369831517 Chr.21: 26919756-26919756 on Build GRCh38
rs767130794 Chr.21: 26919855-26919855 on Build GRCh38
rs567420954 Chr.21: 26920080-26920080 on Build GRCh38
rs922996626 Chr.21: 26919722-26919722 on Build GRCh38
rs539913768 Chr.21: 26919742-26919742 on Build GRCh38
rs1056332339 Chr.21: 26919750-26919750 on Build GRCh38
rs910341898 Chr.21: 26920123-26920123 on Build GRCh38

View Full Product Details