Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 30592353-30592847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRTAP6-3
Entrez Gene ID
337968
Gene Name
keratin associated protein 6-3
Gene Aliases
KAP6.3
UniGene
Hs.553691
Gene Chromosome Location
Chr.21: 30592440-30593055 on Build GRCh38
Transcripts
NM_181605.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4926338 c.203G>C p.G68A Chr.21: 30592669-30592669 on Build GRCh38
725349 c.97G>A p.G33R Chr.21: 30592563-30592563 on Build GRCh38
1029916 c.208G>A p.D70N Chr.21: 30592674-30592674 on Build GRCh38
4697055 c.150C>T p.G50G Chr.21: 30592616-30592616 on Build GRCh38
1029914 c.158G>A p.G53D Chr.21: 30592624-30592624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770399639 Chr.21: 30592543-30592543 on Build GRCh38
rs774211941 Chr.21: 30592675-30592676 on Build GRCh38
rs79025761 Chr.21: 30592578-30592578 on Build GRCh38
rs760560413 Chr.21: 30592638-30592638 on Build GRCh38
rs773866515 Chr.21: 30592425-30592425 on Build GRCh38
rs375662433 Chr.21: 30592467-30592467 on Build GRCh38
rs750023181 Chr.21: 30592661-30592661 on Build GRCh38
rs778670054 Chr.21: 30592525-30592525 on Build GRCh38
rs747154924 Chr.21: 30592768-30592768 on Build GRCh38
rs199859574 Chr.21: 30592551-30592551 on Build GRCh38
rs781155383 Chr.21: 30592398-30592398 on Build GRCh38
rs775725149 Chr.21: 30592749-30592749 on Build GRCh38
rs376835665 Chr.21: 30592411-30592411 on Build GRCh38
rs372192565 Chr.21: 30592795-30592795 on Build GRCh38
rs147261100 Chr.21: 30592558-30592559 on Build GRCh38
rs888410412 Chr.21: 30592510-30592510 on Build GRCh38
rs772792339 Chr.21: 30592702-30592702 on Build GRCh38
rs1048717129 Chr.21: 30592505-30592505 on Build GRCh38
rs376388028 Chr.21: 30592745-30592745 on Build GRCh38
rs75088048 Chr.21: 30592588-30592588 on Build GRCh38
rs370061319 Chr.21: 30592739-30592739 on Build GRCh38
rs759365747 Chr.21: 30592493-30592493 on Build GRCh38
rs770687678 Chr.21: 30592414-30592414 on Build GRCh38
rs748670186 Chr.21: 30592538-30592538 on Build GRCh38
rs775670063 Chr.21: 30592630-30592630 on Build GRCh38
rs768420649 Chr.21: 30592676-30592687 on Build GRCh38
rs761532391 Chr.21: 30592715-30592715 on Build GRCh38
rs777284244 Chr.21: 30592410-30592410 on Build GRCh38
rs776223199 Chr.21: 30592784-30592784 on Build GRCh38
rs904738647 Chr.21: 30592413-30592413 on Build GRCh38
rs753690583 Chr.21: 30592574-30592574 on Build GRCh38
rs751246983 Chr.21: 30592822-30592822 on Build GRCh38
rs748580226 Chr.21: 30592590-30592590 on Build GRCh38
rs114855973 Chr.21: 30592478-30592478 on Build GRCh38
rs747037667 Chr.21: 30592591-30592592 on Build GRCh38
rs754754605 Chr.21: 30592396-30592396 on Build GRCh38
rs767310644 Chr.21: 30592559-30592559 on Build GRCh38
rs368459086 Chr.21: 30592514-30592514 on Build GRCh38
rs767289791 Chr.21: 30592700-30592711 on Build GRCh38
rs376564418 Chr.21: 30592818-30592818 on Build GRCh38
rs751756268 Chr.21: 30592491-30592491 on Build GRCh38
rs749811759 Chr.21: 30592710-30592710 on Build GRCh38
rs13433456 Chr.21: 30592591-30592591 on Build GRCh38
rs113468962 Chr.21: 30592550-30592550 on Build GRCh38
rs9305426 Chr.21: 30592597-30592597 on Build GRCh38
rs13433449 Chr.21: 30592609-30592609 on Build GRCh38
rs987810002 Chr.21: 30592399-30592399 on Build GRCh38
rs774586130 Chr.21: 30592552-30592552 on Build GRCh38
rs141367067 Chr.21: 30592594-30592594 on Build GRCh38
rs568427466 Chr.21: 30592526-30592526 on Build GRCh38
rs777310267 Chr.21: 30592463-30592463 on Build GRCh38
rs772344374 Chr.21: 30592625-30592625 on Build GRCh38
rs932816724 Chr.21: 30592624-30592624 on Build GRCh38
rs761804861 Chr.21: 30592682-30592683 on Build GRCh38
rs957738721 Chr.21: 30592393-30592393 on Build GRCh38
rs781643901 Chr.21: 30592533-30592533 on Build GRCh38
rs371118726 Chr.21: 30592549-30592549 on Build GRCh38
rs1053329444 Chr.21: 30592613-30592613 on Build GRCh38
rs1037016015 Chr.21: 30592717-30592717 on Build GRCh38
rs760377372 Chr.21: 30592566-30592566 on Build GRCh38
rs763761475 Chr.21: 30592642-30592642 on Build GRCh38
rs546860400 Chr.21: 30592816-30592816 on Build GRCh38
rs777148513 Chr.21: 30592513-30592513 on Build GRCh38
rs372952842 Chr.21: 30592451-30592451 on Build GRCh38
rs901383687 Chr.21: 30592453-30592455 on Build GRCh38
rs762936047 Chr.21: 30592475-30592475 on Build GRCh38
rs373786352 Chr.21: 30592495-30592495 on Build GRCh38
rs771164991 Chr.21: 30592597-30592603 on Build GRCh38
rs776795552 Chr.21: 30592605-30592618 on Build GRCh38
rs749123858 Chr.21: 30592631-30592675 on Build GRCh38
rs530698443 Chr.21: 30592689-30592689 on Build GRCh38
rs904985091 Chr.21: 30592750-30592750 on Build GRCh38
rs773276891 Chr.21: 30592469-30592469 on Build GRCh38
rs765246395 Chr.21: 30592468-30592468 on Build GRCh38
rs776655876 Chr.21: 30592645-30592645 on Build GRCh38
rs765325154 Chr.21: 30592800-30592800 on Build GRCh38
rs756532819 Chr.21: 30592690-30592690 on Build GRCh38
rs773260109 Chr.21: 30592801-30592801 on Build GRCh38
rs200489704 Chr.21: 30592674-30592674 on Build GRCh38
rs752651693 Chr.21: 30592502-30592502 on Build GRCh38
rs751139257 Chr.21: 30592684-30592684 on Build GRCh38
rs752418918 Chr.21: 30592564-30592564 on Build GRCh38
rs540161429 Chr.21: 30592577-30592577 on Build GRCh38
rs924940237 Chr.21: 30592375-30592375 on Build GRCh38
rs79130546 Chr.21: 30592693-30592693 on Build GRCh38
rs747429523 Chr.21: 30592427-30592427 on Build GRCh38
rs867876654 Chr.21: 30592618-30592618 on Build GRCh38
rs113124062 Chr.21: 30592735-30592735 on Build GRCh38
rs748045735 Chr.21: 30592401-30592401 on Build GRCh38
rs71321338 Chr.21: 30592527-30592527 on Build GRCh38
rs778383968 Chr.21: 30592595-30592595 on Build GRCh38
rs764844835 Chr.21: 30592660-30592660 on Build GRCh38
rs375609643 Chr.21: 30592606-30592606 on Build GRCh38
rs761599011 Chr.21: 30592646-30592646 on Build GRCh38
rs770779425 Chr.21: 30592530-30592530 on Build GRCh38
rs367824135 Chr.21: 30592519-30592519 on Build GRCh38
rs932904856 Chr.21: 30592497-30592497 on Build GRCh38
rs376602609 Chr.21: 30592726-30592726 on Build GRCh38
rs768510017 Chr.21: 30592775-30592775 on Build GRCh38
rs74715941 Chr.21: 30592570-30592570 on Build GRCh38
rs111531400 Chr.21: 30592740-30592740 on Build GRCh38
rs377389368 Chr.21: 30592555-30592555 on Build GRCh38
rs755650281 Chr.21: 30592405-30592405 on Build GRCh38
rs750115408 Chr.21: 30592582-30592582 on Build GRCh38
rs756017477 Chr.21: 30592506-30592506 on Build GRCh38
rs370739777 Chr.21: 30592448-30592448 on Build GRCh38
rs777949636 Chr.21: 30592697-30592697 on Build GRCh38

View Full Product Details