Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 31558752-31559254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC150051
Entrez Gene ID
150051
Gene Name
uncharacterized LOC150051
Gene Aliases
-
UniGene
Hs.708976
Gene Chromosome Location
Chr.21: 31559245-31560487 on Build GRCh38
Transcripts
NM_001321023.1 NM_003253.2 XM_011529712.1 XM_017028447.1 XM_017028449.1 XM_017028452.1 XM_017028453.1

Gene Symbol
TIAM1
Entrez Gene ID
7074
Gene Name
T-cell lymphoma invasion and metastasis 1
Gene Aliases
-
UniGene
Hs.517228
Gene Chromosome Location
Chr.21: 31118418-31559977 on Build GRCh38
Transcripts
NM_001321023.1 NM_003253.2 XM_011529712.1 XM_017028447.1 XM_017028449.1 XM_017028452.1 XM_017028453.1

SNP Detail

SNP ID Chromosome Location
rs919237739 Chr.21: 31559067-31559067 on Build GRCh38
rs1020538308 Chr.21: 31559012-31559012 on Build GRCh38
rs976557355 Chr.21: 31558875-31558875 on Build GRCh38
rs1046497015 Chr.21: 31559080-31559080 on Build GRCh38
rs1042048595 Chr.21: 31559106-31559106 on Build GRCh38
rs564799762 Chr.21: 31558836-31558836 on Build GRCh38
rs528820322 Chr.21: 31558870-31558870 on Build GRCh38
rs1028478129 Chr.21: 31559024-31559024 on Build GRCh38
rs1028112943 Chr.21: 31559112-31559112 on Build GRCh38
rs937212165 Chr.21: 31558832-31558832 on Build GRCh38
rs942082119 Chr.21: 31558928-31558928 on Build GRCh38
rs932575621 Chr.21: 31558914-31558914 on Build GRCh38
rs930694978 Chr.21: 31558774-31558774 on Build GRCh38
rs1002429285 Chr.21: 31558820-31558820 on Build GRCh38
rs1055608720 Chr.21: 31559018-31559018 on Build GRCh38
rs9983409 Chr.21: 31559109-31559109 on Build GRCh38
rs909837224 Chr.21: 31559061-31559061 on Build GRCh38
rs1041455158 Chr.21: 31559041-31559041 on Build GRCh38
rs1041527890 Chr.21: 31559202-31559202 on Build GRCh38
rs1040694177 Chr.21: 31559054-31559054 on Build GRCh38
rs551101930 Chr.21: 31558971-31558971 on Build GRCh38
rs4817404 Chr.21: 31558915-31558915 on Build GRCh38
rs949411711 Chr.21: 31558958-31558958 on Build GRCh38
rs943787199 Chr.21: 31559217-31559217 on Build GRCh38
rs947182420 Chr.21: 31559038-31559038 on Build GRCh38
rs371887281 Chr.21: 31558821-31558821 on Build GRCh38
rs902848926 Chr.21: 31558880-31558880 on Build GRCh38
rs894039557 Chr.21: 31559002-31559002 on Build GRCh38
rs899502952 Chr.21: 31559227-31559227 on Build GRCh38
rs1025115926 Chr.21: 31558810-31558810 on Build GRCh38
rs111349886 Chr.21: 31559075-31559075 on Build GRCh38
rs940665525 Chr.21: 31558811-31558811 on Build GRCh38
rs568451754 Chr.21: 31558919-31558919 on Build GRCh38
rs961251258 Chr.21: 31559065-31559065 on Build GRCh38
rs914424306 Chr.21: 31559037-31559037 on Build GRCh38
rs958933225 Chr.21: 31559150-31559150 on Build GRCh38
rs1029150788 Chr.21: 31558922-31558922 on Build GRCh38
rs576697880 Chr.21: 31559108-31559108 on Build GRCh38
rs1005395234 Chr.21: 31559131-31559131 on Build GRCh38
rs1017142609 Chr.21: 31559135-31559135 on Build GRCh38
rs987762374 Chr.21: 31558842-31558842 on Build GRCh38
rs1009750341 Chr.21: 31559103-31559103 on Build GRCh38
rs942013451 Chr.21: 31558772-31558772 on Build GRCh38
rs938224748 Chr.21: 31559096-31559096 on Build GRCh38
rs997861116 Chr.21: 31559016-31559016 on Build GRCh38
rs957749096 Chr.21: 31558839-31558839 on Build GRCh38
rs557693432 Chr.21: 31558818-31558818 on Build GRCh38
rs953920716 Chr.21: 31559113-31559113 on Build GRCh38
rs984359041 Chr.21: 31559057-31559057 on Build GRCh38
rs892918084 Chr.21: 31558847-31558847 on Build GRCh38
rs569743265 Chr.21: 31559013-31559013 on Build GRCh38
rs984815778 Chr.21: 31558796-31558796 on Build GRCh38
rs922048494 Chr.21: 31558773-31558773 on Build GRCh38
rs1036472952 Chr.21: 31558779-31558779 on Build GRCh38
rs1013798028 Chr.21: 31558794-31558794 on Build GRCh38
rs1041161861 Chr.21: 31559222-31559222 on Build GRCh38
rs897922258 Chr.21: 31558953-31558953 on Build GRCh38
rs902923360 Chr.21: 31559053-31559053 on Build GRCh38
rs917151243 Chr.21: 31559166-31559166 on Build GRCh38
rs970755691 Chr.21: 31559174-31559174 on Build GRCh38
rs947255867 Chr.21: 31559199-31559199 on Build GRCh38
rs1020695334 Chr.21: 31558857-31558857 on Build GRCh38
rs1009195683 Chr.21: 31559011-31559011 on Build GRCh38
rs928667079 Chr.21: 31559124-31559124 on Build GRCh38
rs953926878 Chr.21: 31559029-31559029 on Build GRCh38
rs552651581 Chr.21: 31559197-31559197 on Build GRCh38
rs907896671 Chr.21: 31558805-31558805 on Build GRCh38
rs1032632965 Chr.21: 31558991-31558991 on Build GRCh38
rs972063819 Chr.21: 31559066-31559066 on Build GRCh38
rs1054329878 Chr.21: 31559097-31559097 on Build GRCh38
rs926733192 Chr.21: 31559230-31559230 on Build GRCh38
rs1013296524 Chr.21: 31559213-31559213 on Build GRCh38
rs926695797 Chr.21: 31559083-31559083 on Build GRCh38
rs972617738 Chr.21: 31559165-31559165 on Build GRCh38
rs982510950 Chr.21: 31559210-31559210 on Build GRCh38
rs961305287 Chr.21: 31559153-31559153 on Build GRCh38
rs921081641 Chr.21: 31558912-31558912 on Build GRCh38
rs1002763677 Chr.21: 31558989-31558989 on Build GRCh38
rs535533000 Chr.21: 31558967-31558967 on Build GRCh38
rs894060371 Chr.21: 31559100-31559100 on Build GRCh38
rs984878977 Chr.21: 31558963-31558963 on Build GRCh38
rs1007859065 Chr.21: 31558908-31558908 on Build GRCh38
rs558408316 Chr.21: 31559084-31559084 on Build GRCh38
rs897912394 Chr.21: 31558782-31558782 on Build GRCh38
rs773039692 Chr.21: 31558837-31558837 on Build GRCh38
rs996442183 Chr.21: 31559064-31559064 on Build GRCh38
rs1036358255 Chr.21: 31558929-31558929 on Build GRCh38

View Full Product Details