Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 31995686-31996200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HUNK
Entrez Gene ID
30811
Gene Name
hormonally up-regulated Neu-associated kinase
Gene Aliases
-
UniGene
Hs.109437
Gene Chromosome Location
Chr.21: 31873315-32004064 on Build GRCh38
Transcripts
NM_014586.1 XM_011529537.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
115692 c.1361C>T p.S454F Chr.21: 31995823-31995823 on Build GRCh38
1686816 c.1306-1G>A p.? Chr.21: 31995767-31995767 on Build GRCh38
2842899 c.1352G>A p.R451Q Chr.21: 31995814-31995814 on Build GRCh38
1315792 c.1381C>T p.L461L Chr.21: 31995843-31995843 on Build GRCh38
6381211 c.1460C>A p.A487D Chr.21: 31995922-31995922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770081341 Chr.21: 31995994-31995994 on Build GRCh38
rs748439256 Chr.21: 31995989-31995989 on Build GRCh38
rs188263586 Chr.21: 31995929-31995929 on Build GRCh38
rs986331856 Chr.21: 31996027-31996027 on Build GRCh38
rs372384937 Chr.21: 31995836-31995836 on Build GRCh38
rs765288551 Chr.21: 31995813-31995813 on Build GRCh38
rs758949628 Chr.21: 31995960-31995960 on Build GRCh38
rs529590745 Chr.21: 31995884-31995884 on Build GRCh38
rs796274193 Chr.21: 31995794-31995794 on Build GRCh38
rs566593564 Chr.21: 31996001-31996001 on Build GRCh38
rs147107512 Chr.21: 31996151-31996151 on Build GRCh38
rs11411823 Chr.21: 31996098-31996099 on Build GRCh38
rs754218827 Chr.21: 31995865-31995865 on Build GRCh38
rs141875933 Chr.21: 31995941-31995941 on Build GRCh38
rs966597632 Chr.21: 31996022-31996022 on Build GRCh38
rs775429945 Chr.21: 31995753-31995753 on Build GRCh38
rs568303971 Chr.21: 31996103-31996103 on Build GRCh38
rs986300625 Chr.21: 31995837-31995837 on Build GRCh38
rs761018227 Chr.21: 31995754-31995754 on Build GRCh38
rs903837431 Chr.21: 31996165-31996165 on Build GRCh38
rs746355478 Chr.21: 31995741-31995741 on Build GRCh38
rs780387782 Chr.21: 31995969-31995969 on Build GRCh38
rs376391627 Chr.21: 31995979-31995979 on Build GRCh38
rs780198562 Chr.21: 31995879-31995879 on Build GRCh38
rs953686896 Chr.21: 31996008-31996008 on Build GRCh38
rs546384374 Chr.21: 31996100-31996100 on Build GRCh38
rs1046724932 Chr.21: 31996037-31996037 on Build GRCh38
rs909526019 Chr.21: 31996039-31996039 on Build GRCh38
rs371144574 Chr.21: 31995983-31995983 on Build GRCh38
rs972177098 Chr.21: 31996057-31996057 on Build GRCh38
rs1044772524 Chr.21: 31996061-31996061 on Build GRCh38
rs868467388 Chr.21: 31995783-31995783 on Build GRCh38
rs953715019 Chr.21: 31995802-31995802 on Build GRCh38
rs369629181 Chr.21: 31995952-31995952 on Build GRCh38
rs922321602 Chr.21: 31995807-31995807 on Build GRCh38
rs1005226062 Chr.21: 31995905-31995905 on Build GRCh38
rs201715104 Chr.21: 31996099-31996099 on Build GRCh38
rs1009453655 Chr.21: 31996137-31996137 on Build GRCh38
rs909375559 Chr.21: 31995848-31995848 on Build GRCh38
rs201787585 Chr.21: 31995935-31995935 on Build GRCh38
rs765969721 Chr.21: 31995850-31995850 on Build GRCh38
rs370008993 Chr.21: 31995919-31995919 on Build GRCh38
rs748932644 Chr.21: 31995994-31995994 on Build GRCh38
rs143593780 Chr.21: 31995946-31995946 on Build GRCh38
rs535722479 Chr.21: 31996117-31996117 on Build GRCh38
rs551684571 Chr.21: 31995871-31995871 on Build GRCh38
rs910744638 Chr.21: 31996017-31996017 on Build GRCh38
rs773144172 Chr.21: 31996000-31996000 on Build GRCh38
rs749585396 Chr.21: 31995719-31995719 on Build GRCh38
rs941813154 Chr.21: 31995869-31995869 on Build GRCh38
rs375328601 Chr.21: 31995958-31995958 on Build GRCh38
rs142391016 Chr.21: 31995814-31995814 on Build GRCh38
rs926270331 Chr.21: 31996167-31996167 on Build GRCh38
rs762978845 Chr.21: 31995831-31995831 on Build GRCh38
rs776491721 Chr.21: 31995761-31995761 on Build GRCh38
rs377150590 Chr.21: 31995942-31995942 on Build GRCh38
rs748626318 Chr.21: 31996045-31996045 on Build GRCh38
rs138721520 Chr.21: 31995878-31995878 on Build GRCh38
rs771455236 Chr.21: 31995731-31995731 on Build GRCh38
rs754660206 Chr.21: 31995852-31995852 on Build GRCh38
rs752390485 Chr.21: 31995944-31995944 on Build GRCh38
rs780665957 Chr.21: 31995857-31995857 on Build GRCh38
rs1046448240 Chr.21: 31995876-31995876 on Build GRCh38
rs545889182 Chr.21: 31995743-31995743 on Build GRCh38
rs2070371 Chr.21: 31995875-31995875 on Build GRCh38
rs1003777366 Chr.21: 31996105-31996105 on Build GRCh38
rs544322969 Chr.21: 31995914-31995914 on Build GRCh38
rs572334874 Chr.21: 31995793-31995793 on Build GRCh38
rs751108454 Chr.21: 31995939-31995939 on Build GRCh38
rs764165478 Chr.21: 31995758-31995758 on Build GRCh38
rs750975114 Chr.21: 31995851-31995851 on Build GRCh38
rs564859765 Chr.21: 31995779-31995779 on Build GRCh38
rs201734640 Chr.21: 31996089-31996097 on Build GRCh38
rs949156500 Chr.21: 31996147-31996147 on Build GRCh38
rs750731147 Chr.21: 31995956-31995956 on Build GRCh38
rs779221441 Chr.21: 31995872-31995872 on Build GRCh38
rs397790622 Chr.21: 31996099-31996100 on Build GRCh38
rs976397404 Chr.21: 31996023-31996023 on Build GRCh38
rs4817444 Chr.21: 31996135-31996135 on Build GRCh38
rs768036040 Chr.21: 31996004-31996004 on Build GRCh38
rs11702842 Chr.21: 31996042-31996042 on Build GRCh38
rs78415327 Chr.21: 31996174-31996174 on Build GRCh38
rs762096000 Chr.21: 31995915-31995915 on Build GRCh38
rs769777844 Chr.21: 31995916-31995916 on Build GRCh38
rs937936134 Chr.21: 31996170-31996170 on Build GRCh38

View Full Product Details