Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33070252-33070742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OLIG1
Entrez Gene ID
116448
Gene Name
oligodendrocyte transcription factor 1
Gene Aliases
BHLHB6, BHLHE21
UniGene
Hs.56663
Gene Chromosome Location
Chr.21: 33070144-33072422 on Build GRCh38
Transcripts
NM_138983.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5986851 c.266A>T p.Q89L Chr.21: 33070560-33070560 on Build GRCh38
6033347 c.274C>T p.R92C Chr.21: 33070568-33070568 on Build GRCh38
5858536 c.77_79delCCT p.S32delS Chr.21: 33070371-33070373 on Build GRCh38
6209305 c.161G>A p.R54H Chr.21: 33070455-33070455 on Build GRCh38
6362678 c.47T>C p.L16P Chr.21: 33070341-33070341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs935888617 Chr.21: 33070387-33070387 on Build GRCh38
rs781464064 Chr.21: 33070342-33070342 on Build GRCh38
rs756942552 Chr.21: 33070386-33070391 on Build GRCh38
rs768992557 Chr.21: 33070547-33070548 on Build GRCh38
rs1034485829 Chr.21: 33070370-33070370 on Build GRCh38
rs760488698 Chr.21: 33070527-33070527 on Build GRCh38
rs577420887 Chr.21: 33070340-33070340 on Build GRCh38
rs745955940 Chr.21: 33070628-33070628 on Build GRCh38
rs916905247 Chr.21: 33070371-33070379 on Build GRCh38
rs934051950 Chr.21: 33070408-33070408 on Build GRCh38
rs1004796618 Chr.21: 33070310-33070310 on Build GRCh38
rs763989022 Chr.21: 33070530-33070530 on Build GRCh38
rs778074163 Chr.21: 33070609-33070609 on Build GRCh38
rs780798020 Chr.21: 33070401-33070415 on Build GRCh38
rs771034018 Chr.21: 33070396-33070396 on Build GRCh38
rs866564583 Chr.21: 33070596-33070596 on Build GRCh38
rs758049165 Chr.21: 33070589-33070589 on Build GRCh38
rs750993532 Chr.21: 33070383-33070391 on Build GRCh38
rs1051132311 Chr.21: 33070420-33070420 on Build GRCh38
rs756462721 Chr.21: 33070402-33070403 on Build GRCh38
rs756520834 Chr.21: 33070604-33070604 on Build GRCh38
rs763674411 Chr.21: 33070371-33070376 on Build GRCh38
rs775216609 Chr.21: 33070499-33070499 on Build GRCh38
rs761899993 Chr.21: 33070698-33070698 on Build GRCh38
rs749811689 Chr.21: 33070432-33070440 on Build GRCh38
rs1034173984 Chr.21: 33070545-33070545 on Build GRCh38
rs538825906 Chr.21: 33070378-33070378 on Build GRCh38
rs753855485 Chr.21: 33070534-33070534 on Build GRCh38
rs759772548 Chr.21: 33070493-33070493 on Build GRCh38
rs753049454 Chr.21: 33070601-33070601 on Build GRCh38
rs749453528 Chr.21: 33070412-33070445 on Build GRCh38
rs762499538 Chr.21: 33070710-33070710 on Build GRCh38
rs889092911 Chr.21: 33070405-33070405 on Build GRCh38
rs758010856 Chr.21: 33070318-33070318 on Build GRCh38
rs945835877 Chr.21: 33070303-33070303 on Build GRCh38
rs1014800611 Chr.21: 33070315-33070315 on Build GRCh38
rs887105812 Chr.21: 33070477-33070477 on Build GRCh38
rs1008881549 Chr.21: 33070434-33070434 on Build GRCh38
rs375959063 Chr.21: 33070402-33070402 on Build GRCh38
rs375917294 Chr.21: 33070672-33070672 on Build GRCh38
rs553283318 Chr.21: 33070454-33070454 on Build GRCh38
rs761773545 Chr.21: 33070535-33070535 on Build GRCh38
rs772224219 Chr.21: 33070634-33070634 on Build GRCh38
rs796943372 Chr.21: 33070418-33070418 on Build GRCh38
rs776510478 Chr.21: 33070678-33070678 on Build GRCh38
rs779722956 Chr.21: 33070600-33070600 on Build GRCh38
rs372626465 Chr.21: 33070657-33070657 on Build GRCh38
rs779026714 Chr.21: 33070625-33070625 on Build GRCh38
rs772885878 Chr.21: 33070708-33070708 on Build GRCh38
rs765325129 Chr.21: 33070371-33070373 on Build GRCh38
rs772325064 Chr.21: 33070494-33070494 on Build GRCh38
rs1039806724 Chr.21: 33070391-33070392 on Build GRCh38
rs1036655291 Chr.21: 33070507-33070507 on Build GRCh38
rs966981434 Chr.21: 33070562-33070562 on Build GRCh38
rs940592425 Chr.21: 33070498-33070498 on Build GRCh38
rs749939573 Chr.21: 33070588-33070588 on Build GRCh38
rs563674754 Chr.21: 33070653-33070653 on Build GRCh38
rs542137414 Chr.21: 33070561-33070561 on Build GRCh38
rs112561383 Chr.21: 33070528-33070528 on Build GRCh38
rs1040806250 Chr.21: 33070448-33070448 on Build GRCh38
rs748537947 Chr.21: 33070375-33070375 on Build GRCh38
rs750104907 Chr.21: 33070316-33070316 on Build GRCh38
rs765257688 Chr.21: 33070702-33070702 on Build GRCh38
rs1040982549 Chr.21: 33070400-33070400 on Build GRCh38
rs778525404 Chr.21: 33070282-33070282 on Build GRCh38
rs977713563 Chr.21: 33070642-33070642 on Build GRCh38
rs764852874 Chr.21: 33070567-33070567 on Build GRCh38
rs552498250 Chr.21: 33070370-33070371 on Build GRCh38
rs757706857 Chr.21: 33070624-33070624 on Build GRCh38
rs74275610 Chr.21: 33070379-33070379 on Build GRCh38
rs774351586 Chr.21: 33070492-33070492 on Build GRCh38
rs749749002 Chr.21: 33070394-33070394 on Build GRCh38
rs778229300 Chr.21: 33070391-33070391 on Build GRCh38
rs575025407 Chr.21: 33070489-33070489 on Build GRCh38
rs367752518 Chr.21: 33070411-33070411 on Build GRCh38
rs1023926991 Chr.21: 33070551-33070551 on Build GRCh38
rs775760606 Chr.21: 33070645-33070645 on Build GRCh38
rs914276953 Chr.21: 33070371-33070394 on Build GRCh38
rs780214215 Chr.21: 33070409-33070411 on Build GRCh38
rs746287972 Chr.21: 33070403-33070411 on Build GRCh38
rs899012163 Chr.21: 33070532-33070532 on Build GRCh38
rs749803949 Chr.21: 33070621-33070621 on Build GRCh38
rs994605314 Chr.21: 33070343-33070343 on Build GRCh38

View Full Product Details