Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33071746-33072289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OLIG1
Entrez Gene ID
116448
Gene Name
oligodendrocyte transcription factor 1
Gene Aliases
BHLHB6, BHLHE21
UniGene
Hs.56663
Gene Chromosome Location
Chr.21: 33070144-33072422 on Build GRCh38
Transcripts
NM_138983.2

SNP Detail

SNP ID Chromosome Location
rs997341531 Chr.21: 33072249-33072249 on Build GRCh38
rs977392827 Chr.21: 33072033-33072034 on Build GRCh38
rs568563980 Chr.21: 33072034-33072034 on Build GRCh38
rs902798291 Chr.21: 33071900-33071900 on Build GRCh38
rs77910593 Chr.21: 33071800-33071800 on Build GRCh38
rs956017391 Chr.21: 33072260-33072260 on Build GRCh38
rs539740659 Chr.21: 33072127-33072127 on Build GRCh38
rs1032673921 Chr.21: 33071910-33071910 on Build GRCh38
rs918070738 Chr.21: 33072100-33072100 on Build GRCh38
rs1031407231 Chr.21: 33072231-33072231 on Build GRCh38
rs1037084731 Chr.21: 33072121-33072121 on Build GRCh38
rs1042639611 Chr.21: 33071877-33071877 on Build GRCh38
rs1049118885 Chr.21: 33071769-33071769 on Build GRCh38
rs570737418 Chr.21: 33071923-33071923 on Build GRCh38
rs184431046 Chr.21: 33072208-33072208 on Build GRCh38
rs914351140 Chr.21: 33071825-33071826 on Build GRCh38
rs551749045 Chr.21: 33071838-33071838 on Build GRCh38
rs981286631 Chr.21: 33071798-33071798 on Build GRCh38
rs981339013 Chr.21: 33072059-33072059 on Build GRCh38
rs1007285726 Chr.21: 33072173-33072173 on Build GRCh38
rs754327172 Chr.21: 33072210-33072210 on Build GRCh38
rs767581828 Chr.21: 33072090-33072090 on Build GRCh38
rs959700866 Chr.21: 33071783-33071783 on Build GRCh38
rs1027551295 Chr.21: 33071986-33071986 on Build GRCh38
rs1013810374 Chr.21: 33071858-33071858 on Build GRCh38
rs9982759 Chr.21: 33072169-33072169 on Build GRCh38
rs1012295133 Chr.21: 33072185-33072185 on Build GRCh38
rs995711435 Chr.21: 33071976-33071976 on Build GRCh38
rs11554600 Chr.21: 33071831-33071831 on Build GRCh38
rs117130421 Chr.21: 33071996-33071996 on Build GRCh38
rs1007729062 Chr.21: 33071939-33071939 on Build GRCh38
rs897332022 Chr.21: 33071968-33071968 on Build GRCh38
rs1012285405 Chr.21: 33072005-33072005 on Build GRCh38
rs1024936885 Chr.21: 33072014-33072014 on Build GRCh38
rs909217590 Chr.21: 33072091-33072091 on Build GRCh38
rs558450994 Chr.21: 33072166-33072166 on Build GRCh38
rs776676271 Chr.21: 33071869-33071869 on Build GRCh38
rs934483629 Chr.21: 33071801-33071801 on Build GRCh38
rs892806885 Chr.21: 33072170-33072170 on Build GRCh38
rs897432555 Chr.21: 33072130-33072130 on Build GRCh38
rs750669801 Chr.21: 33072139-33072139 on Build GRCh38
rs966289825 Chr.21: 33072007-33072007 on Build GRCh38
rs150885224 Chr.21: 33072060-33072060 on Build GRCh38
rs575472310 Chr.21: 33072117-33072117 on Build GRCh38
rs984785965 Chr.21: 33072088-33072088 on Build GRCh38
rs755501135 Chr.21: 33072213-33072213 on Build GRCh38
rs975963415 Chr.21: 33072028-33072028 on Build GRCh38
rs189108894 Chr.21: 33071931-33071931 on Build GRCh38
rs938077311 Chr.21: 33072097-33072097 on Build GRCh38
rs917911361 Chr.21: 33071788-33071788 on Build GRCh38
rs62214174 Chr.21: 33072083-33072083 on Build GRCh38
rs141573781 Chr.21: 33071795-33071795 on Build GRCh38
rs746859831 Chr.21: 33072075-33072076 on Build GRCh38
rs1055633111 Chr.21: 33072098-33072098 on Build GRCh38
rs561598181 Chr.21: 33072222-33072222 on Build GRCh38
rs949565994 Chr.21: 33072109-33072109 on Build GRCh38
rs998465514 Chr.21: 33071901-33071901 on Build GRCh38

View Full Product Details