Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33577403-33577908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DONSON
Entrez Gene ID
29980
Gene Name
downstream neighbor of SON
Gene Aliases
B17, C21orf60
UniGene
Hs.436341
Gene Chromosome Location
Chr.21: 33577551-33588708 on Build GRCh38
Transcripts
NM_017613.3 NM_001291412.1 NM_138927.2 NR_103797.1

Gene Symbol
SON
Entrez Gene ID
6651
Gene Name
SON DNA binding protein
Gene Aliases
BASS1, C21orf50, DBP-5, NREBP, SON3
UniGene
Hs.517262
Gene Chromosome Location
Chr.21: 33543038-33577514 on Build GRCh38
Transcripts
NM_017613.3 NM_001291412.1 NM_138927.2 NR_103797.1

SNP Detail

SNP ID Chromosome Location
rs868725305 Chr.21: 33577660-33577660 on Build GRCh38
rs951679344 Chr.21: 33577661-33577661 on Build GRCh38
rs867862115 Chr.21: 33577664-33577664 on Build GRCh38
rs199830795 Chr.21: 33577703-33577703 on Build GRCh38
rs199567196 Chr.21: 33577854-33577854 on Build GRCh38
rs564668700 Chr.21: 33577616-33577616 on Build GRCh38
rs1046448200 Chr.21: 33577672-33577672 on Build GRCh38
rs1047175849 Chr.21: 33577608-33577608 on Build GRCh38
rs374825441 Chr.21: 33577689-33577689 on Build GRCh38
rs543114585 Chr.21: 33577583-33577583 on Build GRCh38
rs868011052 Chr.21: 33577699-33577699 on Build GRCh38
rs906206340 Chr.21: 33577674-33577674 on Build GRCh38
rs866659692 Chr.21: 33577654-33577654 on Build GRCh38
rs893627047 Chr.21: 33577680-33577680 on Build GRCh38
rs959442215 Chr.21: 33577657-33577657 on Build GRCh38
rs755800440 Chr.21: 33577864-33577864 on Build GRCh38
rs181896718 Chr.21: 33577817-33577817 on Build GRCh38
rs958069235 Chr.21: 33577547-33577547 on Build GRCh38
rs1052340666 Chr.21: 33577753-33577753 on Build GRCh38
rs766987443 Chr.21: 33577785-33577785 on Build GRCh38
rs978363605 Chr.21: 33577523-33577523 on Build GRCh38
rs999416317 Chr.21: 33577676-33577676 on Build GRCh38
rs112887522 Chr.21: 33577487-33577713 on Build GRCh38
rs369318446 Chr.21: 33577568-33577569 on Build GRCh38
rs3221082 Chr.21: 33577606-33577607 on Build GRCh38
rs200055284 Chr.21: 33577851-33577854 on Build GRCh38
rs865931901 Chr.21: 33577653-33577653 on Build GRCh38
rs998653107 Chr.21: 33577859-33577859 on Build GRCh38
rs148105472 Chr.21: 33577847-33577847 on Build GRCh38
rs999446025 Chr.21: 33577648-33577648 on Build GRCh38
rs754296642 Chr.21: 33577699-33577699 on Build GRCh38
rs866224381 Chr.21: 33577638-33577638 on Build GRCh38
rs989573478 Chr.21: 33577557-33577557 on Build GRCh38
rs930154684 Chr.21: 33577605-33577605 on Build GRCh38
rs941754338 Chr.21: 33577612-33577612 on Build GRCh38
rs1004202252 Chr.21: 33577872-33577872 on Build GRCh38
rs962843516 Chr.21: 33577706-33577706 on Build GRCh38
rs190151919 Chr.21: 33577782-33577782 on Build GRCh38
rs532520464 Chr.21: 33577749-33577749 on Build GRCh38
rs927597226 Chr.21: 33577734-33577734 on Build GRCh38
rs992106298 Chr.21: 33577719-33577719 on Build GRCh38
rs879662065 Chr.21: 33577650-33577650 on Build GRCh38
rs947400633 Chr.21: 33577582-33577582 on Build GRCh38
rs749883164 Chr.21: 33577830-33577830 on Build GRCh38
rs964517385 Chr.21: 33577691-33577691 on Build GRCh38
rs935009945 Chr.21: 33577739-33577739 on Build GRCh38
rs62227708 Chr.21: 33577630-33577630 on Build GRCh38
rs184112282 Chr.21: 33577701-33577701 on Build GRCh38
rs1012355090 Chr.21: 33577634-33577634 on Build GRCh38
rs868157092 Chr.21: 33577668-33577668 on Build GRCh38
rs1018800516 Chr.21: 33577656-33577656 on Build GRCh38
rs111856399 Chr.21: 33577861-33577861 on Build GRCh38
rs1030045353 Chr.21: 33577695-33577695 on Build GRCh38
rs563739505 Chr.21: 33577565-33577565 on Build GRCh38
rs1046831628 Chr.21: 33577778-33577778 on Build GRCh38
rs982366760 Chr.21: 33577483-33577485 on Build GRCh38
rs542330977 Chr.21: 33577539-33577539 on Build GRCh38
rs916350184 Chr.21: 33577670-33577670 on Build GRCh38
rs371118775 Chr.21: 33577662-33577662 on Build GRCh38
rs947761037 Chr.21: 33577732-33577732 on Build GRCh38
rs530005818 Chr.21: 33577600-33577600 on Build GRCh38
rs528314057 Chr.21: 33577720-33577720 on Build GRCh38
rs1003336764 Chr.21: 33577851-33577851 on Build GRCh38
rs528837004 Chr.21: 33577686-33577686 on Build GRCh38
rs868781852 Chr.21: 33577649-33577649 on Build GRCh38
rs565144365 Chr.21: 33577733-33577733 on Build GRCh38
rs2284559 Chr.21: 33577614-33577614 on Build GRCh38
rs868121359 Chr.21: 33577603-33577603 on Build GRCh38
rs1035743440 Chr.21: 33577518-33577519 on Build GRCh38
rs866885468 Chr.21: 33577644-33577644 on Build GRCh38
rs764272322 Chr.21: 33577567-33577568 on Build GRCh38
rs984013204 Chr.21: 33577707-33577735 on Build GRCh38
rs977194299 Chr.21: 33577659-33577659 on Build GRCh38
rs947743708 Chr.21: 33577671-33577671 on Build GRCh38
rs2284558 Chr.21: 33577610-33577610 on Build GRCh38
rs879673117 Chr.21: 33577701-33577702 on Build GRCh38
rs899675041 Chr.21: 33577820-33577820 on Build GRCh38
rs1005193067 Chr.21: 33577797-33577797 on Build GRCh38
rs774829318 Chr.21: 33577482-33577482 on Build GRCh38
rs893764491 Chr.21: 33577774-33577774 on Build GRCh38
rs764731922 Chr.21: 33577475-33577475 on Build GRCh38
rs1031089613 Chr.21: 33577544-33577544 on Build GRCh38
rs1025334862 Chr.21: 33577486-33577486 on Build GRCh38
rs900881116 Chr.21: 33577743-33577744 on Build GRCh38
rs957870843 Chr.21: 33577652-33577652 on Build GRCh38
rs568067672 Chr.21: 33577780-33577780 on Build GRCh38
rs964067492 Chr.21: 33577658-33577658 on Build GRCh38
rs530532324 Chr.21: 33577597-33577597 on Build GRCh38
rs3178051 Chr.21: 33577478-33577478 on Build GRCh38
rs749191459 Chr.21: 33577684-33577684 on Build GRCh38
rs887095771 Chr.21: 33577871-33577871 on Build GRCh38
rs867053378 Chr.21: 33577604-33577604 on Build GRCh38
rs13052962 Chr.21: 33577620-33577620 on Build GRCh38
rs906483386 Chr.21: 33577646-33577646 on Build GRCh38
rs941554552 Chr.21: 33577666-33577666 on Build GRCh38
rs1052370513 Chr.21: 33577678-33577678 on Build GRCh38
rs191526771 Chr.21: 33577697-33577697 on Build GRCh38
rs71194853 Chr.21: 33577691-33577702 on Build GRCh38
rs1018355093 Chr.21: 33577688-33577688 on Build GRCh38
rs1010775190 Chr.21: 33577685-33577685 on Build GRCh38
rs998286147 Chr.21: 33577693-33577693 on Build GRCh38
rs970573860 Chr.21: 33577515-33577515 on Build GRCh38

View Full Product Details