Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 34096360-34096870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPS6
Entrez Gene ID
64968
Gene Name
mitochondrial ribosomal protein S6
Gene Aliases
C21orf101, MRP-S6, RPMS6, S6mt
UniGene
Hs.302742
Gene Chromosome Location
Chr.21: 34073523-34143034 on Build GRCh38
Transcripts
NM_032476.3 NM_006933.6

Gene Symbol
SLC5A3
Entrez Gene ID
6526
Gene Name
solute carrier family 5 member 3
Gene Aliases
BCW2, SMIT, SMIT1, SMIT2
UniGene
Hs.302742
Gene Chromosome Location
Chr.21: 34073523-34106262 on Build GRCh38
Transcripts
NM_032476.3 NM_006933.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1413924 c.1508C>T p.P503L Chr.21: 34096706-34096706 on Build GRCh38
1413923 c.1282A>G p.I428V Chr.21: 34096480-34096480 on Build GRCh38
1413926 c.1589G>A p.S530N Chr.21: 34096787-34096787 on Build GRCh38
1413925 c.1565C>T p.T522M Chr.21: 34096763-34096763 on Build GRCh38
7421812 c.1423A>G p.M475V Chr.21: 34096621-34096621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915124504 Chr.21: 34096407-34096407 on Build GRCh38
rs569028619 Chr.21: 34096428-34096428 on Build GRCh38
rs769665990 Chr.21: 34096524-34096524 on Build GRCh38
rs764604067 Chr.21: 34096450-34096450 on Build GRCh38
rs745528477 Chr.21: 34096689-34096689 on Build GRCh38
rs753857258 Chr.21: 34096742-34096742 on Build GRCh38
rs575422533 Chr.21: 34096722-34096722 on Build GRCh38
rs557966542 Chr.21: 34096385-34096385 on Build GRCh38
rs372550004 Chr.21: 34096587-34096587 on Build GRCh38
rs777690400 Chr.21: 34096543-34096543 on Build GRCh38
rs117638044 Chr.21: 34096613-34096613 on Build GRCh38
rs568072128 Chr.21: 34096386-34096386 on Build GRCh38
rs757979616 Chr.21: 34096771-34096771 on Build GRCh38
rs756320010 Chr.21: 34096724-34096724 on Build GRCh38
rs778548781 Chr.21: 34096764-34096764 on Build GRCh38
rs752872402 Chr.21: 34096661-34096661 on Build GRCh38
rs1024914411 Chr.21: 34096668-34096668 on Build GRCh38
rs149914018 Chr.21: 34096833-34096833 on Build GRCh38
rs773718366 Chr.21: 34096708-34096708 on Build GRCh38
rs1030643107 Chr.21: 34096808-34096808 on Build GRCh38
rs755047769 Chr.21: 34096646-34096646 on Build GRCh38
rs765610386 Chr.21: 34096464-34096464 on Build GRCh38
rs777145221 Chr.21: 34096575-34096575 on Build GRCh38
rs749430501 Chr.21: 34096573-34096573 on Build GRCh38
rs368456491 Chr.21: 34096632-34096632 on Build GRCh38
rs762719099 Chr.21: 34096416-34096416 on Build GRCh38
rs757122598 Chr.21: 34096672-34096672 on Build GRCh38
rs778217292 Chr.21: 34096798-34096798 on Build GRCh38
rs768163299 Chr.21: 34096773-34096773 on Build GRCh38
rs774586769 Chr.21: 34096714-34096714 on Build GRCh38
rs780317618 Chr.21: 34096483-34096483 on Build GRCh38
rs759657838 Chr.21: 34096715-34096715 on Build GRCh38
rs764055960 Chr.21: 34096730-34096730 on Build GRCh38
rs774830116 Chr.21: 34096699-34096699 on Build GRCh38
rs751642021 Chr.21: 34096485-34096485 on Build GRCh38
rs370000793 Chr.21: 34096458-34096458 on Build GRCh38
rs1031683487 Chr.21: 34096629-34096629 on Build GRCh38
rs781054160 Chr.21: 34096396-34096396 on Build GRCh38
rs111299643 Chr.21: 34096666-34096666 on Build GRCh38
rs746093403 Chr.21: 34096393-34096393 on Build GRCh38
rs766689462 Chr.21: 34096713-34096713 on Build GRCh38
rs574501087 Chr.21: 34096669-34096669 on Build GRCh38
rs921041492 Chr.21: 34096431-34096431 on Build GRCh38
rs774983348 Chr.21: 34096706-34096706 on Build GRCh38
rs191539072 Chr.21: 34096673-34096673 on Build GRCh38
rs771803912 Chr.21: 34096695-34096695 on Build GRCh38
rs748070770 Chr.21: 34096506-34096506 on Build GRCh38
rs745713093 Chr.21: 34096770-34096770 on Build GRCh38
rs773888906 Chr.21: 34096550-34096550 on Build GRCh38
rs550905038 Chr.21: 34096411-34096411 on Build GRCh38
rs143365196 Chr.21: 34096394-34096394 on Build GRCh38
rs559001332 Chr.21: 34096551-34096551 on Build GRCh38
rs957745254 Chr.21: 34096826-34096826 on Build GRCh38
rs375427676 Chr.21: 34096755-34096755 on Build GRCh38
rs762123888 Chr.21: 34096461-34096461 on Build GRCh38
rs762214834 Chr.21: 34096707-34096707 on Build GRCh38
rs576211592 Chr.21: 34096590-34096590 on Build GRCh38
rs749819511 Chr.21: 34096802-34096802 on Build GRCh38
rs770757642 Chr.21: 34096425-34096425 on Build GRCh38
rs933707054 Chr.21: 34096437-34096437 on Build GRCh38
rs771521450 Chr.21: 34096820-34096820 on Build GRCh38
rs542069855 Chr.21: 34096670-34096670 on Build GRCh38
rs975097049 Chr.21: 34096767-34096767 on Build GRCh38
rs200641030 Chr.21: 34096391-34096391 on Build GRCh38
rs763240947 Chr.21: 34096711-34096711 on Build GRCh38
rs151207838 Chr.21: 34096647-34096647 on Build GRCh38
rs920946715 Chr.21: 34096774-34096774 on Build GRCh38
rs762283242 Chr.21: 34096576-34096576 on Build GRCh38
rs146893213 Chr.21: 34096602-34096602 on Build GRCh38
rs535716171 Chr.21: 34096401-34096401 on Build GRCh38
rs781120517 Chr.21: 34096494-34096494 on Build GRCh38
rs769874431 Chr.21: 34096447-34096447 on Build GRCh38
rs763361171 Chr.21: 34096599-34096599 on Build GRCh38
rs1039028792 Chr.21: 34096519-34096519 on Build GRCh38
rs772962068 Chr.21: 34096412-34096412 on Build GRCh38
rs779810969 Chr.21: 34096772-34096772 on Build GRCh38
rs770702731 Chr.21: 34096549-34096549 on Build GRCh38
rs767822490 Chr.21: 34096718-34096718 on Build GRCh38
rs780017963 Chr.21: 34096480-34096480 on Build GRCh38
rs749135471 Chr.21: 34096546-34096546 on Build GRCh38
rs897218283 Chr.21: 34096588-34096588 on Build GRCh38
rs1050807571 Chr.21: 34096465-34096465 on Build GRCh38
rs771512755 Chr.21: 34096555-34096555 on Build GRCh38
rs967980350 Chr.21: 34096746-34096746 on Build GRCh38

View Full Product Details