Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 40055632-40056170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSCAM
Entrez Gene ID
1826
Gene Name
DS cell adhesion molecule
Gene Aliases
CHD2, CHD2-42, CHD2-52
UniGene
Hs.397800
Gene Chromosome Location
Chr.21: 40010999-40847113 on Build GRCh38
Transcripts
NM_001271534.1 NM_001389.3 NR_073202.1 XM_017028281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5723177 c.4991G>A p.R1664K Chr.21: 40055769-40055769 on Build GRCh38
444494 c.5032A>G p.I1678V Chr.21: 40055728-40055728 on Build GRCh38
280614 c.5022G>A p.T1674T Chr.21: 40055738-40055738 on Build GRCh38
3550795 c.4970G>A p.R1657Q Chr.21: 40055790-40055790 on Build GRCh38
5798291 c.4923G>A p.K1641K Chr.21: 40055837-40055837 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544635744 Chr.21: 40055988-40055988 on Build GRCh38
rs533763779 Chr.21: 40055914-40055914 on Build GRCh38
rs544906142 Chr.21: 40055986-40055986 on Build GRCh38
rs748635134 Chr.21: 40055779-40055779 on Build GRCh38
rs982968350 Chr.21: 40055693-40055693 on Build GRCh38
rs112157214 Chr.21: 40055695-40055695 on Build GRCh38
rs115204168 Chr.21: 40055971-40055971 on Build GRCh38
rs73221349 Chr.21: 40055840-40055840 on Build GRCh38
rs925250311 Chr.21: 40055931-40055931 on Build GRCh38
rs931680035 Chr.21: 40055810-40055810 on Build GRCh38
rs749714856 Chr.21: 40055889-40055889 on Build GRCh38
rs779441338 Chr.21: 40055868-40055868 on Build GRCh38
rs756965040 Chr.21: 40055754-40055754 on Build GRCh38
rs897458213 Chr.21: 40055992-40055992 on Build GRCh38
rs778536164 Chr.21: 40055761-40055761 on Build GRCh38
rs766747740 Chr.21: 40055790-40055790 on Build GRCh38
rs765543937 Chr.21: 40055707-40055707 on Build GRCh38
rs755073776 Chr.21: 40055737-40055737 on Build GRCh38
rs986971232 Chr.21: 40056048-40056048 on Build GRCh38
rs751755553 Chr.21: 40055730-40055730 on Build GRCh38
rs200676569 Chr.21: 40055680-40055680 on Build GRCh38
rs917849698 Chr.21: 40056115-40056115 on Build GRCh38
rs772403484 Chr.21: 40055763-40055763 on Build GRCh38
rs2837400 Chr.21: 40055918-40055918 on Build GRCh38
rs1030078292 Chr.21: 40056020-40056020 on Build GRCh38
rs975609102 Chr.21: 40055998-40055998 on Build GRCh38
rs115907659 Chr.21: 40055953-40055953 on Build GRCh38
rs116520450 Chr.21: 40056042-40056042 on Build GRCh38
rs770276824 Chr.21: 40055780-40055780 on Build GRCh38
rs1023545206 Chr.21: 40055681-40055681 on Build GRCh38
rs746514289 Chr.21: 40055874-40055874 on Build GRCh38
rs1044924786 Chr.21: 40056129-40056129 on Build GRCh38
rs376791154 Chr.21: 40055845-40055845 on Build GRCh38
rs769450380 Chr.21: 40055684-40055684 on Build GRCh38
rs758009044 Chr.21: 40055866-40055866 on Build GRCh38
rs374386822 Chr.21: 40055816-40055816 on Build GRCh38
rs186875566 Chr.21: 40055657-40055657 on Build GRCh38
rs577566088 Chr.21: 40055987-40055987 on Build GRCh38
rs763371653 Chr.21: 40055787-40055787 on Build GRCh38
rs774689701 Chr.21: 40055795-40055795 on Build GRCh38
rs979162819 Chr.21: 40056050-40056050 on Build GRCh38
rs759685690 Chr.21: 40055739-40055739 on Build GRCh38
rs756021849 Chr.21: 40055829-40055829 on Build GRCh38
rs896349597 Chr.21: 40055896-40055896 on Build GRCh38
rs1028885711 Chr.21: 40056019-40056019 on Build GRCh38
rs971817573 Chr.21: 40055791-40055791 on Build GRCh38
rs764601633 Chr.21: 40055878-40055878 on Build GRCh38
rs762288928 Chr.21: 40055701-40055701 on Build GRCh38
rs185585502 Chr.21: 40056123-40056123 on Build GRCh38
rs777436692 Chr.21: 40055740-40055740 on Build GRCh38
rs905008294 Chr.21: 40056138-40056138 on Build GRCh38
rs748921226 Chr.21: 40055686-40055686 on Build GRCh38
rs988498997 Chr.21: 40056077-40056077 on Build GRCh38
rs141507048 Chr.21: 40055738-40055738 on Build GRCh38
rs750731548 Chr.21: 40055712-40055712 on Build GRCh38
rs924965643 Chr.21: 40056069-40056069 on Build GRCh38
rs571311891 Chr.21: 40055904-40055904 on Build GRCh38
rs755022707 Chr.21: 40055675-40055675 on Build GRCh38
rs771549137 Chr.21: 40055767-40055767 on Build GRCh38
rs905291149 Chr.21: 40055946-40055946 on Build GRCh38
rs925533319 Chr.21: 40056131-40056131 on Build GRCh38
rs773867869 Chr.21: 40055690-40055690 on Build GRCh38
rs777092357 Chr.21: 40055699-40055699 on Build GRCh38
rs773504976 Chr.21: 40055783-40055783 on Build GRCh38
rs1026958848 Chr.21: 40055957-40055957 on Build GRCh38
rs775018052 Chr.21: 40055776-40055776 on Build GRCh38
rs192527491 Chr.21: 40055678-40055678 on Build GRCh38
rs759036968 Chr.21: 40055692-40055692 on Build GRCh38
rs776145643 Chr.21: 40055774-40055774 on Build GRCh38
rs753647645 Chr.21: 40055837-40055837 on Build GRCh38
rs773738248 Chr.21: 40055881-40055881 on Build GRCh38
rs766694863 Chr.21: 40055721-40055721 on Build GRCh38
rs879053464 Chr.21: 40056024-40056024 on Build GRCh38
rs116176049 Chr.21: 40055970-40055970 on Build GRCh38
rs766886368 Chr.21: 40055725-40055725 on Build GRCh38
rs538166594 Chr.21: 40055834-40055834 on Build GRCh38
rs749056896 Chr.21: 40055753-40055753 on Build GRCh38
rs921708128 Chr.21: 40055809-40055809 on Build GRCh38
rs1015914796 Chr.21: 40055768-40055768 on Build GRCh38
rs542676642 Chr.21: 40055965-40055965 on Build GRCh38
rs752753543 Chr.21: 40055827-40055827 on Build GRCh38
rs934982225 Chr.21: 40056075-40056075 on Build GRCh38
rs935321275 Chr.21: 40055937-40055937 on Build GRCh38
rs567402114 Chr.21: 40055817-40055817 on Build GRCh38
rs76885144 Chr.21: 40055688-40055688 on Build GRCh38

View Full Product Details