Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 41480134-41480629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS2
Entrez Gene ID
7113
Gene Name
transmembrane protease, serine 2
Gene Aliases
PP9284, PRSS10
UniGene
Hs.439309
Gene Chromosome Location
Chr.21: 41464551-41508158 on Build GRCh38
Transcripts
NM_001135099.1 NM_005656.3 XM_005261043.2 XM_011529731.2 XM_011529732.2 XM_011529733.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3758884 c.589G>A p.V197M Chr.21: 41480570-41480570 on Build GRCh38
6976371 c.568G>A p.G190R Chr.21: 41480591-41480591 on Build GRCh38
6929722 c.604A>T p.R202W Chr.21: 41480555-41480555 on Build GRCh38
6045048 c.554G>T p.C185F Chr.21: 41480494-41480494 on Build GRCh38
6977320 c.653G>A p.G218E Chr.21: 41480506-41480506 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144898971 Chr.21: 41480578-41480578 on Build GRCh38
rs903273822 Chr.21: 41480503-41480503 on Build GRCh38
rs766503231 Chr.21: 41480591-41480591 on Build GRCh38
rs748560513 Chr.21: 41480600-41480600 on Build GRCh38
rs752474554 Chr.21: 41480483-41480483 on Build GRCh38
rs541387461 Chr.21: 41480388-41480388 on Build GRCh38
rs376897988 Chr.21: 41480213-41480213 on Build GRCh38
rs36089629 Chr.21: 41480184-41480185 on Build GRCh38
rs950437284 Chr.21: 41480399-41480399 on Build GRCh38
rs548725701 Chr.21: 41480181-41480181 on Build GRCh38
rs61735789 Chr.21: 41480508-41480508 on Build GRCh38
rs543617195 Chr.21: 41480157-41480157 on Build GRCh38
rs734056 Chr.21: 41480393-41480393 on Build GRCh38
rs1035659632 Chr.21: 41480331-41480331 on Build GRCh38
rs776238499 Chr.21: 41480466-41480466 on Build GRCh38
rs762840890 Chr.21: 41480487-41480487 on Build GRCh38
rs980351849 Chr.21: 41480264-41480264 on Build GRCh38
rs143109187 Chr.21: 41480259-41480259 on Build GRCh38
rs780957271 Chr.21: 41480359-41480359 on Build GRCh38
rs1017949697 Chr.21: 41480180-41480180 on Build GRCh38
rs772335956 Chr.21: 41480315-41480315 on Build GRCh38
rs748571451 Chr.21: 41480525-41480525 on Build GRCh38
rs777857090 Chr.21: 41480383-41480383 on Build GRCh38
rs973116103 Chr.21: 41480203-41480203 on Build GRCh38
rs999855497 Chr.21: 41480457-41480457 on Build GRCh38
rs915100646 Chr.21: 41480491-41480491 on Build GRCh38
rs563209762 Chr.21: 41480537-41480537 on Build GRCh38
rs734055 Chr.21: 41480243-41480243 on Build GRCh38
rs1009173595 Chr.21: 41480406-41480406 on Build GRCh38
rs774013018 Chr.21: 41480535-41480535 on Build GRCh38
rs551277697 Chr.21: 41480224-41480224 on Build GRCh38
rs749577439 Chr.21: 41480514-41480514 on Build GRCh38
rs1032644092 Chr.21: 41480460-41480460 on Build GRCh38
rs566925007 Chr.21: 41480332-41480332 on Build GRCh38
rs372223232 Chr.21: 41480453-41480453 on Build GRCh38
rs371583288 Chr.21: 41480592-41480592 on Build GRCh38
rs764043495 Chr.21: 41480499-41480499 on Build GRCh38
rs780256128 Chr.21: 41480513-41480513 on Build GRCh38
rs759250613 Chr.21: 41480554-41480554 on Build GRCh38
rs763953663 Chr.21: 41480550-41480550 on Build GRCh38
rs546790067 Chr.21: 41480293-41480293 on Build GRCh38
rs1057445884 Chr.21: 41480533-41480533 on Build GRCh38
rs770323541 Chr.21: 41480459-41480459 on Build GRCh38
rs755231989 Chr.21: 41480593-41480593 on Build GRCh38
rs918665794 Chr.21: 41480386-41480386 on Build GRCh38
rs764956413 Chr.21: 41480471-41480471 on Build GRCh38
rs186734573 Chr.21: 41480438-41480438 on Build GRCh38
rs762438307 Chr.21: 41480456-41480456 on Build GRCh38
rs1023216142 Chr.21: 41480583-41480583 on Build GRCh38
rs34561135 Chr.21: 41480384-41480384 on Build GRCh38
rs767499499 Chr.21: 41480565-41480565 on Build GRCh38
rs370312550 Chr.21: 41480343-41480343 on Build GRCh38
rs757262240 Chr.21: 41480504-41480504 on Build GRCh38
rs79397218 Chr.21: 41480455-41480455 on Build GRCh38
rs1023565990 Chr.21: 41480366-41480366 on Build GRCh38
rs899376787 Chr.21: 41480594-41480594 on Build GRCh38
rs12329760 Chr.21: 41480570-41480570 on Build GRCh38
rs779200981 Chr.21: 41480599-41480599 on Build GRCh38
rs376158219 Chr.21: 41480526-41480526 on Build GRCh38
rs562078893 Chr.21: 41480559-41480559 on Build GRCh38
rs762745808 Chr.21: 41480538-41480538 on Build GRCh38
rs768417140 Chr.21: 41480229-41480229 on Build GRCh38
rs946529846 Chr.21: 41480494-41480494 on Build GRCh38
rs370175310 Chr.21: 41480468-41480468 on Build GRCh38
rs1032758518 Chr.21: 41480219-41480219 on Build GRCh38
rs751523924 Chr.21: 41480500-41480500 on Build GRCh38
rs781089181 Chr.21: 41480507-41480507 on Build GRCh38
rs1038932296 Chr.21: 41480426-41480426 on Build GRCh38
rs998847649 Chr.21: 41480326-41480326 on Build GRCh38
rs767345891 Chr.21: 41480564-41480564 on Build GRCh38
rs759090681 Chr.21: 41480529-41480529 on Build GRCh38
rs139181746 Chr.21: 41480207-41480207 on Build GRCh38
rs780343945 Chr.21: 41480425-41480425 on Build GRCh38
rs751428160 Chr.21: 41480557-41480557 on Build GRCh38
rs987441905 Chr.21: 41480228-41480228 on Build GRCh38
rs947481195 Chr.21: 41480277-41480277 on Build GRCh38
rs756213944 Chr.21: 41480577-41480577 on Build GRCh38
rs373847134 Chr.21: 41480519-41480519 on Build GRCh38
rs943261513 Chr.21: 41480230-41480230 on Build GRCh38
rs941354966 Chr.21: 41480364-41480364 on Build GRCh38
rs373852195 Chr.21: 41480389-41480389 on Build GRCh38
rs200120702 Chr.21: 41480517-41480517 on Build GRCh38
rs139092674 Chr.21: 41480516-41480516 on Build GRCh38

View Full Product Details