Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 41800875-41801383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDM15
Entrez Gene ID
63977
Gene Name
PR domain 15
Gene Aliases
C21orf83, PFM15, ZNF298
UniGene
Hs.473893
Gene Chromosome Location
Chr.21: 41798225-41879482 on Build GRCh38
Transcripts
NM_001040424.2 NM_001282934.1 NM_022115.4 NR_104257.1 NR_104258.1 NR_104260.1 NR_135464.1 XM_006724039.2 XM_006724040.2 XM_011529674.1 XM_011529675.1 XM_011529676.2 XM_011529677.2 XM_011529678.2 XM_011529679.2 XM_011529681.2 XM_011529683.1 XM_017028425.1 XM_017028426.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6755040 c.4442C>T p.S1481L Chr.21: 41801322-41801322 on Build GRCh38
7294886 c.4443G>A p.S1481S Chr.21: 41801321-41801321 on Build GRCh38
6755045 c.4446G>C p.Q1482H Chr.21: 41801318-41801318 on Build GRCh38
7292387 c.4398G>A p.P1466P Chr.21: 41801366-41801366 on Build GRCh38
2819074 c.4449G>A p.P1483P Chr.21: 41801315-41801315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751836847 Chr.21: 41801321-41801321 on Build GRCh38
rs146149904 Chr.21: 41801195-41801195 on Build GRCh38
rs748507496 Chr.21: 41801255-41801260 on Build GRCh38
rs760560166 Chr.21: 41801301-41801301 on Build GRCh38
rs769163221 Chr.21: 41801254-41801255 on Build GRCh38
rs748700445 Chr.21: 41801255-41801255 on Build GRCh38
rs776365265 Chr.21: 41801361-41801361 on Build GRCh38
rs993355524 Chr.21: 41800964-41800964 on Build GRCh38
rs534120191 Chr.21: 41800973-41800973 on Build GRCh38
rs564585389 Chr.21: 41801221-41801221 on Build GRCh38
rs147616045 Chr.21: 41801274-41801274 on Build GRCh38
rs546890788 Chr.21: 41801285-41801285 on Build GRCh38
rs779463468 Chr.21: 41801235-41801236 on Build GRCh38
rs3850706 Chr.21: 41801323-41801323 on Build GRCh38
rs142036110 Chr.21: 41801294-41801294 on Build GRCh38
rs191960107 Chr.21: 41801095-41801095 on Build GRCh38
rs994776296 Chr.21: 41801105-41801105 on Build GRCh38
rs1020937058 Chr.21: 41800960-41800960 on Build GRCh38
rs746731880 Chr.21: 41801358-41801358 on Build GRCh38
rs766408138 Chr.21: 41801214-41801214 on Build GRCh38
rs1042151402 Chr.21: 41800979-41800979 on Build GRCh38
rs899992214 Chr.21: 41800926-41800926 on Build GRCh38
rs562620889 Chr.21: 41801109-41801109 on Build GRCh38
rs778292224 Chr.21: 41801230-41801230 on Build GRCh38
rs915405077 Chr.21: 41801347-41801347 on Build GRCh38
rs931051260 Chr.21: 41801072-41801072 on Build GRCh38
rs757665530 Chr.21: 41801322-41801322 on Build GRCh38
rs771684064 Chr.21: 41801353-41801353 on Build GRCh38
rs1035982521 Chr.21: 41800986-41800986 on Build GRCh38
rs758765575 Chr.21: 41801229-41801229 on Build GRCh38
rs773458140 Chr.21: 41801328-41801345 on Build GRCh38
rs200853879 Chr.21: 41801360-41801360 on Build GRCh38
rs950825484 Chr.21: 41801114-41801114 on Build GRCh38
rs768998241 Chr.21: 41801273-41801273 on Build GRCh38
rs943257599 Chr.21: 41800927-41800927 on Build GRCh38
rs573784614 Chr.21: 41801080-41801080 on Build GRCh38
rs914891383 Chr.21: 41800943-41800943 on Build GRCh38
rs919773772 Chr.21: 41801012-41801012 on Build GRCh38
rs752998031 Chr.21: 41801224-41801224 on Build GRCh38
rs753832336 Chr.21: 41801217-41801217 on Build GRCh38
rs974443984 Chr.21: 41800929-41800929 on Build GRCh38
rs768190050 Chr.21: 41801258-41801258 on Build GRCh38
rs754431165 Chr.21: 41801335-41801335 on Build GRCh38
rs749476959 Chr.21: 41801351-41801351 on Build GRCh38
rs974643702 Chr.21: 41801199-41801199 on Build GRCh38
rs991257122 Chr.21: 41801005-41801005 on Build GRCh38
rs778382718 Chr.21: 41801336-41801336 on Build GRCh38
rs758675738 Chr.21: 41801317-41801317 on Build GRCh38
rs1019190319 Chr.21: 41801136-41801136 on Build GRCh38
rs776651379 Chr.21: 41801314-41801314 on Build GRCh38
rs148832213 Chr.21: 41801355-41801355 on Build GRCh38
rs1040721139 Chr.21: 41801097-41801097 on Build GRCh38
rs558209153 Chr.21: 41800942-41800942 on Build GRCh38
rs17113941 Chr.21: 41801079-41801079 on Build GRCh38
rs902222490 Chr.21: 41800995-41800995 on Build GRCh38
rs747462595 Chr.21: 41801234-41801234 on Build GRCh38
rs987354959 Chr.21: 41801125-41801125 on Build GRCh38
rs745379477 Chr.21: 41801040-41801040 on Build GRCh38
rs747724242 Chr.21: 41801341-41801341 on Build GRCh38
rs750318074 Chr.21: 41801194-41801194 on Build GRCh38
rs894004133 Chr.21: 41801167-41801167 on Build GRCh38
rs149697369 Chr.21: 41801316-41801316 on Build GRCh38
rs773930882 Chr.21: 41801267-41801267 on Build GRCh38
rs964646795 Chr.21: 41801186-41801186 on Build GRCh38
rs766202873 Chr.21: 41801308-41801308 on Build GRCh38
rs771662285 Chr.21: 41801090-41801090 on Build GRCh38
rs958616806 Chr.21: 41801001-41801001 on Build GRCh38
rs746395639 Chr.21: 41801333-41801333 on Build GRCh38
rs759556472 Chr.21: 41801363-41801363 on Build GRCh38
rs749735986 Chr.21: 41801255-41801257 on Build GRCh38
rs934853818 Chr.21: 41801011-41801011 on Build GRCh38
rs9983923 Chr.21: 41800951-41800951 on Build GRCh38
rs1026399533 Chr.21: 41801111-41801111 on Build GRCh38
rs1057481962 Chr.21: 41801046-41801046 on Build GRCh38
rs772462348 Chr.21: 41801306-41801323 on Build GRCh38
rs562141560 Chr.21: 41800920-41800920 on Build GRCh38
rs1009685772 Chr.21: 41800955-41800955 on Build GRCh38
rs755603804 Chr.21: 41801233-41801234 on Build GRCh38
rs866462339 Chr.21: 41800913-41800913 on Build GRCh38
rs878918451 Chr.21: 41801289-41801289 on Build GRCh38
rs781749065 Chr.21: 41801249-41801249 on Build GRCh38
rs759648530 Chr.21: 41801315-41801315 on Build GRCh38
rs544583763 Chr.21: 41801220-41801220 on Build GRCh38
rs771335120 Chr.21: 41801235-41801235 on Build GRCh38
rs764440851 Chr.21: 41801319-41801319 on Build GRCh38
rs35395118 Chr.21: 41800959-41800960 on Build GRCh38

View Full Product Details