Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 41801243-41801742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDM15
Entrez Gene ID
63977
Gene Name
PR domain 15
Gene Aliases
C21orf83, PFM15, ZNF298
UniGene
Hs.473893
Gene Chromosome Location
Chr.21: 41798225-41879482 on Build GRCh38
Transcripts
NM_001040424.2 NM_001282934.1 NM_022115.4 NR_104257.1 NR_104258.1 NR_104260.1 NR_135464.1 XM_006724039.2 XM_006724040.2 XM_011529674.1 XM_011529675.1 XM_011529676.2 XM_011529677.2 XM_011529678.2 XM_011529679.2 XM_011529681.2 XM_011529683.1 XM_017028425.1 XM_017028426.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5057598 c.4132G>A p.A1378T Chr.21: 41801632-41801632 on Build GRCh38
2819074 c.4449G>A p.P1483P Chr.21: 41801315-41801315 on Build GRCh38
5858657 c.3469_3471delCAG p.Q1157delQ Chr.21: 41801255-41801257 on Build GRCh38
149230 c.4369C>T p.L1457L Chr.21: 41801395-41801395 on Build GRCh38
6755046 c.3408G>C p.Q1136H Chr.21: 41801318-41801318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751836847 Chr.21: 41801321-41801321 on Build GRCh38
rs757779104 Chr.21: 41801481-41801481 on Build GRCh38
rs760560166 Chr.21: 41801301-41801301 on Build GRCh38
rs780074101 Chr.21: 41801519-41801519 on Build GRCh38
rs77715313 Chr.21: 41801456-41801456 on Build GRCh38
rs776365265 Chr.21: 41801361-41801361 on Build GRCh38
rs546890788 Chr.21: 41801285-41801285 on Build GRCh38
rs571676023 Chr.21: 41801421-41801421 on Build GRCh38
rs3850706 Chr.21: 41801323-41801323 on Build GRCh38
rs746731880 Chr.21: 41801358-41801358 on Build GRCh38
rs376297844 Chr.21: 41801649-41801649 on Build GRCh38
rs994853464 Chr.21: 41801378-41801378 on Build GRCh38
rs757665530 Chr.21: 41801322-41801322 on Build GRCh38
rs750015279 Chr.21: 41801681-41801681 on Build GRCh38
rs771199735 Chr.21: 41801659-41801659 on Build GRCh38
rs751840680 Chr.21: 41801374-41801374 on Build GRCh38
rs142104135 Chr.21: 41801660-41801660 on Build GRCh38
rs772316949 Chr.21: 41801571-41801571 on Build GRCh38
rs764175753 Chr.21: 41801369-41801369 on Build GRCh38
rs746677961 Chr.21: 41801408-41801408 on Build GRCh38
rs754431165 Chr.21: 41801335-41801335 on Build GRCh38
rs370290707 Chr.21: 41801653-41801653 on Build GRCh38
rs758675738 Chr.21: 41801317-41801317 on Build GRCh38
rs774485148 Chr.21: 41801549-41801549 on Build GRCh38
rs760986641 Chr.21: 41801586-41801586 on Build GRCh38
rs373004514 Chr.21: 41801466-41801466 on Build GRCh38
rs769745019 Chr.21: 41801518-41801518 on Build GRCh38
rs147459914 Chr.21: 41801603-41801603 on Build GRCh38
rs747724242 Chr.21: 41801341-41801341 on Build GRCh38
rs116436673 Chr.21: 41801420-41801420 on Build GRCh38
rs745633894 Chr.21: 41801503-41801503 on Build GRCh38
rs906586443 Chr.21: 41801372-41801372 on Build GRCh38
rs773930882 Chr.21: 41801267-41801267 on Build GRCh38
rs757500912 Chr.21: 41801379-41801379 on Build GRCh38
rs746395639 Chr.21: 41801333-41801333 on Build GRCh38
rs759556472 Chr.21: 41801363-41801363 on Build GRCh38
rs772462348 Chr.21: 41801306-41801323 on Build GRCh38
rs36053419 Chr.21: 41801493-41801493 on Build GRCh38
rs745719904 Chr.21: 41801423-41801423 on Build GRCh38
rs372779699 Chr.21: 41801628-41801628 on Build GRCh38
rs761840152 Chr.21: 41801568-41801568 on Build GRCh38
rs768787999 Chr.21: 41801447-41801447 on Build GRCh38
rs778765604 Chr.21: 41801633-41801633 on Build GRCh38
rs755827937 Chr.21: 41801718-41801718 on Build GRCh38
rs74706304 Chr.21: 41801656-41801656 on Build GRCh38
rs766824100 Chr.21: 41801465-41801465 on Build GRCh38
rs760073923 Chr.21: 41801599-41801599 on Build GRCh38
rs769820638 Chr.21: 41801425-41801425 on Build GRCh38
rs781029285 Chr.21: 41801497-41801497 on Build GRCh38
rs2236696 Chr.21: 41801666-41801666 on Build GRCh38
rs766733881 Chr.21: 41801589-41801589 on Build GRCh38
rs147616045 Chr.21: 41801274-41801274 on Build GRCh38
rs780581037 Chr.21: 41801399-41801399 on Build GRCh38
rs768538770 Chr.21: 41801540-41801540 on Build GRCh38
rs777355181 Chr.21: 41801485-41801485 on Build GRCh38
rs142036110 Chr.21: 41801294-41801294 on Build GRCh38
rs112998549 Chr.21: 41801654-41801654 on Build GRCh38
rs1018807517 Chr.21: 41801411-41801411 on Build GRCh38
rs61738718 Chr.21: 41801672-41801672 on Build GRCh38
rs960518024 Chr.21: 41801480-41801480 on Build GRCh38
rs145309149 Chr.21: 41801634-41801634 on Build GRCh38
rs915405077 Chr.21: 41801347-41801347 on Build GRCh38
rs138992396 Chr.21: 41801402-41801402 on Build GRCh38
rs139717863 Chr.21: 41801629-41801629 on Build GRCh38
rs771684064 Chr.21: 41801353-41801353 on Build GRCh38
rs763162674 Chr.21: 41801367-41801367 on Build GRCh38
rs369599731 Chr.21: 41801366-41801366 on Build GRCh38
rs773458140 Chr.21: 41801328-41801345 on Build GRCh38
rs200853879 Chr.21: 41801360-41801360 on Build GRCh38
rs138426773 Chr.21: 41801451-41801451 on Build GRCh38
rs768998241 Chr.21: 41801273-41801273 on Build GRCh38
rs369361257 Chr.21: 41801627-41801627 on Build GRCh38
rs111288587 Chr.21: 41801486-41801486 on Build GRCh38
rs750755650 Chr.21: 41801457-41801457 on Build GRCh38
rs2236694 Chr.21: 41801395-41801395 on Build GRCh38
rs556113305 Chr.21: 41801578-41801578 on Build GRCh38
rs149361413 Chr.21: 41801539-41801539 on Build GRCh38
rs749476959 Chr.21: 41801351-41801351 on Build GRCh38
rs376196876 Chr.21: 41801609-41801609 on Build GRCh38
rs778382718 Chr.21: 41801336-41801336 on Build GRCh38
rs544746085 Chr.21: 41801613-41801613 on Build GRCh38
rs776651379 Chr.21: 41801314-41801314 on Build GRCh38
rs148832213 Chr.21: 41801355-41801355 on Build GRCh38
rs61740056 Chr.21: 41801645-41801645 on Build GRCh38
rs573723961 Chr.21: 41801470-41801470 on Build GRCh38
rs763090405 Chr.21: 41801439-41801439 on Build GRCh38
rs149697369 Chr.21: 41801316-41801316 on Build GRCh38
rs2236695 Chr.21: 41801637-41801637 on Build GRCh38
rs766202873 Chr.21: 41801308-41801308 on Build GRCh38
rs538385630 Chr.21: 41801400-41801400 on Build GRCh38
rs145764491 Chr.21: 41801435-41801435 on Build GRCh38
rs761322637 Chr.21: 41801671-41801671 on Build GRCh38
rs756926245 Chr.21: 41801496-41801496 on Build GRCh38
rs762085177 Chr.21: 41801454-41801454 on Build GRCh38
rs564501415 Chr.21: 41801630-41801630 on Build GRCh38
rs150648109 Chr.21: 41801459-41801459 on Build GRCh38
rs996045746 Chr.21: 41801682-41801682 on Build GRCh38
rs767941572 Chr.21: 41801392-41801392 on Build GRCh38
rs765750235 Chr.21: 41801665-41801665 on Build GRCh38
rs151182384 Chr.21: 41801663-41801663 on Build GRCh38
rs878918451 Chr.21: 41801289-41801289 on Build GRCh38
rs759648530 Chr.21: 41801315-41801315 on Build GRCh38
rs754284282 Chr.21: 41801591-41801591 on Build GRCh38
rs764440851 Chr.21: 41801319-41801319 on Build GRCh38
rs750845569 Chr.21: 41801394-41801394 on Build GRCh38

View Full Product Details