Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 43800775-43801269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRP1
Entrez Gene ID
8568
Gene Name
ribosomal RNA processing 1
Gene Aliases
D21S2056E, NNP-1, NOP52, RRP1A
UniGene
Hs.110757
Gene Chromosome Location
Chr.21: 43789537-43804102 on Build GRCh38
Transcripts
NM_003683.5 XM_017028485.1 XM_017028486.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1632508 c.1009+9C>T p.? Chr.21: 43800890-43800890 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs933059380 Chr.21: 43800931-43800931 on Build GRCh38
rs146860814 Chr.21: 43801000-43801000 on Build GRCh38
rs758441464 Chr.21: 43800816-43800816 on Build GRCh38
rs933170721 Chr.21: 43801206-43801206 on Build GRCh38
rs762106236 Chr.21: 43800845-43800845 on Build GRCh38
rs200895005 Chr.21: 43800847-43800847 on Build GRCh38
rs866034902 Chr.21: 43800803-43800803 on Build GRCh38
rs749501572 Chr.21: 43800921-43800921 on Build GRCh38
rs769045299 Chr.21: 43800833-43800833 on Build GRCh38
rs555831888 Chr.21: 43801233-43801233 on Build GRCh38
rs749697728 Chr.21: 43800832-43800832 on Build GRCh38
rs777299630 Chr.21: 43801205-43801205 on Build GRCh38
rs201331848 Chr.21: 43800831-43800831 on Build GRCh38
rs774781312 Chr.21: 43800838-43800838 on Build GRCh38
rs1021083243 Chr.21: 43801031-43801031 on Build GRCh38
rs973283452 Chr.21: 43800805-43800807 on Build GRCh38
rs749649889 Chr.21: 43800865-43800865 on Build GRCh38
rs781154494 Chr.21: 43800904-43800904 on Build GRCh38
rs555723571 Chr.21: 43801155-43801155 on Build GRCh38
rs755544013 Chr.21: 43801029-43801029 on Build GRCh38
rs891614478 Chr.21: 43800926-43800926 on Build GRCh38
rs943073452 Chr.21: 43800994-43800994 on Build GRCh38
rs199714273 Chr.21: 43800896-43800896 on Build GRCh38
rs111742469 Chr.21: 43801156-43801156 on Build GRCh38
rs563988937 Chr.21: 43801187-43801187 on Build GRCh38
rs374992641 Chr.21: 43800813-43800813 on Build GRCh38
rs943086850 Chr.21: 43801234-43801234 on Build GRCh38
rs547821265 Chr.21: 43800971-43800971 on Build GRCh38
rs777609719 Chr.21: 43800817-43800817 on Build GRCh38
rs922946822 Chr.21: 43801157-43801157 on Build GRCh38
rs563938706 Chr.21: 43801203-43801203 on Build GRCh38
rs183721433 Chr.21: 43801019-43801019 on Build GRCh38
rs749056950 Chr.21: 43801211-43801211 on Build GRCh38
rs768972164 Chr.21: 43800925-43800925 on Build GRCh38
rs1033795156 Chr.21: 43801094-43801094 on Build GRCh38
rs552360721 Chr.21: 43800890-43800890 on Build GRCh38
rs752044959 Chr.21: 43801017-43801017 on Build GRCh38
rs762374598 Chr.21: 43800841-43800841 on Build GRCh38
rs541801631 Chr.21: 43800810-43800810 on Build GRCh38
rs558354171 Chr.21: 43801098-43801098 on Build GRCh38
rs149818702 Chr.21: 43800799-43800799 on Build GRCh38
rs781268090 Chr.21: 43800827-43800827 on Build GRCh38
rs984255576 Chr.21: 43801121-43801121 on Build GRCh38
rs1042918744 Chr.21: 43801210-43801210 on Build GRCh38
rs757162272 Chr.21: 43800900-43800900 on Build GRCh38
rs1035708010 Chr.21: 43801003-43801003 on Build GRCh38
rs758175650 Chr.21: 43800892-43800892 on Build GRCh38
rs373963641 Chr.21: 43801154-43801154 on Build GRCh38
rs960337576 Chr.21: 43801008-43801008 on Build GRCh38
rs371997224 Chr.21: 43800895-43800895 on Build GRCh38
rs1053467329 Chr.21: 43801242-43801242 on Build GRCh38
rs894076590 Chr.21: 43801006-43801006 on Build GRCh38
rs755141251 Chr.21: 43800841-43800842 on Build GRCh38
rs758692824 Chr.21: 43800932-43800932 on Build GRCh38
rs140711499 Chr.21: 43801133-43801133 on Build GRCh38
rs114739249 Chr.21: 43800905-43800905 on Build GRCh38
rs759280639 Chr.21: 43800859-43800859 on Build GRCh38
rs560395660 Chr.21: 43800824-43800824 on Build GRCh38
rs780307424 Chr.21: 43800917-43800917 on Build GRCh38
rs972996283 Chr.21: 43801027-43801027 on Build GRCh38
rs537584360 Chr.21: 43801188-43801188 on Build GRCh38
rs368121544 Chr.21: 43800823-43800823 on Build GRCh38
rs780644754 Chr.21: 43801050-43801050 on Build GRCh38
rs2329576 Chr.21: 43800906-43800906 on Build GRCh38
rs192661788 Chr.21: 43800894-43800894 on Build GRCh38
rs535969042 Chr.21: 43800928-43800928 on Build GRCh38
rs906748243 Chr.21: 43801036-43801036 on Build GRCh38
rs776219551 Chr.21: 43800853-43800853 on Build GRCh38
rs535840216 Chr.21: 43800958-43800958 on Build GRCh38
rs910321149 Chr.21: 43801221-43801221 on Build GRCh38
rs905949947 Chr.21: 43800995-43800995 on Build GRCh38
rs865808723 Chr.21: 43801005-43801005 on Build GRCh38
rs970795525 Chr.21: 43800828-43800828 on Build GRCh38
rs1017320276 Chr.21: 43801163-43801163 on Build GRCh38
rs572330031 Chr.21: 43801072-43801072 on Build GRCh38
rs752366383 Chr.21: 43800891-43800891 on Build GRCh38

View Full Product Details