Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44074246-44074757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAPPC10
Entrez Gene ID
7109
Gene Name
trafficking protein particle complex 10
Gene Aliases
EHOC-1, EHOC1, GT334, TMEM1, TRS130, TRS30
UniGene
Hs.126221
Gene Chromosome Location
Chr.21: 44012325-44106552 on Build GRCh38
Transcripts
NM_003274.4 XM_005261168.4 XM_011529714.2 XM_011529715.2 XM_011529716.2 XM_011529717.2 XM_011529718.2 XM_011529719.2 XM_011529720.2 XM_011529721.2 XM_011529722.2 XM_011529723.2 XM_011529725.2 XM_011529726.2 XM_011529727.2 XM_011529728.2 XM_011529729.2 XM_011529730.2 XM_017028454.1 XM_017028455.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1632509 c.1138T>G p.S380A Chr.21: 44074423-44074423 on Build GRCh38
4101892 c.1160G>T p.G387V Chr.21: 44074445-44074445 on Build GRCh38
3732337 c.1159G>T p.G387W Chr.21: 44074444-44074444 on Build GRCh38
5392997 c.1134C>T p.I378I Chr.21: 44074419-44074419 on Build GRCh38
5858721 c.1072G>A p.V358M Chr.21: 44074357-44074357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755356650 Chr.21: 44074499-44074499 on Build GRCh38
rs761480668 Chr.21: 44074614-44074615 on Build GRCh38
rs778305787 Chr.21: 44074293-44074293 on Build GRCh38
rs145786098 Chr.21: 44074576-44074576 on Build GRCh38
rs144074948 Chr.21: 44074409-44074409 on Build GRCh38
rs779405137 Chr.21: 44074412-44074412 on Build GRCh38
rs529220159 Chr.21: 44074718-44074718 on Build GRCh38
rs745439012 Chr.21: 44074508-44074508 on Build GRCh38
rs1020467210 Chr.21: 44074533-44074533 on Build GRCh38
rs918682946 Chr.21: 44074658-44074658 on Build GRCh38
rs998255320 Chr.21: 44074273-44074273 on Build GRCh38
rs143514819 Chr.21: 44074722-44074722 on Build GRCh38
rs779869749 Chr.21: 44074510-44074510 on Build GRCh38
rs774372325 Chr.21: 44074460-44074460 on Build GRCh38
rs560661863 Chr.21: 44074615-44074615 on Build GRCh38
rs867900349 Chr.21: 44074380-44074380 on Build GRCh38
rs563444965 Chr.21: 44074613-44074613 on Build GRCh38
rs771966857 Chr.21: 44074521-44074521 on Build GRCh38
rs17004626 Chr.21: 44074305-44074305 on Build GRCh38
rs193063951 Chr.21: 44074408-44074408 on Build GRCh38
rs199640878 Chr.21: 44074432-44074432 on Build GRCh38
rs1055057348 Chr.21: 44074433-44074433 on Build GRCh38
rs778332550 Chr.21: 44074507-44074507 on Build GRCh38
rs767462894 Chr.21: 44074474-44074474 on Build GRCh38
rs958651163 Chr.21: 44074591-44074591 on Build GRCh38
rs545072822 Chr.21: 44074590-44074590 on Build GRCh38
rs974154827 Chr.21: 44074653-44074653 on Build GRCh38
rs769365241 Chr.21: 44074509-44074509 on Build GRCh38
rs755495651 Chr.21: 44074604-44074604 on Build GRCh38
rs756211847 Chr.21: 44074482-44074482 on Build GRCh38
rs557906343 Chr.21: 44074319-44074319 on Build GRCh38
rs111399180 Chr.21: 44074649-44074649 on Build GRCh38
rs751477354 Chr.21: 44074320-44074320 on Build GRCh38
rs144085894 Chr.21: 44074339-44074339 on Build GRCh38
rs776045087 Chr.21: 44074419-44074419 on Build GRCh38
rs1029720085 Chr.21: 44074516-44074516 on Build GRCh38
rs930088202 Chr.21: 44074714-44074714 on Build GRCh38
rs992460648 Chr.21: 44074627-44074627 on Build GRCh38
rs767450860 Chr.21: 44074338-44074338 on Build GRCh38
rs781483680 Chr.21: 44074274-44074274 on Build GRCh38
rs981296424 Chr.21: 44074362-44074362 on Build GRCh38
rs761619583 Chr.21: 44074462-44074462 on Build GRCh38
rs941426583 Chr.21: 44074651-44074651 on Build GRCh38
rs200239699 Chr.21: 44074285-44074285 on Build GRCh38
rs766590796 Chr.21: 44074483-44074483 on Build GRCh38
rs757376416 Chr.21: 44074336-44074336 on Build GRCh38
rs763927095 Chr.21: 44074457-44074457 on Build GRCh38
rs748903488 Chr.21: 44074511-44074511 on Build GRCh38
rs768606992 Chr.21: 44074512-44074512 on Build GRCh38
rs957151459 Chr.21: 44074310-44074310 on Build GRCh38
rs555772453 Chr.21: 44074435-44074435 on Build GRCh38
rs367596961 Chr.21: 44074461-44074461 on Build GRCh38
rs145499248 Chr.21: 44074630-44074630 on Build GRCh38
rs758879039 Chr.21: 44074503-44074503 on Build GRCh38
rs754563228 Chr.21: 44074473-44074474 on Build GRCh38
rs114213621 Chr.21: 44074629-44074629 on Build GRCh38
rs761686846 Chr.21: 44074577-44074577 on Build GRCh38
rs184195379 Chr.21: 44074485-44074485 on Build GRCh38
rs1011355017 Chr.21: 44074529-44074529 on Build GRCh38
rs778649600 Chr.21: 44074502-44074504 on Build GRCh38
rs559357832 Chr.21: 44074565-44074565 on Build GRCh38
rs775477142 Chr.21: 44074307-44074307 on Build GRCh38
rs201286049 Chr.21: 44074502-44074502 on Build GRCh38
rs148845575 Chr.21: 44074720-44074720 on Build GRCh38
rs1038927957 Chr.21: 44074477-44074477 on Build GRCh38
rs768558481 Chr.21: 44074574-44074574 on Build GRCh38
rs200218438 Chr.21: 44074501-44074501 on Build GRCh38
rs370368380 Chr.21: 44074306-44074306 on Build GRCh38
rs746024576 Chr.21: 44074279-44074279 on Build GRCh38
rs779816335 Chr.21: 44074384-44074384 on Build GRCh38
rs751195148 Chr.21: 44074641-44074641 on Build GRCh38
rs746864230 Chr.21: 44074517-44074517 on Build GRCh38
rs941522002 Chr.21: 44074713-44074713 on Build GRCh38
rs775279067 Chr.21: 44074441-44074441 on Build GRCh38
rs967254481 Chr.21: 44074347-44074347 on Build GRCh38
rs756335355 Chr.21: 44074344-44074344 on Build GRCh38
rs770987965 Chr.21: 44074579-44074579 on Build GRCh38
rs188849410 Chr.21: 44074636-44074636 on Build GRCh38
rs762830316 Chr.21: 44074317-44074317 on Build GRCh38
rs148635981 Chr.21: 44074445-44074445 on Build GRCh38
rs369287519 Chr.21: 44074513-44074513 on Build GRCh38
rs780239636 Chr.21: 44074357-44074357 on Build GRCh38
rs748553382 Chr.21: 44074414-44074414 on Build GRCh38
rs551453075 Chr.21: 44074727-44074727 on Build GRCh38
rs770505686 Chr.21: 44074415-44074415 on Build GRCh38
rs942971257 Chr.21: 44074731-44074731 on Build GRCh38
rs148031004 Chr.21: 44074723-44074723 on Build GRCh38
rs756527087 Chr.21: 44074283-44074283 on Build GRCh38
rs750522692 Chr.21: 44074475-44074475 on Build GRCh38
rs749626397 Chr.21: 44074297-44074297 on Build GRCh38
rs2277805 Chr.21: 44074302-44074302 on Build GRCh38
rs370838084 Chr.21: 44074301-44074301 on Build GRCh38

View Full Product Details