Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44457415-44457914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC3
Entrez Gene ID
81543
Gene Name
leucine rich repeat containing 3
Gene Aliases
C21orf102
UniGene
Hs.326579
Gene Chromosome Location
Chr.21: 44455486-44458856 on Build GRCh38
Transcripts
NM_030891.4

SNP Detail

SNP ID Chromosome Location
rs556850981 Chr.21: 44457433-44457433 on Build GRCh38
rs1056961952 Chr.21: 44457819-44457819 on Build GRCh38
rs191465790 Chr.21: 44457631-44457631 on Build GRCh38
rs1004889397 Chr.21: 44457444-44457444 on Build GRCh38
rs953963611 Chr.21: 44457555-44457555 on Build GRCh38
rs374190254 Chr.21: 44457634-44457634 on Build GRCh38
rs769644671 Chr.21: 44457622-44457622 on Build GRCh38
rs999852024 Chr.21: 44457556-44457556 on Build GRCh38
rs983798910 Chr.21: 44457649-44457649 on Build GRCh38
rs891465679 Chr.21: 44457767-44457767 on Build GRCh38
rs568833892 Chr.21: 44457827-44457827 on Build GRCh38
rs146419808 Chr.21: 44457443-44457443 on Build GRCh38
rs369421218 Chr.21: 44457439-44457439 on Build GRCh38
rs998347051 Chr.21: 44457690-44457690 on Build GRCh38
rs564182143 Chr.21: 44457817-44457817 on Build GRCh38
rs977330110 Chr.21: 44457499-44457499 on Build GRCh38
rs1042925896 Chr.21: 44457596-44457596 on Build GRCh38
rs984618254 Chr.21: 44457887-44457887 on Build GRCh38
rs74756726 Chr.21: 44457639-44457639 on Build GRCh38
rs766327180 Chr.21: 44457676-44457676 on Build GRCh38
rs772368879 Chr.21: 44457442-44457442 on Build GRCh38
rs761171347 Chr.21: 44457465-44457465 on Build GRCh38
rs964654935 Chr.21: 44457654-44457654 on Build GRCh38
rs775581847 Chr.21: 44457836-44457836 on Build GRCh38
rs979937841 Chr.21: 44457624-44457624 on Build GRCh38
rs147063689 Chr.21: 44457774-44457774 on Build GRCh38
rs754607869 Chr.21: 44457593-44457593 on Build GRCh38
rs917048640 Chr.21: 44457618-44457618 on Build GRCh38
rs988660834 Chr.21: 44457699-44457699 on Build GRCh38
rs909887238 Chr.21: 44457658-44457658 on Build GRCh38
rs547614858 Chr.21: 44457515-44457515 on Build GRCh38
rs1018500551 Chr.21: 44457652-44457652 on Build GRCh38
rs1049951125 Chr.21: 44457700-44457700 on Build GRCh38
rs946694891 Chr.21: 44457731-44457731 on Build GRCh38
rs1024166505 Chr.21: 44457790-44457790 on Build GRCh38
rs138446409 Chr.21: 44457849-44457849 on Build GRCh38
rs935449443 Chr.21: 44457549-44457549 on Build GRCh38
rs534393890 Chr.21: 44457855-44457855 on Build GRCh38
rs749309270 Chr.21: 44457434-44457434 on Build GRCh38
rs528275921 Chr.21: 44457677-44457677 on Build GRCh38
rs751038855 Chr.21: 44457518-44457518 on Build GRCh38
rs550602788 Chr.21: 44457800-44457800 on Build GRCh38
rs554625916 Chr.21: 44457841-44457841 on Build GRCh38
rs935412443 Chr.21: 44457694-44457694 on Build GRCh38
rs374789026 Chr.21: 44457530-44457530 on Build GRCh38
rs1048366215 Chr.21: 44457640-44457640 on Build GRCh38
rs761052387 Chr.21: 44457434-44457435 on Build GRCh38
rs141623228 Chr.21: 44457678-44457678 on Build GRCh38
rs147881682 Chr.21: 44457594-44457594 on Build GRCh38
rs751021764 Chr.21: 44457440-44457440 on Build GRCh38
rs911377761 Chr.21: 44457480-44457480 on Build GRCh38
rs994360770 Chr.21: 44457635-44457635 on Build GRCh38
rs538891424 Chr.21: 44457775-44457775 on Build GRCh38
rs747898726 Chr.21: 44457621-44457621 on Build GRCh38
rs959455333 Chr.21: 44457874-44457874 on Build GRCh38
rs901464451 Chr.21: 44457689-44457689 on Build GRCh38
rs895661992 Chr.21: 44457825-44457825 on Build GRCh38
rs993916327 Chr.21: 44457828-44457828 on Build GRCh38
rs1052445668 Chr.21: 44457571-44457571 on Build GRCh38
rs768748576 Chr.21: 44457435-44457435 on Build GRCh38
rs575110561 Chr.21: 44457485-44457485 on Build GRCh38
rs926746944 Chr.21: 44457436-44457436 on Build GRCh38
rs938016561 Chr.21: 44457438-44457438 on Build GRCh38
rs1035938745 Chr.21: 44457632-44457632 on Build GRCh38
rs971314739 Chr.21: 44457823-44457823 on Build GRCh38
rs1000374229 Chr.21: 44457738-44457738 on Build GRCh38
rs112717208 Chr.21: 44457540-44457540 on Build GRCh38
rs1033117410 Chr.21: 44457586-44457586 on Build GRCh38
rs758684147 Chr.21: 44457644-44457644 on Build GRCh38
rs536372793 Chr.21: 44457835-44457835 on Build GRCh38
rs889527122 Chr.21: 44457735-44457735 on Build GRCh38
rs1032651190 Chr.21: 44457747-44457747 on Build GRCh38
rs867483919 Chr.21: 44457587-44457587 on Build GRCh38

View Full Product Details