Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 45455405-45455905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL18A1
Entrez Gene ID
80781
Gene Name
collagen type XVIII alpha 1 chain
Gene Aliases
KNO, KNO1, KS
UniGene
Hs.517356
Gene Chromosome Location
Chr.21: 45405137-45513720 on Build GRCh38
Transcripts
NM_030582.3 NM_130444.2 NM_130445.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3939528 c.292G>A p.E98K Chr.21: 45455822-45455822 on Build GRCh38
4582129 c.179C>T p.T60I Chr.21: 45455709-45455709 on Build GRCh38
3939529 c.292G>A p.E98K Chr.21: 45455822-45455822 on Build GRCh38
6094832 c.19G>A p.G7S Chr.21: 45455549-45455549 on Build GRCh38
6632599 c.194delC p.R67fs*49 Chr.21: 45455724-45455724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750868738 Chr.21: 45455867-45455867 on Build GRCh38
rs370000815 Chr.21: 45455854-45455854 on Build GRCh38
rs529405045 Chr.21: 45455844-45455844 on Build GRCh38
rs749775709 Chr.21: 45455542-45455542 on Build GRCh38
rs114139997 Chr.21: 45455861-45455861 on Build GRCh38
rs114899519 Chr.21: 45455509-45455509 on Build GRCh38
rs748462347 Chr.21: 45455502-45455505 on Build GRCh38
rs748399816 Chr.21: 45455554-45455554 on Build GRCh38
rs753936486 Chr.21: 45455611-45455611 on Build GRCh38
rs375958675 Chr.21: 45455769-45455770 on Build GRCh38
rs867129226 Chr.21: 45455853-45455853 on Build GRCh38
rs775587295 Chr.21: 45455850-45455850 on Build GRCh38
rs200354859 Chr.21: 45455701-45455701 on Build GRCh38
rs772982058 Chr.21: 45455768-45455768 on Build GRCh38
rs554366449 Chr.21: 45455496-45455496 on Build GRCh38
rs752886739 Chr.21: 45455793-45455793 on Build GRCh38
rs756147946 Chr.21: 45455529-45455529 on Build GRCh38
rs199984023 Chr.21: 45455661-45455661 on Build GRCh38
rs369389768 Chr.21: 45455621-45455621 on Build GRCh38
rs779335371 Chr.21: 45455550-45455550 on Build GRCh38
rs773963114 Chr.21: 45455757-45455757 on Build GRCh38
rs554930126 Chr.21: 45455609-45455609 on Build GRCh38
rs764301199 Chr.21: 45455855-45455855 on Build GRCh38
rs764638296 Chr.21: 45455812-45455812 on Build GRCh38
rs557923229 Chr.21: 45455879-45455879 on Build GRCh38
rs781638267 Chr.21: 45455822-45455822 on Build GRCh38
rs749001958 Chr.21: 45455751-45455751 on Build GRCh38
rs1016891053 Chr.21: 45455497-45455497 on Build GRCh38
rs377623821 Chr.21: 45455842-45455842 on Build GRCh38
rs748007378 Chr.21: 45455762-45455762 on Build GRCh38
rs769404140 Chr.21: 45455575-45455575 on Build GRCh38
rs887924468 Chr.21: 45455520-45455520 on Build GRCh38
rs770111210 Chr.21: 45455849-45455849 on Build GRCh38
rs73909563 Chr.21: 45455438-45455438 on Build GRCh38
rs928576283 Chr.21: 45455644-45455644 on Build GRCh38
rs767745134 Chr.21: 45455866-45455866 on Build GRCh38
rs1002894782 Chr.21: 45455459-45455459 on Build GRCh38
rs781334856 Chr.21: 45455519-45455519 on Build GRCh38
rs1031661837 Chr.21: 45455587-45455587 on Build GRCh38
rs965991021 Chr.21: 45455471-45455471 on Build GRCh38
rs375087150 Chr.21: 45455729-45455729 on Build GRCh38
rs931065734 Chr.21: 45455511-45455511 on Build GRCh38
rs757851697 Chr.21: 45455883-45455883 on Build GRCh38
rs779506536 Chr.21: 45455748-45455748 on Build GRCh38
rs370812546 Chr.21: 45455830-45455830 on Build GRCh38
rs979960718 Chr.21: 45455796-45455796 on Build GRCh38
rs762845756 Chr.21: 45455508-45455508 on Build GRCh38
rs775178517 Chr.21: 45455582-45455582 on Build GRCh38
rs756378916 Chr.21: 45455606-45455606 on Build GRCh38
rs61739237 Chr.21: 45455769-45455769 on Build GRCh38
rs374229287 Chr.21: 45455500-45455500 on Build GRCh38
rs61735029 Chr.21: 45455676-45455676 on Build GRCh38
rs375714903 Chr.21: 45455846-45455846 on Build GRCh38
rs149777818 Chr.21: 45455833-45455833 on Build GRCh38
rs948641424 Chr.21: 45455493-45455493 on Build GRCh38
rs7277693 Chr.21: 45455586-45455586 on Build GRCh38
rs748689940 Chr.21: 45455662-45455662 on Build GRCh38
rs757709318 Chr.21: 45455731-45455731 on Build GRCh38
rs754968843 Chr.21: 45455710-45455710 on Build GRCh38
rs753119196 Chr.21: 45455719-45455719 on Build GRCh38
rs75503020 Chr.21: 45455503-45455503 on Build GRCh38
rs768003538 Chr.21: 45455752-45455752 on Build GRCh38
rs568608028 Chr.21: 45455464-45455464 on Build GRCh38
rs758784072 Chr.21: 45455643-45455643 on Build GRCh38
rs777643778 Chr.21: 45455836-45455836 on Build GRCh38
rs752078007 Chr.21: 45455594-45455595 on Build GRCh38
rs760347610 Chr.21: 45455492-45455492 on Build GRCh38
rs758958927 Chr.21: 45455795-45455795 on Build GRCh38
rs752631911 Chr.21: 45455504-45455504 on Build GRCh38
rs750596734 Chr.21: 45455604-45455604 on Build GRCh38
rs938697545 Chr.21: 45455652-45455652 on Build GRCh38
rs934448081 Chr.21: 45455521-45455521 on Build GRCh38
rs759282506 Chr.21: 45455572-45455572 on Build GRCh38
rs765197743 Chr.21: 45455788-45455788 on Build GRCh38
rs779217825 Chr.21: 45455627-45455627 on Build GRCh38
rs761835450 Chr.21: 45455674-45455674 on Build GRCh38
rs557984883 Chr.21: 45455522-45455522 on Build GRCh38
rs769637337 Chr.21: 45455654-45455654 on Build GRCh38
rs1031791883 Chr.21: 45455734-45455734 on Build GRCh38
rs576327205 Chr.21: 45455530-45455530 on Build GRCh38
rs760886724 Chr.21: 45455684-45455684 on Build GRCh38
rs746967987 Chr.21: 45455839-45455839 on Build GRCh38
rs61749002 Chr.21: 45455672-45455672 on Build GRCh38
rs763995210 Chr.21: 45455594-45455594 on Build GRCh38
rs776380175 Chr.21: 45455563-45455563 on Build GRCh38
rs764004091 Chr.21: 45455723-45455724 on Build GRCh38
rs948575157 Chr.21: 45455778-45455778 on Build GRCh38
rs980024376 Chr.21: 45455617-45455617 on Build GRCh38
rs754701304 Chr.21: 45455829-45455829 on Build GRCh38
rs367609600 Chr.21: 45455501-45455501 on Build GRCh38
rs767332921 Chr.21: 45455602-45455602 on Build GRCh38
rs201529109 Chr.21: 45455821-45455821 on Build GRCh38
rs373586271 Chr.21: 45455720-45455720 on Build GRCh38
rs541819056 Chr.21: 45455449-45455449 on Build GRCh38
rs748717777 Chr.21: 45455641-45455641 on Build GRCh38
rs200976185 Chr.21: 45455620-45455620 on Build GRCh38
rs893054570 Chr.21: 45455434-45455434 on Build GRCh38
rs970127668 Chr.21: 45455807-45455808 on Build GRCh38
rs375363285 Chr.21: 45455612-45455612 on Build GRCh38
rs776665393 Chr.21: 45455771-45455771 on Build GRCh38
rs745484237 Chr.21: 45455651-45455651 on Build GRCh38
rs199929202 Chr.21: 45455709-45455709 on Build GRCh38
rs1032131702 Chr.21: 45455486-45455487 on Build GRCh38
rs752470 Chr.21: 45455453-45455453 on Build GRCh38
rs553986145 Chr.21: 45455488-45455488 on Build GRCh38
rs368343259 Chr.21: 45455559-45455559 on Build GRCh38
rs1037159818 Chr.21: 45455433-45455433 on Build GRCh38
rs751529142 Chr.21: 45455595-45455595 on Build GRCh38
rs762647237 Chr.21: 45455682-45455691 on Build GRCh38
rs563619668 Chr.21: 45455770-45455770 on Build GRCh38
rs775537458 Chr.21: 45455637-45455650 on Build GRCh38
rs747435605 Chr.21: 45455730-45455730 on Build GRCh38
rs753411491 Chr.21: 45455827-45455827 on Build GRCh38
rs777309478 Chr.21: 45455884-45455884 on Build GRCh38
rs976198093 Chr.21: 45455476-45455476 on Build GRCh38
rs772223103 Chr.21: 45455560-45455560 on Build GRCh38
rs370300539 Chr.21: 45455495-45455495 on Build GRCh38
rs769018929 Chr.21: 45455487-45455487 on Build GRCh38
rs759848880 Chr.21: 45455781-45455781 on Build GRCh38
rs368696106 Chr.21: 45455665-45455665 on Build GRCh38
rs1027815493 Chr.21: 45455447-45455447 on Build GRCh38
rs773035029 Chr.21: 45455678-45455678 on Build GRCh38
rs751376691 Chr.21: 45455514-45455514 on Build GRCh38
rs765377882 Chr.21: 45455714-45455714 on Build GRCh38
rs761449651 Chr.21: 45455596-45455596 on Build GRCh38
rs62000459 Chr.21: 45455878-45455878 on Build GRCh38
rs773455356 Chr.21: 45455569-45455569 on Build GRCh38
rs914230 Chr.21: 45455650-45455650 on Build GRCh38
rs759495424 Chr.21: 45455575-45455577 on Build GRCh38
rs771588051 Chr.21: 45455834-45455834 on Build GRCh38
rs938462922 Chr.21: 45455428-45455433 on Build GRCh38
rs374634058 Chr.21: 45455860-45455860 on Build GRCh38
rs754577294 Chr.21: 45455875-45455875 on Build GRCh38
rs537048520 Chr.21: 45455764-45455764 on Build GRCh38
rs769086878 Chr.21: 45455545-45455545 on Build GRCh38

View Full Product Details