Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24321978-24322486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPECC1L
Entrez Gene ID
23384
Gene Name
sperm antigen with calponin homology and coiled-coil domains 1 like
Gene Aliases
CYTSA, GBBB2, OBLFC1
UniGene
Hs.474384
Gene Chromosome Location
Chr.22: 24270817-24417740 on Build GRCh38
Transcripts
NM_001145468.3 NM_001254732.2 NM_015330.4 NR_103546.1

Gene Symbol
SPECC1L-ADORA2A
Entrez Gene ID
101730217
Gene Name
SPECC1L-ADORA2A readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.474384
Gene Chromosome Location
Chr.22: 24270817-24442360 on Build GRCh38
Transcripts
NM_001145468.3 NM_001254732.2 NM_015330.4 NR_103546.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6161700 c.1482T>A p.R494R Chr.22: 24322462-24322462 on Build GRCh38
6094953 c.1142G>T p.R381L Chr.22: 24322122-24322122 on Build GRCh38
4103063 c.1063C>T p.P355S Chr.22: 24322043-24322043 on Build GRCh38
7414204 c.1055T>A p.V352D Chr.22: 24322035-24322035 on Build GRCh38
5525617 c.1156G>A p.G386R Chr.22: 24322136-24322136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757073861 Chr.22: 24322037-24322037 on Build GRCh38
rs368577395 Chr.22: 24322466-24322466 on Build GRCh38
rs201737560 Chr.22: 24322446-24322446 on Build GRCh38
rs200935130 Chr.22: 24322066-24322066 on Build GRCh38
rs112546920 Chr.22: 24322387-24322387 on Build GRCh38
rs555677665 Chr.22: 24322430-24322430 on Build GRCh38
rs199964094 Chr.22: 24322075-24322075 on Build GRCh38
rs189698315 Chr.22: 24322320-24322320 on Build GRCh38
rs200004718 Chr.22: 24322383-24322383 on Build GRCh38
rs199776299 Chr.22: 24322271-24322271 on Build GRCh38
rs201503447 Chr.22: 24322460-24322460 on Build GRCh38
rs200189237 Chr.22: 24322439-24322439 on Build GRCh38
rs375824180 Chr.22: 24322003-24322003 on Build GRCh38
rs200112322 Chr.22: 24322136-24322136 on Build GRCh38
rs200113830 Chr.22: 24322053-24322053 on Build GRCh38
rs201832371 Chr.22: 24322043-24322043 on Build GRCh38
rs201734525 Chr.22: 24322264-24322264 on Build GRCh38
rs201943531 Chr.22: 24322109-24322109 on Build GRCh38
rs774014798 Chr.22: 24322396-24322396 on Build GRCh38
rs202073031 Chr.22: 24322069-24322069 on Build GRCh38
rs200756643 Chr.22: 24322021-24322021 on Build GRCh38
rs778187636 Chr.22: 24322253-24322253 on Build GRCh38
rs573041479 Chr.22: 24322353-24322353 on Build GRCh38
rs201856077 Chr.22: 24322135-24322135 on Build GRCh38
rs775820123 Chr.22: 24322292-24322292 on Build GRCh38
rs184380299 Chr.22: 24322241-24322241 on Build GRCh38
rs761342220 Chr.22: 24322397-24322397 on Build GRCh38
rs201419442 Chr.22: 24322126-24322126 on Build GRCh38
rs764710331 Chr.22: 24322121-24322121 on Build GRCh38
rs961990425 Chr.22: 24322350-24322350 on Build GRCh38
rs572646048 Chr.22: 24322038-24322038 on Build GRCh38
rs200078151 Chr.22: 24322285-24322285 on Build GRCh38
rs762544209 Chr.22: 24322294-24322294 on Build GRCh38
rs113210669 Chr.22: 24322129-24322129 on Build GRCh38
rs755829276 Chr.22: 24322321-24322321 on Build GRCh38
rs774478874 Chr.22: 24322065-24322065 on Build GRCh38
rs760081963 Chr.22: 24322115-24322115 on Build GRCh38
rs370468668 Chr.22: 24322099-24322099 on Build GRCh38
rs149701047 Chr.22: 24322357-24322357 on Build GRCh38
rs374718404 Chr.22: 24322411-24322411 on Build GRCh38
rs140238402 Chr.22: 24322403-24322403 on Build GRCh38
rs786201030 Chr.22: 24322169-24322169 on Build GRCh38
rs200390255 Chr.22: 24322222-24322222 on Build GRCh38
rs776109901 Chr.22: 24322375-24322375 on Build GRCh38
rs201343103 Chr.22: 24322093-24322093 on Build GRCh38
rs111653895 Chr.22: 24322229-24322229 on Build GRCh38
rs770001710 Chr.22: 24322367-24322367 on Build GRCh38
rs115222940 Chr.22: 24322128-24322128 on Build GRCh38
rs199621175 Chr.22: 24322243-24322243 on Build GRCh38
rs143055101 Chr.22: 24322339-24322339 on Build GRCh38
rs756538892 Chr.22: 24322461-24322461 on Build GRCh38
rs767223178 Chr.22: 24322310-24322310 on Build GRCh38
rs55723436 Chr.22: 24322440-24322440 on Build GRCh38
rs55945050 Chr.22: 24322309-24322309 on Build GRCh38
rs776965771 Chr.22: 24322244-24322244 on Build GRCh38
rs780064334 Chr.22: 24322322-24322322 on Build GRCh38
rs930899878 Chr.22: 24322134-24322134 on Build GRCh38
rs760568337 Chr.22: 24322406-24322406 on Build GRCh38
rs768688210 Chr.22: 24322057-24322057 on Build GRCh38
rs781499696 Chr.22: 24322448-24322448 on Build GRCh38
rs551676689 Chr.22: 24322111-24322111 on Build GRCh38
rs756093513 Chr.22: 24322182-24322182 on Build GRCh38
rs781324727 Chr.22: 24322361-24322361 on Build GRCh38
rs754533880 Chr.22: 24322004-24322004 on Build GRCh38
rs779793276 Chr.22: 24322201-24322201 on Build GRCh38
rs778019038 Chr.22: 24322234-24322234 on Build GRCh38
rs200327822 Chr.22: 24322122-24322122 on Build GRCh38
rs757163782 Chr.22: 24322156-24322156 on Build GRCh38
rs761050341 Chr.22: 24322090-24322090 on Build GRCh38
rs200775992 Chr.22: 24322255-24322255 on Build GRCh38
rs1002082218 Chr.22: 24322380-24322380 on Build GRCh38
rs907675222 Chr.22: 24322340-24322340 on Build GRCh38
rs781100664 Chr.22: 24322162-24322162 on Build GRCh38
rs180674130 Chr.22: 24322006-24322006 on Build GRCh38
rs763671168 Chr.22: 24322305-24322305 on Build GRCh38
rs145257997 Chr.22: 24322019-24322019 on Build GRCh38
rs768345667 Chr.22: 24322381-24322381 on Build GRCh38
rs202216724 Chr.22: 24322407-24322407 on Build GRCh38
rs750446613 Chr.22: 24322181-24322181 on Build GRCh38
rs767313233 Chr.22: 24322401-24322401 on Build GRCh38
rs535069429 Chr.22: 24322245-24322245 on Build GRCh38
rs751295195 Chr.22: 24322307-24322307 on Build GRCh38
rs751641675 Chr.22: 24322445-24322445 on Build GRCh38
rs759759553 Chr.22: 24322257-24322257 on Build GRCh38
rs145506280 Chr.22: 24322379-24322379 on Build GRCh38
rs200081627 Chr.22: 24322331-24322331 on Build GRCh38
rs777827708 Chr.22: 24322344-24322344 on Build GRCh38
rs777232246 Chr.22: 24322014-24322014 on Build GRCh38
rs375126702 Chr.22: 24322171-24322171 on Build GRCh38
rs200018524 Chr.22: 24322011-24322011 on Build GRCh38
rs748294687 Chr.22: 24322219-24322219 on Build GRCh38
rs1018962862 Chr.22: 24322116-24322116 on Build GRCh38
rs387907108 Chr.22: 24322224-24322224 on Build GRCh38
rs537661552 Chr.22: 24322008-24322008 on Build GRCh38
rs201026999 Chr.22: 24322228-24322228 on Build GRCh38

View Full Product Details