Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24324046-24324572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPECC1L
Entrez Gene ID
23384
Gene Name
sperm antigen with calponin homology and coiled-coil domains 1 like
Gene Aliases
CYTSA, GBBB2, OBLFC1
UniGene
Hs.474384
Gene Chromosome Location
Chr.22: 24270817-24417740 on Build GRCh38
Transcripts
NM_001145468.3 NM_001254732.2 NM_015330.4 NR_103546.1

Gene Symbol
SPECC1L-ADORA2A
Entrez Gene ID
101730217
Gene Name
SPECC1L-ADORA2A readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.474384
Gene Chromosome Location
Chr.22: 24270817-24442360 on Build GRCh38
Transcripts
NM_001145468.3 NM_001254732.2 NM_015330.4 NR_103546.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4103064 c.1991A>G p.D664G Chr.22: 24324272-24324272 on Build GRCh38
5774379 c.2102G>A p.R701H Chr.22: 24324383-24324383 on Build GRCh38
5501692 c.2137G>T p.D713Y Chr.22: 24324418-24324418 on Build GRCh38
1249634 c.2003C>T p.T668M Chr.22: 24324284-24324284 on Build GRCh38
4729492 c.2062G>T p.E688* Chr.22: 24324343-24324343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561062132 Chr.22: 24324160-24324160 on Build GRCh38
rs764487800 Chr.22: 24324223-24324223 on Build GRCh38
rs200188011 Chr.22: 24324204-24324204 on Build GRCh38
rs764424646 Chr.22: 24324291-24324291 on Build GRCh38
rs901127816 Chr.22: 24324519-24324519 on Build GRCh38
rs778366845 Chr.22: 24324406-24324406 on Build GRCh38
rs201384488 Chr.22: 24324209-24324209 on Build GRCh38
rs200617696 Chr.22: 24324272-24324272 on Build GRCh38
rs147450336 Chr.22: 24324285-24324285 on Build GRCh38
rs767041166 Chr.22: 24324466-24324466 on Build GRCh38
rs748008686 Chr.22: 24324408-24324408 on Build GRCh38
rs773838156 Chr.22: 24324073-24324073 on Build GRCh38
rs774199671 Chr.22: 24324170-24324170 on Build GRCh38
rs867329096 Chr.22: 24324397-24324397 on Build GRCh38
rs753492834 Chr.22: 24324382-24324382 on Build GRCh38
rs369515461 Chr.22: 24324478-24324478 on Build GRCh38
rs772143480 Chr.22: 24324267-24324267 on Build GRCh38
rs199606998 Chr.22: 24324260-24324260 on Build GRCh38
rs1042605676 Chr.22: 24324490-24324490 on Build GRCh38
rs553800098 Chr.22: 24324125-24324125 on Build GRCh38
rs1027989165 Chr.22: 24324169-24324169 on Build GRCh38
rs200054678 Chr.22: 24324367-24324367 on Build GRCh38
rs770116658 Chr.22: 24324193-24324194 on Build GRCh38
rs757765534 Chr.22: 24324237-24324237 on Build GRCh38
rs774791001 Chr.22: 24324295-24324295 on Build GRCh38
rs576552629 Chr.22: 24324196-24324196 on Build GRCh38
rs749938286 Chr.22: 24324245-24324245 on Build GRCh38
rs200147217 Chr.22: 24324290-24324290 on Build GRCh38
rs759942931 Chr.22: 24324182-24324183 on Build GRCh38
rs201193901 Chr.22: 24324476-24324476 on Build GRCh38
rs200829994 Chr.22: 24324240-24324240 on Build GRCh38
rs767950904 Chr.22: 24324239-24324239 on Build GRCh38
rs765585355 Chr.22: 24324215-24324215 on Build GRCh38
rs997895011 Chr.22: 24324166-24324166 on Build GRCh38
rs747843492 Chr.22: 24324261-24324261 on Build GRCh38
rs572113918 Chr.22: 24324128-24324128 on Build GRCh38
rs779734266 Chr.22: 24324250-24324250 on Build GRCh38
rs761567078 Chr.22: 24324079-24324079 on Build GRCh38
rs201728404 Chr.22: 24324337-24324337 on Build GRCh38
rs560143239 Chr.22: 24324497-24324497 on Build GRCh38
rs199736149 Chr.22: 24324236-24324236 on Build GRCh38
rs1022277414 Chr.22: 24324182-24324182 on Build GRCh38
rs200810244 Chr.22: 24324329-24324329 on Build GRCh38
rs940727108 Chr.22: 24324507-24324507 on Build GRCh38
rs542672184 Chr.22: 24324139-24324139 on Build GRCh38
rs148577244 Chr.22: 24324302-24324302 on Build GRCh38
rs199810315 Chr.22: 24324172-24324172 on Build GRCh38
rs755662712 Chr.22: 24324355-24324355 on Build GRCh38
rs763275175 Chr.22: 24324217-24324217 on Build GRCh38
rs199697138 Chr.22: 24324249-24324249 on Build GRCh38
rs775804377 Chr.22: 24324216-24324216 on Build GRCh38
rs745965415 Chr.22: 24324274-24324274 on Build GRCh38
rs1026603859 Chr.22: 24324326-24324326 on Build GRCh38
rs765870412 Chr.22: 24324372-24324372 on Build GRCh38
rs754800303 Chr.22: 24324252-24324252 on Build GRCh38
rs143004795 Chr.22: 24324284-24324284 on Build GRCh38
rs931782267 Chr.22: 24324424-24324424 on Build GRCh38
rs889425153 Chr.22: 24324550-24324550 on Build GRCh38
rs754390959 Chr.22: 24324535-24324535 on Build GRCh38
rs766105646 Chr.22: 24324324-24324327 on Build GRCh38
rs754676542 Chr.22: 24324393-24324393 on Build GRCh38
rs1002876548 Chr.22: 24324111-24324111 on Build GRCh38
rs5760340 Chr.22: 24324416-24324416 on Build GRCh38
rs551393813 Chr.22: 24324479-24324479 on Build GRCh38
rs187324735 Chr.22: 24324251-24324251 on Build GRCh38
rs777903202 Chr.22: 24324475-24324475 on Build GRCh38
rs767977884 Chr.22: 24324165-24324165 on Build GRCh38
rs909117573 Chr.22: 24324484-24324484 on Build GRCh38
rs374091889 Chr.22: 24324098-24324098 on Build GRCh38
rs1009917439 Chr.22: 24324143-24324144 on Build GRCh38
rs971386147 Chr.22: 24324370-24324370 on Build GRCh38
rs762015892 Chr.22: 24324315-24324315 on Build GRCh38
rs139099983 Chr.22: 24324248-24324248 on Build GRCh38
rs758216249 Chr.22: 24324453-24324453 on Build GRCh38
rs776864833 Chr.22: 24324202-24324202 on Build GRCh38
rs1049430087 Chr.22: 24324526-24324526 on Build GRCh38

View Full Product Details