Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24719664-24720171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIWIL3
Entrez Gene ID
440822
Gene Name
piwi like RNA-mediated gene silencing 3
Gene Aliases
HIWI3
UniGene
Hs.448343
Gene Chromosome Location
Chr.22: 24719034-24774720 on Build GRCh38
Transcripts
NM_001008496.3 NM_001255975.1 NR_045648.1 NR_045649.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
137242 c.2476C>T p.P826S Chr.22: 24719804-24719804 on Build GRCh38
726120 c.2389G>A p.D797N Chr.22: 24719891-24719891 on Build GRCh38
5631051 c.2529G>T p.L843F Chr.22: 24719751-24719751 on Build GRCh38
391494 c.2530_2531delCC p.P844fs*>39 Chr.22: 24719749-24719750 on Build GRCh38
3310302 c.2447A>C p.N816T Chr.22: 24719833-24719833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754534538 Chr.22: 24719865-24719866 on Build GRCh38
rs762922028 Chr.22: 24719849-24719849 on Build GRCh38
rs778583073 Chr.22: 24719882-24719883 on Build GRCh38
rs768715411 Chr.22: 24719853-24719853 on Build GRCh38
rs1016521957 Chr.22: 24719769-24719769 on Build GRCh38
rs754053811 Chr.22: 24719872-24719872 on Build GRCh38
rs1000066715 Chr.22: 24720045-24720048 on Build GRCh38
rs763933056 Chr.22: 24719795-24719795 on Build GRCh38
rs1022445001 Chr.22: 24719778-24719778 on Build GRCh38
rs775536418 Chr.22: 24719788-24719788 on Build GRCh38
rs997214093 Chr.22: 24719694-24719694 on Build GRCh38
rs749888929 Chr.22: 24720051-24720051 on Build GRCh38
rs778999933 Chr.22: 24719818-24719818 on Build GRCh38
rs368104028 Chr.22: 24719701-24719701 on Build GRCh38
rs200544443 Chr.22: 24719780-24719780 on Build GRCh38
rs139040174 Chr.22: 24719782-24719782 on Build GRCh38
rs778241701 Chr.22: 24719904-24719904 on Build GRCh38
rs779455110 Chr.22: 24719729-24719729 on Build GRCh38
rs758541245 Chr.22: 24719888-24719888 on Build GRCh38
rs748697671 Chr.22: 24719735-24719735 on Build GRCh38
rs377157661 Chr.22: 24719834-24719834 on Build GRCh38
rs199855233 Chr.22: 24719813-24719813 on Build GRCh38
rs5760605 Chr.22: 24719992-24719992 on Build GRCh38
rs906907725 Chr.22: 24719730-24719730 on Build GRCh38
rs372720948 Chr.22: 24719941-24719941 on Build GRCh38
rs781439779 Chr.22: 24719926-24719926 on Build GRCh38
rs368305582 Chr.22: 24719883-24719883 on Build GRCh38
rs766025061 Chr.22: 24719771-24719772 on Build GRCh38
rs201599301 Chr.22: 24719750-24719750 on Build GRCh38
rs752778767 Chr.22: 24719886-24719886 on Build GRCh38
rs200698651 Chr.22: 24719762-24719762 on Build GRCh38
rs756220842 Chr.22: 24719803-24719803 on Build GRCh38
rs764020941 Chr.22: 24719707-24719707 on Build GRCh38
rs757176094 Chr.22: 24719962-24719962 on Build GRCh38
rs901526499 Chr.22: 24719692-24719692 on Build GRCh38
rs61740100 Chr.22: 24719832-24719832 on Build GRCh38
rs757608299 Chr.22: 24719920-24719920 on Build GRCh38
rs915909613 Chr.22: 24719827-24719827 on Build GRCh38
rs745475363 Chr.22: 24719927-24719927 on Build GRCh38
rs754246262 Chr.22: 24719708-24719708 on Build GRCh38
rs779936341 Chr.22: 24719714-24719714 on Build GRCh38
rs753694000 Chr.22: 24719844-24719844 on Build GRCh38
rs769584264 Chr.22: 24719938-24719938 on Build GRCh38
rs895375087 Chr.22: 24719757-24719757 on Build GRCh38
rs142951647 Chr.22: 24719801-24719801 on Build GRCh38
rs759702806 Chr.22: 24719880-24719880 on Build GRCh38
rs888753818 Chr.22: 24719984-24719984 on Build GRCh38
rs756377564 Chr.22: 24719711-24719711 on Build GRCh38
rs773593639 Chr.22: 24719749-24719749 on Build GRCh38
rs766616676 Chr.22: 24719812-24719812 on Build GRCh38
rs897719198 Chr.22: 24720052-24720052 on Build GRCh38
rs750634294 Chr.22: 24719708-24719708 on Build GRCh38
rs201985483 Chr.22: 24719742-24719742 on Build GRCh38
rs188580272 Chr.22: 24720106-24720106 on Build GRCh38
rs755077231 Chr.22: 24719728-24719728 on Build GRCh38
rs368816009 Chr.22: 24719859-24719859 on Build GRCh38
rs1037603110 Chr.22: 24720042-24720042 on Build GRCh38
rs752057987 Chr.22: 24719911-24719911 on Build GRCh38
rs780873182 Chr.22: 24719838-24719838 on Build GRCh38
rs767258800 Chr.22: 24719865-24719865 on Build GRCh38
rs749387770 Chr.22: 24719715-24719715 on Build GRCh38
rs933056044 Chr.22: 24719891-24719891 on Build GRCh38
rs1043950638 Chr.22: 24720013-24720014 on Build GRCh38
rs960150936 Chr.22: 24720144-24720144 on Build GRCh38
rs778045733 Chr.22: 24719833-24719833 on Build GRCh38
rs201971834 Chr.22: 24719789-24719789 on Build GRCh38
rs770622234 Chr.22: 24719753-24719753 on Build GRCh38
rs140053359 Chr.22: 24719844-24719844 on Build GRCh38
rs868332493 Chr.22: 24719864-24719864 on Build GRCh38
rs745546773 Chr.22: 24719842-24719842 on Build GRCh38
rs755057734 Chr.22: 24719817-24719817 on Build GRCh38
rs771259692 Chr.22: 24719836-24719836 on Build GRCh38
rs377313898 Chr.22: 24719947-24719947 on Build GRCh38
rs201604636 Chr.22: 24719831-24719831 on Build GRCh38
rs552241699 Chr.22: 24719994-24719994 on Build GRCh38
rs772389045 Chr.22: 24719736-24719736 on Build GRCh38
rs149655408 Chr.22: 24720064-24720064 on Build GRCh38
rs937037143 Chr.22: 24719917-24719917 on Build GRCh38
rs779636092 Chr.22: 24719939-24719939 on Build GRCh38
rs774493681 Chr.22: 24719856-24719856 on Build GRCh38
rs902681066 Chr.22: 24720020-24720020 on Build GRCh38
rs993318310 Chr.22: 24720104-24720104 on Build GRCh38
rs142279392 Chr.22: 24719847-24719847 on Build GRCh38

View Full Product Details