Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 26589843-26590334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPST2
Entrez Gene ID
8459
Gene Name
tyrosylprotein sulfotransferase 2
Gene Aliases
TANGO13B
UniGene
Hs.632768
Gene Chromosome Location
Chr.22: 26524082-26590125 on Build GRCh38
Transcripts
NM_003595.3 XM_006724338.3

SNP Detail

SNP ID Chromosome Location
rs1014363985 Chr.22: 26590183-26590183 on Build GRCh38
rs561828229 Chr.22: 26590091-26590091 on Build GRCh38
rs1048976967 Chr.22: 26590160-26590160 on Build GRCh38
rs988895732 Chr.22: 26590129-26590129 on Build GRCh38
rs925137082 Chr.22: 26590119-26590119 on Build GRCh38
rs559292494 Chr.22: 26590292-26590292 on Build GRCh38
rs894573172 Chr.22: 26590181-26590181 on Build GRCh38
rs891821190 Chr.22: 26590027-26590027 on Build GRCh38
rs947960658 Chr.22: 26589978-26589978 on Build GRCh38
rs548318500 Chr.22: 26590203-26590203 on Build GRCh38
rs11090449 Chr.22: 26590011-26590011 on Build GRCh38
rs535337559 Chr.22: 26590312-26590312 on Build GRCh38
rs1014864327 Chr.22: 26590095-26590095 on Build GRCh38
rs960392527 Chr.22: 26590100-26590100 on Build GRCh38
rs1005519548 Chr.22: 26590081-26590081 on Build GRCh38
rs565869769 Chr.22: 26590131-26590131 on Build GRCh38
rs1051538984 Chr.22: 26590158-26590158 on Build GRCh38
rs918975502 Chr.22: 26590285-26590285 on Build GRCh38
rs898690937 Chr.22: 26590263-26590263 on Build GRCh38
rs541233178 Chr.22: 26590168-26590168 on Build GRCh38
rs564243527 Chr.22: 26590250-26590250 on Build GRCh38
rs892001075 Chr.22: 26590191-26590191 on Build GRCh38
rs527543811 Chr.22: 26590139-26590139 on Build GRCh38
rs367783254 Chr.22: 26590299-26590299 on Build GRCh38
rs898638318 Chr.22: 26590083-26590083 on Build GRCh38
rs543299135 Chr.22: 26590019-26590019 on Build GRCh38
rs547234029 Chr.22: 26590109-26590109 on Build GRCh38
rs935148143 Chr.22: 26590126-26590126 on Build GRCh38
rs934044692 Chr.22: 26590141-26590141 on Build GRCh38
rs748654598 Chr.22: 26590255-26590255 on Build GRCh38
rs1007105495 Chr.22: 26590075-26590075 on Build GRCh38
rs553425323 Chr.22: 26589959-26589959 on Build GRCh38
rs998984820 Chr.22: 26590197-26590197 on Build GRCh38
rs1017539669 Chr.22: 26590082-26590082 on Build GRCh38
rs1033300874 Chr.22: 26590213-26590213 on Build GRCh38
rs985810996 Chr.22: 26590121-26590121 on Build GRCh38
rs903359869 Chr.22: 26590192-26590192 on Build GRCh38
rs1023260215 Chr.22: 26590110-26590110 on Build GRCh38
rs1023734334 Chr.22: 26590311-26590311 on Build GRCh38
rs1040208543 Chr.22: 26590147-26590147 on Build GRCh38
rs897776537 Chr.22: 26590031-26590031 on Build GRCh38
rs972455853 Chr.22: 26589918-26589918 on Build GRCh38
rs979343623 Chr.22: 26590117-26590117 on Build GRCh38
rs537069852 Chr.22: 26590262-26590262 on Build GRCh38
rs574801916 Chr.22: 26589970-26589970 on Build GRCh38
rs1051385240 Chr.22: 26590045-26590045 on Build GRCh38
rs1017253738 Chr.22: 26590246-26590246 on Build GRCh38
rs1055784730 Chr.22: 26590171-26590171 on Build GRCh38
rs565079853 Chr.22: 26590073-26590073 on Build GRCh38
rs941400079 Chr.22: 26590171-26590171 on Build GRCh38
rs994326586 Chr.22: 26590283-26590283 on Build GRCh38
rs375415384 Chr.22: 26590277-26590277 on Build GRCh38
rs960350081 Chr.22: 26589953-26589953 on Build GRCh38
rs1024459372 Chr.22: 26590096-26590096 on Build GRCh38
rs969727687 Chr.22: 26590111-26590111 on Build GRCh38
rs933990804 Chr.22: 26590035-26590035 on Build GRCh38
rs975819663 Chr.22: 26590256-26590256 on Build GRCh38
rs993948703 Chr.22: 26590087-26590087 on Build GRCh38
rs889772096 Chr.22: 26590163-26590163 on Build GRCh38
rs1025330238 Chr.22: 26589929-26589929 on Build GRCh38
rs979676191 Chr.22: 26589997-26589997 on Build GRCh38
rs1047225103 Chr.22: 26590290-26590290 on Build GRCh38
rs1038606197 Chr.22: 26590179-26590179 on Build GRCh38
rs910328495 Chr.22: 26590133-26590133 on Build GRCh38
rs60407213 Chr.22: 26590107-26590107 on Build GRCh38
rs946115117 Chr.22: 26590287-26590287 on Build GRCh38
rs1048945827 Chr.22: 26590072-26590072 on Build GRCh38
rs935651429 Chr.22: 26589917-26589917 on Build GRCh38
rs535481688 Chr.22: 26590191-26590192 on Build GRCh38
rs1013312156 Chr.22: 26590300-26590300 on Build GRCh38
rs992112737 Chr.22: 26589954-26589954 on Build GRCh38
rs541924429 Chr.22: 26589983-26589983 on Build GRCh38
rs563315576 Chr.22: 26590008-26590008 on Build GRCh38
rs967513334 Chr.22: 26590099-26590099 on Build GRCh38
rs986231073 Chr.22: 26590239-26590239 on Build GRCh38
rs1024364668 Chr.22: 26590191-26590195 on Build GRCh38
rs993357496 Chr.22: 26590056-26590056 on Build GRCh38
rs1057486286 Chr.22: 26590186-26590186 on Build GRCh38

View Full Product Details