Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89116903-89117405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP2
Entrez Gene ID
2634
Gene Name
guanylate binding protein 2
Gene Aliases
-
UniGene
Hs.386567
Gene Chromosome Location
Chr.1: 89106132-89126159 on Build GRCh38
Transcripts
NM_004120.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2258842 c.702C>T p.F234F Chr.1: 89117158-89117158 on Build GRCh38
4010438 c.809A>G p.Y270C Chr.1: 89117051-89117051 on Build GRCh38
7369284 c.661C>T p.R221W Chr.1: 89117199-89117199 on Build GRCh38
7369285 c.629C>T p.T210I Chr.1: 89117231-89117231 on Build GRCh38
912973 c.798A>T p.E266D Chr.1: 89117062-89117062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151053556 Chr.1: 89117127-89117127 on Build GRCh38
rs200947484 Chr.1: 89117258-89117258 on Build GRCh38
rs936846993 Chr.1: 89117303-89117303 on Build GRCh38
rs189899416 Chr.1: 89117187-89117187 on Build GRCh38
rs199744522 Chr.1: 89117157-89117157 on Build GRCh38
rs769514280 Chr.1: 89117024-89117024 on Build GRCh38
rs756205791 Chr.1: 89116973-89116973 on Build GRCh38
rs751517507 Chr.1: 89116970-89116970 on Build GRCh38
rs370854667 Chr.1: 89117296-89117296 on Build GRCh38
rs778042009 Chr.1: 89117106-89117106 on Build GRCh38
rs781730060 Chr.1: 89117137-89117137 on Build GRCh38
rs761688294 Chr.1: 89117153-89117153 on Build GRCh38
rs943121524 Chr.1: 89117345-89117345 on Build GRCh38
rs751814530 Chr.1: 89117186-89117186 on Build GRCh38
rs894232187 Chr.1: 89117351-89117351 on Build GRCh38
rs781547775 Chr.1: 89117194-89117194 on Build GRCh38
rs1026163428 Chr.1: 89117145-89117145 on Build GRCh38
rs761695136 Chr.1: 89117042-89117042 on Build GRCh38
rs145069034 Chr.1: 89117231-89117231 on Build GRCh38
rs754059516 Chr.1: 89116989-89116989 on Build GRCh38
rs770889028 Chr.1: 89117234-89117234 on Build GRCh38
rs968360796 Chr.1: 89117097-89117097 on Build GRCh38
rs775287224 Chr.1: 89117026-89117026 on Build GRCh38
rs764057835 Chr.1: 89116954-89116954 on Build GRCh38
rs773564810 Chr.1: 89117302-89117302 on Build GRCh38
rs781516751 Chr.1: 89117229-89117229 on Build GRCh38
rs774321393 Chr.1: 89117036-89117036 on Build GRCh38
rs778639191 Chr.1: 89117217-89117217 on Build GRCh38
rs778334754 Chr.1: 89117006-89117006 on Build GRCh38
rs773114942 Chr.1: 89117230-89117230 on Build GRCh38
rs745314942 Chr.1: 89117139-89117139 on Build GRCh38
rs762071363 Chr.1: 89117270-89117270 on Build GRCh38
rs767518146 Chr.1: 89117043-89117043 on Build GRCh38
rs185011739 Chr.1: 89116991-89116991 on Build GRCh38
rs753809869 Chr.1: 89117073-89117073 on Build GRCh38
rs975475138 Chr.1: 89117212-89117212 on Build GRCh38
rs912652375 Chr.1: 89117202-89117202 on Build GRCh38
rs149789032 Chr.1: 89116992-89116992 on Build GRCh38
rs181512309 Chr.1: 89117371-89117371 on Build GRCh38
rs140113988 Chr.1: 89117169-89117169 on Build GRCh38
rs2230336 Chr.1: 89117019-89117019 on Build GRCh38
rs888847270 Chr.1: 89117054-89117054 on Build GRCh38
rs764400050 Chr.1: 89117176-89117176 on Build GRCh38
rs752887173 Chr.1: 89117168-89117168 on Build GRCh38
rs778174916 Chr.1: 89116968-89116968 on Build GRCh38
rs765372923 Chr.1: 89117079-89117079 on Build GRCh38
rs868380613 Chr.1: 89117192-89117192 on Build GRCh38
rs766417117 Chr.1: 89116976-89116976 on Build GRCh38
rs375563316 Chr.1: 89117337-89117337 on Build GRCh38
rs763853855 Chr.1: 89117033-89117033 on Build GRCh38
rs755060182 Chr.1: 89117078-89117078 on Build GRCh38
rs762836659 Chr.1: 89117032-89117032 on Build GRCh38
rs773133578 Chr.1: 89117158-89117158 on Build GRCh38
rs748450335 Chr.1: 89117000-89117000 on Build GRCh38
rs765354242 Chr.1: 89117161-89117161 on Build GRCh38
rs143042903 Chr.1: 89117142-89117142 on Build GRCh38
rs1050347858 Chr.1: 89117331-89117331 on Build GRCh38
rs918214740 Chr.1: 89116980-89116980 on Build GRCh38
rs369251246 Chr.1: 89117201-89117201 on Build GRCh38
rs372035050 Chr.1: 89117199-89117199 on Build GRCh38
rs370885169 Chr.1: 89117012-89117012 on Build GRCh38
rs945336496 Chr.1: 89117255-89117255 on Build GRCh38
rs372892988 Chr.1: 89117151-89117151 on Build GRCh38
rs758744642 Chr.1: 89117089-89117089 on Build GRCh38
rs771627137 Chr.1: 89117036-89117037 on Build GRCh38
rs777496524 Chr.1: 89116941-89116941 on Build GRCh38
rs748837721 Chr.1: 89117204-89117204 on Build GRCh38
rs142698251 Chr.1: 89117003-89117003 on Build GRCh38
rs762855111 Chr.1: 89116951-89116951 on Build GRCh38
rs1803632 Chr.1: 89117007-89117007 on Build GRCh38
rs1040086434 Chr.1: 89117369-89117369 on Build GRCh38
rs369681409 Chr.1: 89117198-89117198 on Build GRCh38
rs141308557 Chr.1: 89117143-89117143 on Build GRCh38
rs138950344 Chr.1: 89117262-89117262 on Build GRCh38
rs115143257 Chr.1: 89117149-89117149 on Build GRCh38
rs775502366 Chr.1: 89117257-89117257 on Build GRCh38
rs375023862 Chr.1: 89117088-89117088 on Build GRCh38
rs866032998 Chr.1: 89117236-89117236 on Build GRCh38
rs749738138 Chr.1: 89116991-89116992 on Build GRCh38
rs1046971244 Chr.1: 89116982-89116982 on Build GRCh38
rs886838509 Chr.1: 89116999-89116999 on Build GRCh38
rs768142276 Chr.1: 89117208-89117208 on Build GRCh38
rs774976541 Chr.1: 89117254-89117254 on Build GRCh38
rs746702811 Chr.1: 89117287-89117289 on Build GRCh38
rs1033793206 Chr.1: 89117184-89117184 on Build GRCh38
rs775965374 Chr.1: 89117014-89117014 on Build GRCh38
rs137908444 Chr.1: 89117124-89117124 on Build GRCh38

View Full Product Details