Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 91262127-91262646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HFM1
Entrez Gene ID
164045
Gene Name
HFM1, ATP dependent DNA helicase homolog
Gene Aliases
MER3, POF9, SEC63D1, Si-11, Si-11-6, helicase
UniGene
Hs.454818
Gene Chromosome Location
Chr.1: 91260766-91408008 on Build GRCh38
Transcripts
NM_001017975.4 XM_011540849.1 XM_011540850.2 XM_011540851.1 XM_011540852.2 XM_011540855.1 XM_011540857.1 XM_011540859.2 XM_017000490.1 XM_017000491.1 XM_017000492.1 XM_017000493.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2259616 c.4188delA p.V1397fs*28 Chr.1: 91262291-91262291 on Build GRCh38
6378473 c.4021G>T p.A1341S Chr.1: 91262546-91262546 on Build GRCh38
1627240 c.4032G>T p.K1344N Chr.1: 91262535-91262535 on Build GRCh38
6679635 c.4116T>G p.N1372K Chr.1: 91262363-91262363 on Build GRCh38
5946803 c.4199_4200insT p.I1401fs*2 Chr.1: 91262279-91262280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376450130 Chr.1: 91262439-91262439 on Build GRCh38
rs769090046 Chr.1: 91262360-91262360 on Build GRCh38
rs919783140 Chr.1: 91262508-91262508 on Build GRCh38
rs377088273 Chr.1: 91262170-91262170 on Build GRCh38
rs750456971 Chr.1: 91262574-91262574 on Build GRCh38
rs755325879 Chr.1: 91262610-91262610 on Build GRCh38
rs761322247 Chr.1: 91262279-91262279 on Build GRCh38
rs752719889 Chr.1: 91262453-91262453 on Build GRCh38
rs904090001 Chr.1: 91262379-91262379 on Build GRCh38
rs766581680 Chr.1: 91262599-91262599 on Build GRCh38
rs749864304 Chr.1: 91262258-91262258 on Build GRCh38
rs752618488 Chr.1: 91262406-91262406 on Build GRCh38
rs372738158 Chr.1: 91262196-91262196 on Build GRCh38
rs777271687 Chr.1: 91262507-91262507 on Build GRCh38
rs776456072 Chr.1: 91262523-91262523 on Build GRCh38
rs754896938 Chr.1: 91262307-91262307 on Build GRCh38
rs764224022 Chr.1: 91262457-91262460 on Build GRCh38
rs200535976 Chr.1: 91262516-91262516 on Build GRCh38
rs997135340 Chr.1: 91262405-91262405 on Build GRCh38
rs585898 Chr.1: 91262496-91262496 on Build GRCh38
rs756421054 Chr.1: 91262342-91262342 on Build GRCh38
rs372406769 Chr.1: 91262303-91262303 on Build GRCh38
rs980029659 Chr.1: 91262521-91262521 on Build GRCh38
rs1024337600 Chr.1: 91262182-91262185 on Build GRCh38
rs766258698 Chr.1: 91262297-91262297 on Build GRCh38
rs771622773 Chr.1: 91262397-91262397 on Build GRCh38
rs750606251 Chr.1: 91262472-91262472 on Build GRCh38
rs41286777 Chr.1: 91262171-91262171 on Build GRCh38
rs765280416 Chr.1: 91262320-91262320 on Build GRCh38
rs148254635 Chr.1: 91262568-91262568 on Build GRCh38
rs772730124 Chr.1: 91262411-91262411 on Build GRCh38
rs201980911 Chr.1: 91262569-91262569 on Build GRCh38
rs373714322 Chr.1: 91262255-91262255 on Build GRCh38
rs766204131 Chr.1: 91262278-91262279 on Build GRCh38
rs781501756 Chr.1: 91262219-91262219 on Build GRCh38
rs767387412 Chr.1: 91262405-91262406 on Build GRCh38
rs545329938 Chr.1: 91262235-91262235 on Build GRCh38
rs769759236 Chr.1: 91262555-91262555 on Build GRCh38
rs747628741 Chr.1: 91262381-91262381 on Build GRCh38
rs982619284 Chr.1: 91262375-91262375 on Build GRCh38
rs779319738 Chr.1: 91262613-91262613 on Build GRCh38
rs182338923 Chr.1: 91262438-91262438 on Build GRCh38
rs530895801 Chr.1: 91262328-91262328 on Build GRCh38
rs1022906634 Chr.1: 91262374-91262374 on Build GRCh38
rs756289532 Chr.1: 91262473-91262473 on Build GRCh38
rs770569328 Chr.1: 91262519-91262519 on Build GRCh38
rs760369118 Chr.1: 91262294-91262294 on Build GRCh38
rs12733308 Chr.1: 91262265-91262265 on Build GRCh38
rs200694100 Chr.1: 91262225-91262225 on Build GRCh38
rs143059423 Chr.1: 91262597-91262597 on Build GRCh38
rs774112248 Chr.1: 91262279-91262279 on Build GRCh38
rs529157230 Chr.1: 91262198-91262198 on Build GRCh38
rs781238872 Chr.1: 91262338-91262338 on Build GRCh38
rs754076804 Chr.1: 91262470-91262472 on Build GRCh38
rs757527550 Chr.1: 91262607-91262608 on Build GRCh38
rs749502424 Chr.1: 91262564-91262564 on Build GRCh38
rs757498375 Chr.1: 91262218-91262218 on Build GRCh38
rs139164398 Chr.1: 91262573-91262573 on Build GRCh38
rs201894202 Chr.1: 91262487-91262487 on Build GRCh38
rs906674475 Chr.1: 91262209-91262209 on Build GRCh38
rs765227359 Chr.1: 91262440-91262440 on Build GRCh38
rs746150403 Chr.1: 91262339-91262339 on Build GRCh38
rs201393479 Chr.1: 91262488-91262488 on Build GRCh38
rs150756783 Chr.1: 91262571-91262571 on Build GRCh38
rs190831701 Chr.1: 91262325-91262325 on Build GRCh38
rs185069773 Chr.1: 91262483-91262483 on Build GRCh38
rs767120204 Chr.1: 91262283-91262283 on Build GRCh38
rs370715340 Chr.1: 91262464-91262464 on Build GRCh38
rs580070 Chr.1: 91262224-91262224 on Build GRCh38
rs922527503 Chr.1: 91262316-91262319 on Build GRCh38
rs756165466 Chr.1: 91262420-91262421 on Build GRCh38
rs768147001 Chr.1: 91262268-91262268 on Build GRCh38
rs369277085 Chr.1: 91262289-91262289 on Build GRCh38
rs749654723 Chr.1: 91262350-91262350 on Build GRCh38
rs759278255 Chr.1: 91262315-91262316 on Build GRCh38
rs771482382 Chr.1: 91262502-91262502 on Build GRCh38
rs112115414 Chr.1: 91262539-91262539 on Build GRCh38
rs565616503 Chr.1: 91262365-91262365 on Build GRCh38
rs759372572 Chr.1: 91262543-91262543 on Build GRCh38
rs939436793 Chr.1: 91262175-91262175 on Build GRCh38
rs773779864 Chr.1: 91262276-91262276 on Build GRCh38
rs374822368 Chr.1: 91262421-91262421 on Build GRCh38
rs780529773 Chr.1: 91262345-91262345 on Build GRCh38
rs754110441 Chr.1: 91262601-91262601 on Build GRCh38

View Full Product Details