Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92483255-92483750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFI1
Entrez Gene ID
2672
Gene Name
growth factor independent 1 transcriptional repressor
Gene Aliases
GFI-1, GFI1A, SCN2, ZNF163
UniGene
Hs.73172
Gene Chromosome Location
Chr.1: 92473043-92486876 on Build GRCh38
Transcripts
NM_001127215.1 NM_001127216.1 NM_005263.3 XM_005270749.3 XM_011541245.2 XM_011541246.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4010615 c.19G>A p.V7I Chr.1: 92483469-92483469 on Build GRCh38
3747114 c.107G>A p.S36N Chr.1: 92483381-92483381 on Build GRCh38
6670501 c.7C>T p.R3C Chr.1: 92483481-92483481 on Build GRCh38
6127850 c.110G>C p.R37P Chr.1: 92483378-92483378 on Build GRCh38
3866247 c.82C>T p.R28C Chr.1: 92483406-92483406 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565821550 Chr.1: 92483577-92483577 on Build GRCh38
rs139390290 Chr.1: 92483541-92483541 on Build GRCh38
rs953620894 Chr.1: 92483505-92483505 on Build GRCh38
rs550414938 Chr.1: 92483448-92483448 on Build GRCh38
rs1026606429 Chr.1: 92483375-92483375 on Build GRCh38
rs558771685 Chr.1: 92483719-92483719 on Build GRCh38
rs779119308 Chr.1: 92483435-92483435 on Build GRCh38
rs372866021 Chr.1: 92483405-92483405 on Build GRCh38
rs753626698 Chr.1: 92483334-92483334 on Build GRCh38
rs780411373 Chr.1: 92483348-92483348 on Build GRCh38
rs760413467 Chr.1: 92483370-92483370 on Build GRCh38
rs749600503 Chr.1: 92483349-92483349 on Build GRCh38
rs142779589 Chr.1: 92483425-92483425 on Build GRCh38
rs757405460 Chr.1: 92483340-92483340 on Build GRCh38
rs540674112 Chr.1: 92483593-92483593 on Build GRCh38
rs997008948 Chr.1: 92483677-92483677 on Build GRCh38
rs758263613 Chr.1: 92483389-92483389 on Build GRCh38
rs564226915 Chr.1: 92483333-92483333 on Build GRCh38
rs375422035 Chr.1: 92483414-92483414 on Build GRCh38
rs1044908227 Chr.1: 92483632-92483632 on Build GRCh38
rs1019755675 Chr.1: 92483692-92483692 on Build GRCh38
rs896290404 Chr.1: 92483308-92483308 on Build GRCh38
rs150506480 Chr.1: 92483344-92483363 on Build GRCh38
rs751611941 Chr.1: 92483338-92483338 on Build GRCh38
rs765179520 Chr.1: 92483336-92483336 on Build GRCh38
rs376535724 Chr.1: 92483499-92483499 on Build GRCh38
rs1013438206 Chr.1: 92483319-92483319 on Build GRCh38
rs764916969 Chr.1: 92483387-92483387 on Build GRCh38
rs912070040 Chr.1: 92483315-92483315 on Build GRCh38
rs754873642 Chr.1: 92483335-92483335 on Build GRCh38
rs536865411 Chr.1: 92483693-92483693 on Build GRCh38
rs987571985 Chr.1: 92483291-92483291 on Build GRCh38
rs934670995 Chr.1: 92483545-92483545 on Build GRCh38
rs746155170 Chr.1: 92483346-92483346 on Build GRCh38
rs939370891 Chr.1: 92483578-92483578 on Build GRCh38
rs769076534 Chr.1: 92483357-92483357 on Build GRCh38
rs890790824 Chr.1: 92483680-92483680 on Build GRCh38
rs576417597 Chr.1: 92483455-92483455 on Build GRCh38
rs767641663 Chr.1: 92483401-92483401 on Build GRCh38
rs952239453 Chr.1: 92483562-92483562 on Build GRCh38
rs1057265156 Chr.1: 92483299-92483299 on Build GRCh38
rs34631763 Chr.1: 92483381-92483381 on Build GRCh38
rs762961809 Chr.1: 92483508-92483508 on Build GRCh38
rs149059364 Chr.1: 92483328-92483328 on Build GRCh38
rs371677124 Chr.1: 92483449-92483449 on Build GRCh38
rs903626943 Chr.1: 92483462-92483462 on Build GRCh38
rs756315448 Chr.1: 92483347-92483347 on Build GRCh38
rs1009107667 Chr.1: 92483521-92483521 on Build GRCh38
rs766059107 Chr.1: 92483378-92483378 on Build GRCh38
rs752718949 Chr.1: 92483388-92483388 on Build GRCh38
rs759324139 Chr.1: 92483384-92483384 on Build GRCh38
rs769697146 Chr.1: 92483497-92483497 on Build GRCh38
rs774875013 Chr.1: 92483365-92483365 on Build GRCh38
rs763952921 Chr.1: 92483514-92483514 on Build GRCh38
rs760698768 Chr.1: 92483535-92483535 on Build GRCh38
rs772849128 Chr.1: 92483369-92483369 on Build GRCh38
rs369812626 Chr.1: 92483419-92483419 on Build GRCh38
rs772445175 Chr.1: 92483332-92483332 on Build GRCh38
rs1009439096 Chr.1: 92483428-92483428 on Build GRCh38
rs898100389 Chr.1: 92483707-92483707 on Build GRCh38
rs184691209 Chr.1: 92483494-92483494 on Build GRCh38
rs755423368 Chr.1: 92483426-92483426 on Build GRCh38
rs554666315 Chr.1: 92483649-92483649 on Build GRCh38
rs781217351 Chr.1: 92483341-92483341 on Build GRCh38
rs750251158 Chr.1: 92483439-92483439 on Build GRCh38
rs920666138 Chr.1: 92483557-92483557 on Build GRCh38
rs1056390017 Chr.1: 92483598-92483598 on Build GRCh38
rs746474854 Chr.1: 92483477-92483477 on Build GRCh38
rs770727114 Chr.1: 92483480-92483480 on Build GRCh38
rs1043973649 Chr.1: 92483644-92483644 on Build GRCh38
rs945175494 Chr.1: 92483643-92483643 on Build GRCh38
rs1056268413 Chr.1: 92483423-92483423 on Build GRCh38
rs751035973 Chr.1: 92483678-92483678 on Build GRCh38
rs980760683 Chr.1: 92483575-92483575 on Build GRCh38
rs200972824 Chr.1: 92483501-92483501 on Build GRCh38
rs761138582 Chr.1: 92483514-92483514 on Build GRCh38
rs766331686 Chr.1: 92483322-92483322 on Build GRCh38
rs972227733 Chr.1: 92483552-92483552 on Build GRCh38
rs772644896 Chr.1: 92483442-92483442 on Build GRCh38
rs913850483 Chr.1: 92483625-92483625 on Build GRCh38
rs549580373 Chr.1: 92483305-92483305 on Build GRCh38
rs759078700 Chr.1: 92483496-92483496 on Build GRCh38
rs748584016 Chr.1: 92483368-92483368 on Build GRCh38
rs750535225 Chr.1: 92483526-92483526 on Build GRCh38
rs766426942 Chr.1: 92483538-92483538 on Build GRCh38
rs12094593 Chr.1: 92483683-92483683 on Build GRCh38

View Full Product Details