Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92842433-92842941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM69A
Entrez Gene ID
388650
Gene Name
family with sequence similarity 69 member A
Gene Aliases
-
UniGene
Hs.180946
Gene Chromosome Location
Chr.1: 92832729-92961522 on Build GRCh38
Transcripts
NM_001006605.4 NM_001252269.1 NM_001252270.1 NM_001252271.1 NM_001252273.1

SNP Detail

SNP ID Chromosome Location
rs796386150 Chr.1: 92842566-92842567 on Build GRCh38
rs958993284 Chr.1: 92842689-92842692 on Build GRCh38
rs1000449240 Chr.1: 92842540-92842540 on Build GRCh38
rs576668218 Chr.1: 92842628-92842628 on Build GRCh38
rs35488718 Chr.1: 92842478-92842479 on Build GRCh38
rs939156725 Chr.1: 92842466-92842466 on Build GRCh38
rs941237559 Chr.1: 92842493-92842494 on Build GRCh38
rs909177814 Chr.1: 92842704-92842704 on Build GRCh38
rs1009046291 Chr.1: 92842679-92842679 on Build GRCh38
rs892189895 Chr.1: 92842503-92842503 on Build GRCh38
rs548237895 Chr.1: 92842771-92842771 on Build GRCh38
rs746300828 Chr.1: 92842822-92842822 on Build GRCh38
rs138521228 Chr.1: 92842776-92842776 on Build GRCh38
rs757656957 Chr.1: 92842819-92842819 on Build GRCh38
rs1061634 Chr.1: 92842516-92842516 on Build GRCh38
rs939204187 Chr.1: 92842806-92842806 on Build GRCh38
rs181801314 Chr.1: 92842859-92842859 on Build GRCh38
rs958717121 Chr.1: 92842487-92842487 on Build GRCh38
rs892896768 Chr.1: 92842576-92842576 on Build GRCh38
rs1051480665 Chr.1: 92842775-92842775 on Build GRCh38
rs756570186 Chr.1: 92842708-92842708 on Build GRCh38
rs75748040 Chr.1: 92842611-92842611 on Build GRCh38
rs962140259 Chr.1: 92842701-92842701 on Build GRCh38
rs1034595673 Chr.1: 92842669-92842669 on Build GRCh38
rs529818568 Chr.1: 92842716-92842716 on Build GRCh38
rs1020427671 Chr.1: 92842473-92842473 on Build GRCh38
rs541201916 Chr.1: 92842686-92842686 on Build GRCh38
rs781769607 Chr.1: 92842775-92842776 on Build GRCh38
rs980665504 Chr.1: 92842790-92842790 on Build GRCh38
rs11443244 Chr.1: 92842612-92842613 on Build GRCh38
rs920605459 Chr.1: 92842756-92842756 on Build GRCh38
rs765847439 Chr.1: 92842502-92842502 on Build GRCh38
rs972092216 Chr.1: 92842741-92842741 on Build GRCh38
rs892779823 Chr.1: 92842845-92842845 on Build GRCh38
rs933819522 Chr.1: 92842758-92842758 on Build GRCh38
rs553833270 Chr.1: 92842647-92842647 on Build GRCh38
rs141134097 Chr.1: 92842861-92842861 on Build GRCh38
rs972499216 Chr.1: 92842734-92842734 on Build GRCh38
rs368038056 Chr.1: 92842569-92842570 on Build GRCh38
rs571522089 Chr.1: 92842611-92842612 on Build GRCh38
rs971567364 Chr.1: 92842613-92842614 on Build GRCh38
rs34993468 Chr.1: 92842613-92842613 on Build GRCh38
rs189403253 Chr.1: 92842575-92842575 on Build GRCh38
rs1029198435 Chr.1: 92842599-92842599 on Build GRCh38
rs1041472199 Chr.1: 92842874-92842874 on Build GRCh38
rs953534327 Chr.1: 92842659-92842659 on Build GRCh38
rs1009958380 Chr.1: 92842586-92842586 on Build GRCh38
rs889470940 Chr.1: 92842556-92842556 on Build GRCh38
rs764732161 Chr.1: 92842762-92842762 on Build GRCh38
rs1056141976 Chr.1: 92842476-92842476 on Build GRCh38
rs926504787 Chr.1: 92842797-92842797 on Build GRCh38
rs981690697 Chr.1: 92842486-92842486 on Build GRCh38
rs754224559 Chr.1: 92842492-92842495 on Build GRCh38
rs35649260 Chr.1: 92842614-92842614 on Build GRCh38
rs1003561245 Chr.1: 92842626-92842626 on Build GRCh38
rs113760248 Chr.1: 92842486-92842486 on Build GRCh38
rs945126691 Chr.1: 92842868-92842868 on Build GRCh38
rs1043821737 Chr.1: 92842918-92842918 on Build GRCh38
rs1019053391 Chr.1: 92842700-92842700 on Build GRCh38
rs761457699 Chr.1: 92842501-92842506 on Build GRCh38
rs1054132523 Chr.1: 92842830-92842831 on Build GRCh38
rs185134144 Chr.1: 92842507-92842507 on Build GRCh38
rs985310987 Chr.1: 92842490-92842490 on Build GRCh38
rs1010010721 Chr.1: 92842862-92842863 on Build GRCh38
rs904795859 Chr.1: 92842539-92842539 on Build GRCh38
rs376231510 Chr.1: 92842478-92842478 on Build GRCh38
rs909293677 Chr.1: 92842491-92842491 on Build GRCh38

View Full Product Details