Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29310724-29311228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS2L1
Entrez Gene ID
10634
Gene Name
growth arrest specific 2 like 1
Gene Aliases
GAR22
UniGene
Hs.322852
Gene Chromosome Location
Chr.22: 29306618-29312789 on Build GRCh38
Transcripts
NM_001278730.1 NM_006478.4 NM_152236.2 XM_011529824.1 XM_011529825.1 XM_017028533.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4963246 c.951G>C p.R317R Chr.22: 29310939-29310939 on Build GRCh38
6669384 c.941G>A p.R314Q Chr.22: 29310929-29310929 on Build GRCh38
5016676 c.983_984insA p.E330fs*>9 Chr.22: 29310971-29310972 on Build GRCh38
1616337 c.926C>T p.P309L Chr.22: 29310914-29310914 on Build GRCh38
7161065 c.882G>C p.Q294H Chr.22: 29310870-29310870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375768444 Chr.22: 29310885-29310885 on Build GRCh38
rs755687063 Chr.22: 29310758-29310758 on Build GRCh38
rs778796753 Chr.22: 29310772-29310772 on Build GRCh38
rs202200489 Chr.22: 29310985-29310985 on Build GRCh38
rs752085097 Chr.22: 29310819-29310819 on Build GRCh38
rs370162582 Chr.22: 29310912-29310912 on Build GRCh38
rs769488137 Chr.22: 29311107-29311107 on Build GRCh38
rs867817981 Chr.22: 29310938-29310938 on Build GRCh38
rs771984129 Chr.22: 29310777-29310777 on Build GRCh38
rs756838859 Chr.22: 29311039-29311039 on Build GRCh38
rs757066429 Chr.22: 29311029-29311029 on Build GRCh38
rs957090698 Chr.22: 29311058-29311058 on Build GRCh38
rs568890997 Chr.22: 29310833-29310833 on Build GRCh38
rs971802163 Chr.22: 29311169-29311169 on Build GRCh38
rs1026264695 Chr.22: 29311181-29311181 on Build GRCh38
rs762223857 Chr.22: 29310821-29310821 on Build GRCh38
rs777743539 Chr.22: 29310918-29310918 on Build GRCh38
rs542120982 Chr.22: 29310936-29310936 on Build GRCh38
rs777064773 Chr.22: 29310778-29310778 on Build GRCh38
rs777655731 Chr.22: 29310858-29310858 on Build GRCh38
rs1030988884 Chr.22: 29311099-29311099 on Build GRCh38
rs139027113 Chr.22: 29310917-29310917 on Build GRCh38
rs61737777 Chr.22: 29310929-29310929 on Build GRCh38
rs764549668 Chr.22: 29310808-29310808 on Build GRCh38
rs528617895 Chr.22: 29311053-29311053 on Build GRCh38
rs539403505 Chr.22: 29310846-29310846 on Build GRCh38
rs750062239 Chr.22: 29310753-29310753 on Build GRCh38
rs748675322 Chr.22: 29310933-29310933 on Build GRCh38
rs764742161 Chr.22: 29310914-29310914 on Build GRCh38
rs897174842 Chr.22: 29311168-29311168 on Build GRCh38
rs908979313 Chr.22: 29310852-29310852 on Build GRCh38
rs753796600 Chr.22: 29310806-29310807 on Build GRCh38
rs138534671 Chr.22: 29310901-29310901 on Build GRCh38
rs963157702 Chr.22: 29311148-29311148 on Build GRCh38
rs371349829 Chr.22: 29310928-29310928 on Build GRCh38
rs1001423787 Chr.22: 29310892-29310892 on Build GRCh38
rs200255354 Chr.22: 29311049-29311049 on Build GRCh38
rs754474897 Chr.22: 29310838-29310838 on Build GRCh38
rs908844610 Chr.22: 29311141-29311141 on Build GRCh38
rs991848193 Chr.22: 29311005-29311005 on Build GRCh38
rs766684667 Chr.22: 29310845-29310845 on Build GRCh38
rs572882535 Chr.22: 29310896-29310896 on Build GRCh38
rs774528060 Chr.22: 29311130-29311131 on Build GRCh38
rs760349118 Chr.22: 29310900-29310900 on Build GRCh38
rs773763317 Chr.22: 29311040-29311040 on Build GRCh38
rs200449509 Chr.22: 29310822-29310822 on Build GRCh38
rs772000384 Chr.22: 29310849-29310849 on Build GRCh38
rs868863984 Chr.22: 29310927-29310927 on Build GRCh38
rs959117593 Chr.22: 29310974-29310974 on Build GRCh38
rs997801819 Chr.22: 29311081-29311081 on Build GRCh38
rs1056936174 Chr.22: 29311198-29311198 on Build GRCh38
rs749259779 Chr.22: 29310764-29310764 on Build GRCh38
rs779636685 Chr.22: 29310832-29310832 on Build GRCh38
rs758047109 Chr.22: 29311019-29311022 on Build GRCh38
rs201760797 Chr.22: 29310744-29310744 on Build GRCh38
rs763360466 Chr.22: 29311037-29311038 on Build GRCh38
rs746116850 Chr.22: 29310862-29310862 on Build GRCh38
rs746842839 Chr.22: 29310923-29310923 on Build GRCh38
rs778992140 Chr.22: 29311038-29311038 on Build GRCh38
rs866799249 Chr.22: 29310869-29310869 on Build GRCh38
rs752375604 Chr.22: 29310891-29310892 on Build GRCh38
rs759146077 Chr.22: 29311045-29311045 on Build GRCh38
rs61737776 Chr.22: 29310937-29310937 on Build GRCh38
rs780199857 Chr.22: 29310926-29310926 on Build GRCh38
rs776157308 Chr.22: 29310955-29310955 on Build GRCh38
rs757054075 Chr.22: 29310925-29310925 on Build GRCh38
rs752215253 Chr.22: 29310984-29310984 on Build GRCh38
rs549624789 Chr.22: 29310780-29310780 on Build GRCh38
rs145956439 Chr.22: 29310898-29310898 on Build GRCh38
rs763549289 Chr.22: 29310805-29310805 on Build GRCh38
rs575253990 Chr.22: 29311033-29311033 on Build GRCh38
rs1045023516 Chr.22: 29310992-29310992 on Build GRCh38
rs376426006 Chr.22: 29310765-29310765 on Build GRCh38
rs750079919 Chr.22: 29310823-29310823 on Build GRCh38
rs116700559 Chr.22: 29310895-29310895 on Build GRCh38
rs925526154 Chr.22: 29311047-29311047 on Build GRCh38
rs200876928 Chr.22: 29310794-29310794 on Build GRCh38
rs769868490 Chr.22: 29310870-29310870 on Build GRCh38
rs564502100 Chr.22: 29311043-29311043 on Build GRCh38
rs746450559 Chr.22: 29311038-29311039 on Build GRCh38
rs777008077 Chr.22: 29311044-29311044 on Build GRCh38
rs375963140 Chr.22: 29310954-29310954 on Build GRCh38
rs985794524 Chr.22: 29311134-29311134 on Build GRCh38
rs768767404 Chr.22: 29310887-29310887 on Build GRCh38
rs550907092 Chr.22: 29311182-29311182 on Build GRCh38
rs372445961 Chr.22: 29310824-29310824 on Build GRCh38
rs1006053723 Chr.22: 29311187-29311187 on Build GRCh38
rs919108845 Chr.22: 29310773-29310773 on Build GRCh38
rs994570351 Chr.22: 29311172-29311172 on Build GRCh38
rs1012673264 Chr.22: 29310988-29310988 on Build GRCh38
rs755067226 Chr.22: 29310907-29310907 on Build GRCh38
rs752697484 Chr.22: 29310915-29310915 on Build GRCh38
rs779562868 Chr.22: 29311035-29311035 on Build GRCh38
rs917597284 Chr.22: 29311006-29311006 on Build GRCh38
rs748264714 Chr.22: 29311039-29311039 on Build GRCh38
rs866861278 Chr.22: 29311031-29311031 on Build GRCh38
rs944114986 Chr.22: 29311130-29311130 on Build GRCh38
rs181138891 Chr.22: 29311121-29311121 on Build GRCh38
rs754764166 Chr.22: 29310813-29310814 on Build GRCh38
rs926372639 Chr.22: 29310812-29310812 on Build GRCh38
rs1043006289 Chr.22: 29310834-29310834 on Build GRCh38
rs764176497 Chr.22: 29311008-29311008 on Build GRCh38
rs751530307 Chr.22: 29311010-29311010 on Build GRCh38
rs775384927 Chr.22: 29310804-29310804 on Build GRCh38
rs771139403 Chr.22: 29310960-29310960 on Build GRCh38
rs201715173 Chr.22: 29311042-29311042 on Build GRCh38
rs373123433 Chr.22: 29310762-29310762 on Build GRCh38
rs370542455 Chr.22: 29310786-29310786 on Build GRCh38
rs938499222 Chr.22: 29311186-29311186 on Build GRCh38
rs889921344 Chr.22: 29311103-29311103 on Build GRCh38
rs547031376 Chr.22: 29311071-29311071 on Build GRCh38
rs778618265 Chr.22: 29310884-29310884 on Build GRCh38
rs755178203 Chr.22: 29311037-29311037 on Build GRCh38
rs372837332 Chr.22: 29311052-29311052 on Build GRCh38
rs764679654 Chr.22: 29310965-29310965 on Build GRCh38
rs370281351 Chr.22: 29310964-29310964 on Build GRCh38
rs775750363 Chr.22: 29310879-29310879 on Build GRCh38
rs529068456 Chr.22: 29311123-29311123 on Build GRCh38

View Full Product Details