Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29441326-29441830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFPL1
Entrez Gene ID
5988
Gene Name
ret finger protein like 1
Gene Aliases
RNF78
UniGene
Hs.648249
Gene Chromosome Location
Chr.22: 29403031-29442491 on Build GRCh38
Transcripts
NM_021026.2 XM_011530298.2 XM_011530299.2 XM_017028900.1 XM_017028901.1 XM_017028902.1 NR_002727.2

Gene Symbol
RFPL1S
Entrez Gene ID
10740
Gene Name
RFPL1 antisense RNA 1
Gene Aliases
NCRNA00006, RFPL1-AS, RFPL1-AS1, RFPL1AS
UniGene
Hs.167750
Gene Chromosome Location
Chr.22: 29437015-29442129 on Build GRCh38
Transcripts
NM_021026.2 XM_011530298.2 XM_011530299.2 XM_017028900.1 XM_017028901.1 XM_017028902.1 NR_002727.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
149276 c.535C>T p.R179C Chr.22: 29441703-29441703 on Build GRCh38
1415508 c.493G>A p.V165M Chr.22: 29441661-29441661 on Build GRCh38
3749761 c.412C>T p.L138F Chr.22: 29441580-29441580 on Build GRCh38
3912575 c.576G>A p.W192* Chr.22: 29441744-29441744 on Build GRCh38
1565972 c.387G>A p.L129L Chr.22: 29441555-29441555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374370867 Chr.22: 29441743-29441743 on Build GRCh38
rs372400572 Chr.22: 29441561-29441561 on Build GRCh38
rs769285543 Chr.22: 29441651-29441651 on Build GRCh38
rs3804076 Chr.22: 29441548-29441548 on Build GRCh38
rs769340571 Chr.22: 29441723-29441723 on Build GRCh38
rs757566785 Chr.22: 29441781-29441781 on Build GRCh38
rs765197061 Chr.22: 29441776-29441776 on Build GRCh38
rs749377519 Chr.22: 29441718-29441718 on Build GRCh38
rs764755856 Chr.22: 29441579-29441579 on Build GRCh38
rs745998736 Chr.22: 29441640-29441640 on Build GRCh38
rs577925684 Chr.22: 29441724-29441724 on Build GRCh38
rs775942038 Chr.22: 29441649-29441650 on Build GRCh38
rs772589466 Chr.22: 29441741-29441741 on Build GRCh38
rs770614019 Chr.22: 29441766-29441766 on Build GRCh38
rs556139209 Chr.22: 29441688-29441688 on Build GRCh38
rs199689739 Chr.22: 29441798-29441798 on Build GRCh38
rs763559961 Chr.22: 29441704-29441704 on Build GRCh38
rs3842466 Chr.22: 29441583-29441585 on Build GRCh38
rs148461269 Chr.22: 29441658-29441658 on Build GRCh38
rs140801070 Chr.22: 29441713-29441713 on Build GRCh38
rs758156629 Chr.22: 29441702-29441702 on Build GRCh38
rs113035887 Chr.22: 29441496-29441496 on Build GRCh38
rs567075255 Chr.22: 29441628-29441628 on Build GRCh38
rs771073234 Chr.22: 29441522-29441522 on Build GRCh38
rs373256620 Chr.22: 29441660-29441660 on Build GRCh38
rs776282061 Chr.22: 29441524-29441524 on Build GRCh38
rs78769943 Chr.22: 29441577-29441577 on Build GRCh38
rs748629312 Chr.22: 29441739-29441739 on Build GRCh38
rs1047145651 Chr.22: 29441470-29441470 on Build GRCh38
rs781367721 Chr.22: 29441639-29441639 on Build GRCh38
rs779997850 Chr.22: 29441717-29441717 on Build GRCh38
rs977381144 Chr.22: 29441597-29441597 on Build GRCh38
rs987861736 Chr.22: 29441478-29441478 on Build GRCh38
rs757067958 Chr.22: 29441606-29441606 on Build GRCh38
rs762825785 Chr.22: 29441792-29441792 on Build GRCh38
rs765874853 Chr.22: 29441611-29441611 on Build GRCh38
rs757533010 Chr.22: 29441634-29441634 on Build GRCh38
rs531245500 Chr.22: 29441389-29441389 on Build GRCh38
rs753907886 Chr.22: 29441805-29441805 on Build GRCh38
rs371079248 Chr.22: 29441689-29441689 on Build GRCh38
rs150980053 Chr.22: 29441704-29441704 on Build GRCh38
rs755084718 Chr.22: 29441667-29441667 on Build GRCh38
rs754887014 Chr.22: 29441622-29441622 on Build GRCh38
rs752660368 Chr.22: 29441779-29441779 on Build GRCh38
rs754763262 Chr.22: 29441500-29441500 on Build GRCh38
rs753323830 Chr.22: 29441675-29441675 on Build GRCh38
rs778630330 Chr.22: 29441501-29441501 on Build GRCh38
rs779087751 Chr.22: 29441806-29441806 on Build GRCh38
rs200883191 Chr.22: 29441709-29441709 on Build GRCh38
rs764349354 Chr.22: 29441748-29441755 on Build GRCh38
rs142632131 Chr.22: 29441703-29441703 on Build GRCh38
rs143498009 Chr.22: 29441618-29441618 on Build GRCh38
rs774918682 Chr.22: 29441656-29441656 on Build GRCh38
rs753743834 Chr.22: 29441612-29441612 on Build GRCh38
rs879906073 Chr.22: 29441414-29441414 on Build GRCh38
rs16987628 Chr.22: 29441610-29441610 on Build GRCh38
rs147773965 Chr.22: 29441768-29441768 on Build GRCh38
rs752027912 Chr.22: 29441770-29441774 on Build GRCh38
rs571279619 Chr.22: 29441551-29441551 on Build GRCh38
rs769792808 Chr.22: 29441643-29441643 on Build GRCh38
rs759457554 Chr.22: 29441680-29441680 on Build GRCh38
rs933652692 Chr.22: 29441463-29441463 on Build GRCh38
rs71324764 Chr.22: 29441578-29441580 on Build GRCh38
rs759024645 Chr.22: 29441775-29441775 on Build GRCh38
rs762249369 Chr.22: 29441657-29441657 on Build GRCh38
rs780304763 Chr.22: 29441802-29441802 on Build GRCh38
rs149680066 Chr.22: 29441749-29441749 on Build GRCh38
rs901450255 Chr.22: 29441355-29441355 on Build GRCh38
rs778685247 Chr.22: 29441623-29441623 on Build GRCh38
rs368609774 Chr.22: 29441800-29441800 on Build GRCh38
rs749768595 Chr.22: 29441650-29441650 on Build GRCh38
rs775826347 Chr.22: 29441552-29441552 on Build GRCh38
rs748228731 Chr.22: 29441690-29441690 on Build GRCh38
rs780195784 Chr.22: 29441661-29441661 on Build GRCh38
rs750004982 Chr.22: 29441607-29441607 on Build GRCh38
rs769214763 Chr.22: 29441635-29441635 on Build GRCh38
rs776758768 Chr.22: 29441538-29441538 on Build GRCh38
rs767270016 Chr.22: 29441605-29441605 on Build GRCh38
rs747246215 Chr.22: 29441514-29441514 on Build GRCh38
rs79401928 Chr.22: 29441580-29441580 on Build GRCh38
rs761628262 Chr.22: 29441595-29441595 on Build GRCh38
rs60550593 Chr.22: 29441649-29441649 on Build GRCh38
rs10211999 Chr.22: 29441594-29441594 on Build GRCh38

View Full Product Details