Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29788887-29789395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASCC2
Entrez Gene ID
84164
Gene Name
activating signal cointegrator 1 complex subunit 2
Gene Aliases
ASC1p100, p100
UniGene
Hs.731754
Gene Chromosome Location
Chr.22: 29788608-29838304 on Build GRCh38
Transcripts
NM_001242906.1 NM_032204.4 XM_005261775.2 XM_011530442.2 XM_011530443.2 XM_011530444.2 XM_011530445.2 XM_011530446.2 XM_011530448.2 XM_011530449.2 XM_011530450.2 XM_011530451.2 XM_011530452.2 XM_011530453.2 XM_011530454.2 XM_011530455.2 XM_017028991.1 XM_017028992.1 XM_017028993.1 XM_017028994.1 XM_017028995.1 XM_017028996.1 XM_017028997.1 XM_017028998.1 XM_017028999.1 XM_017029000.1 XM_017029001.1 XM_017029002.1 XM_017029003.1 XM_017029004.1 XM_017029005.1 XM_017029006.1 XM_017029007.1 XM_017029008.1 XM_017029009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4977226 c.2216A>G p.N739S Chr.22: 29789071-29789071 on Build GRCh38
5521016 c.2164G>A p.E722K Chr.22: 29789123-29789123 on Build GRCh38
5953628 c.2262G>A p.M754I Chr.22: 29789025-29789025 on Build GRCh38
6605626 c.2224C>T p.R742W Chr.22: 29789063-29789063 on Build GRCh38
6605625 c.2239G>A p.D747N Chr.22: 29789048-29789048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767831875 Chr.22: 29789002-29789002 on Build GRCh38
rs777789941 Chr.22: 29789137-29789137 on Build GRCh38
rs923421600 Chr.22: 29788957-29788957 on Build GRCh38
rs750799199 Chr.22: 29789009-29789009 on Build GRCh38
rs746904268 Chr.22: 29789143-29789143 on Build GRCh38
rs781203641 Chr.22: 29789225-29789225 on Build GRCh38
rs778352767 Chr.22: 29789123-29789123 on Build GRCh38
rs1047511741 Chr.22: 29788966-29788966 on Build GRCh38
rs569798788 Chr.22: 29789063-29789063 on Build GRCh38
rs7954 Chr.22: 29788961-29788961 on Build GRCh38
rs761111398 Chr.22: 29789178-29789178 on Build GRCh38
rs1011053985 Chr.22: 29788999-29788999 on Build GRCh38
rs748093374 Chr.22: 29789045-29789045 on Build GRCh38
rs112954721 Chr.22: 29788932-29788932 on Build GRCh38
rs770802570 Chr.22: 29789061-29789061 on Build GRCh38
rs909991173 Chr.22: 29789130-29789130 on Build GRCh38
rs561036256 Chr.22: 29789310-29789310 on Build GRCh38
rs957365365 Chr.22: 29788914-29788914 on Build GRCh38
rs776546215 Chr.22: 29789225-29789225 on Build GRCh38
rs373433240 Chr.22: 29789107-29789107 on Build GRCh38
rs902256047 Chr.22: 29789048-29789048 on Build GRCh38
rs758316876 Chr.22: 29789133-29789133 on Build GRCh38
rs139016848 Chr.22: 29789125-29789125 on Build GRCh38
rs752220173 Chr.22: 29789124-29789124 on Build GRCh38
rs776975840 Chr.22: 29789180-29789180 on Build GRCh38
rs748049554 Chr.22: 29788962-29788962 on Build GRCh38
rs752244074 Chr.22: 29789200-29789200 on Build GRCh38
rs762325834 Chr.22: 29789000-29789000 on Build GRCh38
rs543364505 Chr.22: 29789312-29789312 on Build GRCh38
rs779699735 Chr.22: 29789029-29789029 on Build GRCh38
rs749582866 Chr.22: 29789148-29789148 on Build GRCh38
rs367692949 Chr.22: 29788971-29788971 on Build GRCh38
rs902462491 Chr.22: 29789267-29789267 on Build GRCh38
rs2269971 Chr.22: 29789281-29789281 on Build GRCh38
rs1054767449 Chr.22: 29789207-29789207 on Build GRCh38
rs754488227 Chr.22: 29789035-29789035 on Build GRCh38
rs554784267 Chr.22: 29789083-29789083 on Build GRCh38
rs930458946 Chr.22: 29788951-29788951 on Build GRCh38
rs773046865 Chr.22: 29788965-29788965 on Build GRCh38
rs757251260 Chr.22: 29789224-29789224 on Build GRCh38
rs143919306 Chr.22: 29789110-29789110 on Build GRCh38
rs1018560653 Chr.22: 29789069-29789069 on Build GRCh38
rs368440043 Chr.22: 29788995-29788995 on Build GRCh38
rs774480702 Chr.22: 29789079-29789079 on Build GRCh38
rs558636205 Chr.22: 29789093-29789093 on Build GRCh38
rs757124680 Chr.22: 29789144-29789144 on Build GRCh38
rs1029338654 Chr.22: 29789353-29789353 on Build GRCh38
rs759743064 Chr.22: 29788968-29788968 on Build GRCh38
rs542881588 Chr.22: 29789279-29789279 on Build GRCh38
rs775761549 Chr.22: 29788982-29788982 on Build GRCh38
rs750079104 Chr.22: 29789200-29789204 on Build GRCh38
rs558631179 Chr.22: 29788927-29788927 on Build GRCh38
rs768041884 Chr.22: 29789092-29789092 on Build GRCh38
rs767351562 Chr.22: 29788982-29788983 on Build GRCh38
rs1040068138 Chr.22: 29789291-29789291 on Build GRCh38
rs753447337 Chr.22: 29789034-29789034 on Build GRCh38
rs768554781 Chr.22: 29789115-29789115 on Build GRCh38
rs140034829 Chr.22: 29789044-29789044 on Build GRCh38
rs748278181 Chr.22: 29789153-29789153 on Build GRCh38
rs572622935 Chr.22: 29789244-29789244 on Build GRCh38
rs377260532 Chr.22: 29789106-29789106 on Build GRCh38
rs755809868 Chr.22: 29789021-29789021 on Build GRCh38
rs748090819 Chr.22: 29789239-29789239 on Build GRCh38
rs757018121 Chr.22: 29788971-29788972 on Build GRCh38
rs201748529 Chr.22: 29789209-29789209 on Build GRCh38
rs916218879 Chr.22: 29789229-29789229 on Build GRCh38
rs768922568 Chr.22: 29789149-29789149 on Build GRCh38
rs6006255 Chr.22: 29789082-29789082 on Build GRCh38
rs745631809 Chr.22: 29789234-29789234 on Build GRCh38
rs182901427 Chr.22: 29789337-29789337 on Build GRCh38
rs537037042 Chr.22: 29789074-29789074 on Build GRCh38
rs946901637 Chr.22: 29789238-29789238 on Build GRCh38
rs370623853 Chr.22: 29788983-29788983 on Build GRCh38
rs774338814 Chr.22: 29789150-29789150 on Build GRCh38
rs781078504 Chr.22: 29789147-29789147 on Build GRCh38
rs368423719 Chr.22: 29789154-29789154 on Build GRCh38
rs374255582 Chr.22: 29788963-29788963 on Build GRCh38
rs146167118 Chr.22: 29788993-29788993 on Build GRCh38
rs775888160 Chr.22: 29789067-29789067 on Build GRCh38
rs766787036 Chr.22: 29789179-29789179 on Build GRCh38
rs755790631 Chr.22: 29789207-29789208 on Build GRCh38
rs757959494 Chr.22: 29789201-29789201 on Build GRCh38
rs565880057 Chr.22: 29788929-29788929 on Build GRCh38
rs751558882 Chr.22: 29789212-29789212 on Build GRCh38
rs763203660 Chr.22: 29789068-29789068 on Build GRCh38
rs764921465 Chr.22: 29789198-29789198 on Build GRCh38
rs990117562 Chr.22: 29788917-29788917 on Build GRCh38
rs372345494 Chr.22: 29789049-29789049 on Build GRCh38
rs984614605 Chr.22: 29788952-29788952 on Build GRCh38
rs1053568641 Chr.22: 29789304-29789304 on Build GRCh38
rs759226865 Chr.22: 29789182-29789182 on Build GRCh38
rs28544456 Chr.22: 29789057-29789057 on Build GRCh38
rs939326269 Chr.22: 29789171-29789171 on Build GRCh38
rs774561617 Chr.22: 29789362-29789363 on Build GRCh38
rs974641618 Chr.22: 29788921-29788921 on Build GRCh38
rs531737086 Chr.22: 29789311-29789311 on Build GRCh38
rs901521034 Chr.22: 29789333-29789333 on Build GRCh38
rs1017713893 Chr.22: 29789373-29789373 on Build GRCh38
rs902313306 Chr.22: 29789240-29789240 on Build GRCh38
rs746798422 Chr.22: 29789051-29789051 on Build GRCh38
rs941776547 Chr.22: 29789206-29789206 on Build GRCh38

View Full Product Details