Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 32229110-32229512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC5A4
Entrez Gene ID
6527
Gene Name
solute carrier family 5 member 4
Gene Aliases
DJ90G24.4, SAAT1, SGLT3
UniGene
Hs.130101
Gene Chromosome Location
Chr.22: 32218423-32355057 on Build GRCh38
Transcripts
NM_014227.2 XM_006724308.3 XM_011530342.2 XM_011530343.2 XM_011530344.2 XM_017028920.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1682225 c.1250C>T p.A417V Chr.22: 32229224-32229224 on Build GRCh38
726544 c.1233C>T p.T411T Chr.22: 32229241-32229241 on Build GRCh38
4425870 c.1251G>A p.A417A Chr.22: 32229223-32229223 on Build GRCh38
4854989 c.1136G>C p.R379P Chr.22: 32229338-32229338 on Build GRCh38
1161423 c.1241G>A p.R414Q Chr.22: 32229233-32229233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199753375 Chr.22: 32229365-32229365 on Build GRCh38
rs766094490 Chr.22: 32229244-32229244 on Build GRCh38
rs202084255 Chr.22: 32229353-32229353 on Build GRCh38
rs759749538 Chr.22: 32229323-32229323 on Build GRCh38
rs367930190 Chr.22: 32229183-32229183 on Build GRCh38
rs374348399 Chr.22: 32229145-32229145 on Build GRCh38
rs751258625 Chr.22: 32229334-32229334 on Build GRCh38
rs750533251 Chr.22: 32229465-32229465 on Build GRCh38
rs748464500 Chr.22: 32229193-32229193 on Build GRCh38
rs188087739 Chr.22: 32229360-32229360 on Build GRCh38
rs747178458 Chr.22: 32229274-32229274 on Build GRCh38
rs780502091 Chr.22: 32229243-32229243 on Build GRCh38
rs775299064 Chr.22: 32229382-32229382 on Build GRCh38
rs200221311 Chr.22: 32229176-32229176 on Build GRCh38
rs759567213 Chr.22: 32229298-32229298 on Build GRCh38
rs765052623 Chr.22: 32229303-32229303 on Build GRCh38
rs201065219 Chr.22: 32229164-32229164 on Build GRCh38
rs756557197 Chr.22: 32229234-32229234 on Build GRCh38
rs769578771 Chr.22: 32229375-32229375 on Build GRCh38
rs746200908 Chr.22: 32229167-32229167 on Build GRCh38
rs770778449 Chr.22: 32229277-32229277 on Build GRCh38
rs553643566 Chr.22: 32229455-32229455 on Build GRCh38
rs754133867 Chr.22: 32229339-32229339 on Build GRCh38
rs180925414 Chr.22: 32229221-32229221 on Build GRCh38
rs781079504 Chr.22: 32229369-32229369 on Build GRCh38
rs745681694 Chr.22: 32229373-32229373 on Build GRCh38
rs5998322 Chr.22: 32229223-32229223 on Build GRCh38
rs779319917 Chr.22: 32229346-32229346 on Build GRCh38
rs539716961 Chr.22: 32229439-32229439 on Build GRCh38
rs755553343 Chr.22: 32229345-32229345 on Build GRCh38
rs762387986 Chr.22: 32229320-32229320 on Build GRCh38
rs772166813 Chr.22: 32229194-32229194 on Build GRCh38
rs759544577 Chr.22: 32229166-32229167 on Build GRCh38
rs765029888 Chr.22: 32229234-32229235 on Build GRCh38
rs372543267 Chr.22: 32229196-32229196 on Build GRCh38
rs370114297 Chr.22: 32229227-32229227 on Build GRCh38
rs879083808 Chr.22: 32229411-32229411 on Build GRCh38
rs188360318 Chr.22: 32229195-32229195 on Build GRCh38
rs748146452 Chr.22: 32229254-32229254 on Build GRCh38
rs772099258 Chr.22: 32229265-32229265 on Build GRCh38
rs144357030 Chr.22: 32229273-32229273 on Build GRCh38
rs147703693 Chr.22: 32229310-32229310 on Build GRCh38
rs970284096 Chr.22: 32229239-32229239 on Build GRCh38
rs752543253 Chr.22: 32229359-32229359 on Build GRCh38
rs759385971 Chr.22: 32229220-32229220 on Build GRCh38
rs563541123 Chr.22: 32229184-32229184 on Build GRCh38
rs373289806 Chr.22: 32229233-32229233 on Build GRCh38
rs771118078 Chr.22: 32229367-32229367 on Build GRCh38
rs776395556 Chr.22: 32229284-32229284 on Build GRCh38
rs756967934 Chr.22: 32229337-32229337 on Build GRCh38
rs758336899 Chr.22: 32229374-32229374 on Build GRCh38
rs896509274 Chr.22: 32229335-32229335 on Build GRCh38
rs780726775 Chr.22: 32229178-32229178 on Build GRCh38
rs141751922 Chr.22: 32229250-32229250 on Build GRCh38
rs903138066 Chr.22: 32229370-32229370 on Build GRCh38
rs776564701 Chr.22: 32229203-32229203 on Build GRCh38
rs377053159 Chr.22: 32229199-32229199 on Build GRCh38
rs755080959 Chr.22: 32229401-32229401 on Build GRCh38
rs757973477 Chr.22: 32229153-32229153 on Build GRCh38
rs562170792 Chr.22: 32229146-32229146 on Build GRCh38
rs760482444 Chr.22: 32229229-32229229 on Build GRCh38
rs201129927 Chr.22: 32229322-32229322 on Build GRCh38
rs752459330 Chr.22: 32229150-32229150 on Build GRCh38
rs142416109 Chr.22: 32229338-32229338 on Build GRCh38
rs748640245 Chr.22: 32229349-32229349 on Build GRCh38
rs758108963 Chr.22: 32229224-32229224 on Build GRCh38
rs774293686 Chr.22: 32229189-32229189 on Build GRCh38
rs932540964 Chr.22: 32229410-32229410 on Build GRCh38

View Full Product Details