Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 35265259-35265782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGXB4
Entrez Gene ID
10042
Gene Name
HMG-box containing 4
Gene Aliases
HMG2L1, HMGBCG, THC211630
UniGene
Hs.588815
Gene Chromosome Location
Chr.22: 35257274-35295807 on Build GRCh38
Transcripts
NM_001003681.2 NR_027780.1 XM_006724100.3 XM_006724101.3 XM_006724102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5714413 c.945A>T p.E315D Chr.22: 35265333-35265333 on Build GRCh38
6877181 c.1090C>T p.P364S Chr.22: 35265478-35265478 on Build GRCh38
2939404 c.960G>T p.K320N Chr.22: 35265348-35265348 on Build GRCh38
5742104 c.1021A>G p.K341E Chr.22: 35265409-35265409 on Build GRCh38
5921847 c.941G>T p.R314L Chr.22: 35265329-35265329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778144838 Chr.22: 35265498-35265498 on Build GRCh38
rs201132869 Chr.22: 35265396-35265396 on Build GRCh38
rs762914579 Chr.22: 35265343-35265343 on Build GRCh38
rs757738800 Chr.22: 35265502-35265502 on Build GRCh38
rs867115631 Chr.22: 35265560-35265560 on Build GRCh38
rs770934352 Chr.22: 35265430-35265430 on Build GRCh38
rs14659 Chr.22: 35265431-35265431 on Build GRCh38
rs749919403 Chr.22: 35265478-35265478 on Build GRCh38
rs977250362 Chr.22: 35265720-35265720 on Build GRCh38
rs755881528 Chr.22: 35265486-35265486 on Build GRCh38
rs974899867 Chr.22: 35265561-35265561 on Build GRCh38
rs759704863 Chr.22: 35265437-35265437 on Build GRCh38
rs77847678 Chr.22: 35265305-35265305 on Build GRCh38
rs745486695 Chr.22: 35265498-35265500 on Build GRCh38
rs760260091 Chr.22: 35265604-35265604 on Build GRCh38
rs773519729 Chr.22: 35265580-35265581 on Build GRCh38
rs940949641 Chr.22: 35265669-35265669 on Build GRCh38
rs374649728 Chr.22: 35265328-35265328 on Build GRCh38
rs28522155 Chr.22: 35265523-35265523 on Build GRCh38
rs774994177 Chr.22: 35265566-35265568 on Build GRCh38
rs76572304 Chr.22: 35265551-35265551 on Build GRCh38
rs573464766 Chr.22: 35265329-35265329 on Build GRCh38
rs776719649 Chr.22: 35265434-35265434 on Build GRCh38
rs768348738 Chr.22: 35265529-35265529 on Build GRCh38
rs201799099 Chr.22: 35265500-35265500 on Build GRCh38
rs992811244 Chr.22: 35265470-35265470 on Build GRCh38
rs899372111 Chr.22: 35265708-35265708 on Build GRCh38
rs968950499 Chr.22: 35265697-35265697 on Build GRCh38
rs774159141 Chr.22: 35265466-35265466 on Build GRCh38
rs772765503 Chr.22: 35265416-35265416 on Build GRCh38
rs749813736 Chr.22: 35265561-35265561 on Build GRCh38
rs761752000 Chr.22: 35265473-35265473 on Build GRCh38
rs761697027 Chr.22: 35265552-35265552 on Build GRCh38
rs775682430 Chr.22: 35265513-35265513 on Build GRCh38
rs770057050 Chr.22: 35265511-35265511 on Build GRCh38
rs75595324 Chr.22: 35265614-35265614 on Build GRCh38
rs747487173 Chr.22: 35265428-35265428 on Build GRCh38
rs763993813 Chr.22: 35265362-35265362 on Build GRCh38
rs781713491 Chr.22: 35265648-35265648 on Build GRCh38
rs762721652 Chr.22: 35265447-35265447 on Build GRCh38
rs769970545 Chr.22: 35265438-35265438 on Build GRCh38
rs769388508 Chr.22: 35265550-35265551 on Build GRCh38
rs766118403 Chr.22: 35265491-35265491 on Build GRCh38
rs753805876 Chr.22: 35265627-35265627 on Build GRCh38
rs762325326 Chr.22: 35265580-35265582 on Build GRCh38
rs764394032 Chr.22: 35265628-35265628 on Build GRCh38
rs71314937 Chr.22: 35265578-35265578 on Build GRCh38
rs1005071135 Chr.22: 35265303-35265303 on Build GRCh38
rs760766503 Chr.22: 35265582-35265583 on Build GRCh38
rs368101268 Chr.22: 35265421-35265421 on Build GRCh38
rs371525087 Chr.22: 35265492-35265492 on Build GRCh38
rs200058620 Chr.22: 35265510-35265510 on Build GRCh38
rs975232457 Chr.22: 35265406-35265411 on Build GRCh38
rs774824276 Chr.22: 35265551-35265551 on Build GRCh38
rs924360569 Chr.22: 35265739-35265739 on Build GRCh38
rs767967598 Chr.22: 35265580-35265583 on Build GRCh38
rs764083722 Chr.22: 35265450-35265450 on Build GRCh38
rs796781409 Chr.22: 35265622-35265622 on Build GRCh38
rs899312352 Chr.22: 35265562-35265565 on Build GRCh38
rs146235655 Chr.22: 35265338-35265338 on Build GRCh38
rs760661153 Chr.22: 35265326-35265326 on Build GRCh38
rs143712163 Chr.22: 35265493-35265493 on Build GRCh38
rs925147863 Chr.22: 35265554-35265554 on Build GRCh38
rs751136807 Chr.22: 35265378-35265378 on Build GRCh38
rs773475840 Chr.22: 35265315-35265315 on Build GRCh38
rs765923155 Chr.22: 35265606-35265606 on Build GRCh38
rs759750637 Chr.22: 35265332-35265332 on Build GRCh38
rs571369776 Chr.22: 35265627-35265627 on Build GRCh38
rs1037949183 Chr.22: 35265670-35265670 on Build GRCh38
rs781165463 Chr.22: 35265507-35265507 on Build GRCh38
rs780881420 Chr.22: 35265367-35265369 on Build GRCh38
rs201865608 Chr.22: 35265374-35265374 on Build GRCh38
rs755788789 Chr.22: 35265394-35265394 on Build GRCh38
rs1017533103 Chr.22: 35265586-35265586 on Build GRCh38
rs201048393 Chr.22: 35265567-35265567 on Build GRCh38
rs528331783 Chr.22: 35265439-35265439 on Build GRCh38
rs766968478 Chr.22: 35265476-35265476 on Build GRCh38
rs74570053 Chr.22: 35265613-35265613 on Build GRCh38
rs960317734 Chr.22: 35265404-35265404 on Build GRCh38
rs769146475 Chr.22: 35265294-35265294 on Build GRCh38
rs772656699 Chr.22: 35265593-35265593 on Build GRCh38
rs757046387 Chr.22: 35265358-35265359 on Build GRCh38
rs748502651 Chr.22: 35265400-35265400 on Build GRCh38
rs766505979 Chr.22: 35265581-35265582 on Build GRCh38
rs559493829 Chr.22: 35265425-35265425 on Build GRCh38
rs749444386 Chr.22: 35265516-35265516 on Build GRCh38
rs749527106 Chr.22: 35265487-35265487 on Build GRCh38
rs748696767 Chr.22: 35265312-35265312 on Build GRCh38
rs148970891 Chr.22: 35265334-35265334 on Build GRCh38
rs1008855865 Chr.22: 35265582-35265585 on Build GRCh38
rs769217942 Chr.22: 35265562-35265564 on Build GRCh38
rs1048268320 Chr.22: 35265577-35265577 on Build GRCh38
rs768436683 Chr.22: 35265379-35265379 on Build GRCh38
rs921664933 Chr.22: 35265632-35265632 on Build GRCh38
rs767468760 Chr.22: 35265580-35265580 on Build GRCh38
rs936513972 Chr.22: 35265556-35265556 on Build GRCh38
rs746743674 Chr.22: 35265354-35265356 on Build GRCh38
rs776506029 Chr.22: 35265644-35265644 on Build GRCh38
rs182885536 Chr.22: 35265658-35265658 on Build GRCh38
rs993634894 Chr.22: 35265564-35265568 on Build GRCh38
rs190257561 Chr.22: 35265380-35265380 on Build GRCh38
rs372642793 Chr.22: 35265624-35265624 on Build GRCh38

View Full Product Details