Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 35553301-35553795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASD2
Entrez Gene ID
23551
Gene Name
RASD family member 2
Gene Aliases
MGC:4834, Rhes, TEM2
UniGene
Hs.474711
Gene Chromosome Location
Chr.22: 35540492-35554003 on Build GRCh38
Transcripts
NM_014310.3 XM_005261442.4 XM_011530040.2 XM_017028701.1 XM_017028702.1

SNP Detail

SNP ID Chromosome Location
rs916104338 Chr.22: 35553691-35553691 on Build GRCh38
rs919306196 Chr.22: 35553646-35553646 on Build GRCh38
rs1022511204 Chr.22: 35553535-35553535 on Build GRCh38
rs377381797 Chr.22: 35553596-35553596 on Build GRCh38
rs999003326 Chr.22: 35553465-35553465 on Build GRCh38
rs111835551 Chr.22: 35553396-35553396 on Build GRCh38
rs906896271 Chr.22: 35553450-35553450 on Build GRCh38
rs986761495 Chr.22: 35553529-35553529 on Build GRCh38
rs111456836 Chr.22: 35553332-35553332 on Build GRCh38
rs1029545390 Chr.22: 35553417-35553417 on Build GRCh38
rs114508755 Chr.22: 35553335-35553335 on Build GRCh38
rs934290055 Chr.22: 35553762-35553762 on Build GRCh38
rs939036054 Chr.22: 35553650-35553650 on Build GRCh38
rs575945877 Chr.22: 35553689-35553689 on Build GRCh38
rs966108367 Chr.22: 35553629-35553629 on Build GRCh38
rs1017714451 Chr.22: 35553507-35553507 on Build GRCh38
rs555995548 Chr.22: 35553688-35553688 on Build GRCh38
rs17722036 Chr.22: 35553574-35553574 on Build GRCh38
rs147106033 Chr.22: 35553648-35553648 on Build GRCh38
rs766165290 Chr.22: 35553605-35553605 on Build GRCh38
rs1046015490 Chr.22: 35553747-35553747 on Build GRCh38
rs763079242 Chr.22: 35553372-35553372 on Build GRCh38
rs930726304 Chr.22: 35553647-35553647 on Build GRCh38
rs928199374 Chr.22: 35553594-35553594 on Build GRCh38
rs1028127228 Chr.22: 35553515-35553515 on Build GRCh38
rs77877826 Chr.22: 35553543-35553543 on Build GRCh38
rs954077190 Chr.22: 35553525-35553525 on Build GRCh38
rs990473330 Chr.22: 35553651-35553651 on Build GRCh38
rs894689195 Chr.22: 35553404-35553404 on Build GRCh38
rs978758935 Chr.22: 35553587-35553587 on Build GRCh38
rs949016574 Chr.22: 35553411-35553411 on Build GRCh38
rs111935750 Chr.22: 35553764-35553764 on Build GRCh38
rs919420896 Chr.22: 35553359-35553359 on Build GRCh38
rs890452373 Chr.22: 35553426-35553426 on Build GRCh38
rs775694927 Chr.22: 35553339-35553339 on Build GRCh38
rs1049264648 Chr.22: 35553373-35553373 on Build GRCh38
rs906861341 Chr.22: 35553761-35553761 on Build GRCh38
rs768978346 Chr.22: 35553514-35553514 on Build GRCh38
rs571194454 Chr.22: 35553485-35553485 on Build GRCh38
rs1045790728 Chr.22: 35553428-35553428 on Build GRCh38
rs762223124 Chr.22: 35553598-35553598 on Build GRCh38
rs141556070 Chr.22: 35553472-35553472 on Build GRCh38
rs892891215 Chr.22: 35553490-35553490 on Build GRCh38
rs184860677 Chr.22: 35553724-35553724 on Build GRCh38
rs112590454 Chr.22: 35553481-35553481 on Build GRCh38
rs553932451 Chr.22: 35553542-35553542 on Build GRCh38
rs972060373 Chr.22: 35553632-35553632 on Build GRCh38
rs554352942 Chr.22: 35553731-35553742 on Build GRCh38

View Full Product Details