Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36265172-36265679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOL1
Entrez Gene ID
8542
Gene Name
apolipoprotein L1
Gene Aliases
APO-L, APOL, APOL-I, FSGS4
UniGene
Hs.114309
Gene Chromosome Location
Chr.22: 36253071-36267531 on Build GRCh38
Transcripts
NM_001136540.1 NM_001136541.1 NM_003661.3 NM_145343.2 XM_005261796.3 XM_011530478.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
79506 c.621C>T p.L207L Chr.22: 36265409-36265409 on Build GRCh38
3992134 c.831C>T p.N277N Chr.22: 36265619-36265619 on Build GRCh38
1725573 c.472A>T p.K158* Chr.22: 36265260-36265260 on Build GRCh38
419573 c.469C>T p.L157L Chr.22: 36265257-36265257 on Build GRCh38
3842599 c.623T>A p.V208D Chr.22: 36265411-36265411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750895237 Chr.22: 36265587-36265587 on Build GRCh38
rs763545816 Chr.22: 36265581-36265581 on Build GRCh38
rs136174 Chr.22: 36265490-36265490 on Build GRCh38
rs753437298 Chr.22: 36265466-36265466 on Build GRCh38
rs566062739 Chr.22: 36265526-36265526 on Build GRCh38
rs747549024 Chr.22: 36265314-36265314 on Build GRCh38
rs982350405 Chr.22: 36265343-36265343 on Build GRCh38
rs751558663 Chr.22: 36265265-36265265 on Build GRCh38
rs150472632 Chr.22: 36265614-36265614 on Build GRCh38
rs781500424 Chr.22: 36265544-36265545 on Build GRCh38
rs371826870 Chr.22: 36265405-36265405 on Build GRCh38
rs775912698 Chr.22: 36265356-36265356 on Build GRCh38
rs139210712 Chr.22: 36265321-36265321 on Build GRCh38
rs750232721 Chr.22: 36265294-36265294 on Build GRCh38
rs776382706 Chr.22: 36265548-36265548 on Build GRCh38
rs375964420 Chr.22: 36265452-36265452 on Build GRCh38
rs147973675 Chr.22: 36265555-36265555 on Build GRCh38
rs116136671 Chr.22: 36265363-36265363 on Build GRCh38
rs149947719 Chr.22: 36265346-36265346 on Build GRCh38
rs550306376 Chr.22: 36265410-36265410 on Build GRCh38
rs757047533 Chr.22: 36265273-36265273 on Build GRCh38
rs749708519 Chr.22: 36265500-36265500 on Build GRCh38
rs73403889 Chr.22: 36265645-36265645 on Build GRCh38
rs771257687 Chr.22: 36265566-36265566 on Build GRCh38
rs541348507 Chr.22: 36265328-36265328 on Build GRCh38
rs773763181 Chr.22: 36265207-36265207 on Build GRCh38
rs760970273 Chr.22: 36265226-36265226 on Build GRCh38
rs373383439 Chr.22: 36265426-36265426 on Build GRCh38
rs770051900 Chr.22: 36265354-36265354 on Build GRCh38
rs761864974 Chr.22: 36265444-36265444 on Build GRCh38
rs199608929 Chr.22: 36265227-36265227 on Build GRCh38
rs767476842 Chr.22: 36265448-36265448 on Build GRCh38
rs757661619 Chr.22: 36265344-36265344 on Build GRCh38
rs995899996 Chr.22: 36265642-36265642 on Build GRCh38
rs752185325 Chr.22: 36265260-36265261 on Build GRCh38
rs867106900 Chr.22: 36265226-36265227 on Build GRCh38
rs136176 Chr.22: 36265600-36265600 on Build GRCh38
rs772560002 Chr.22: 36265204-36265204 on Build GRCh38
rs779639286 Chr.22: 36265299-36265299 on Build GRCh38
rs138853743 Chr.22: 36265649-36265649 on Build GRCh38
rs146280751 Chr.22: 36265312-36265312 on Build GRCh38
rs541546105 Chr.22: 36265430-36265430 on Build GRCh38
rs774540447 Chr.22: 36265441-36265441 on Build GRCh38
rs136175 Chr.22: 36265520-36265520 on Build GRCh38
rs41311348 Chr.22: 36265632-36265632 on Build GRCh38
rs763193282 Chr.22: 36265245-36265245 on Build GRCh38
rs73885316 Chr.22: 36265628-36265628 on Build GRCh38
rs746261971 Chr.22: 36265337-36265337 on Build GRCh38
rs879140668 Chr.22: 36265345-36265345 on Build GRCh38
rs778796242 Chr.22: 36265559-36265559 on Build GRCh38
rs746232040 Chr.22: 36265412-36265412 on Build GRCh38
rs770867795 Chr.22: 36265498-36265498 on Build GRCh38
rs757814781 Chr.22: 36265296-36265296 on Build GRCh38
rs149073330 Chr.22: 36265364-36265364 on Build GRCh38
rs764721621 Chr.22: 36265323-36265323 on Build GRCh38
rs768163564 Chr.22: 36265311-36265311 on Build GRCh38
rs754605430 Chr.22: 36265470-36265470 on Build GRCh38
rs772138899 Chr.22: 36265632-36265632 on Build GRCh38
rs747154822 Chr.22: 36265638-36265638 on Build GRCh38
rs752305718 Chr.22: 36265560-36265560 on Build GRCh38
rs766190062 Chr.22: 36265453-36265453 on Build GRCh38
rs757904186 Chr.22: 36265483-36265483 on Build GRCh38
rs150846072 Chr.22: 36265409-36265409 on Build GRCh38
rs764668096 Chr.22: 36265241-36265241 on Build GRCh38
rs758612193 Chr.22: 36265358-36265358 on Build GRCh38
rs372242460 Chr.22: 36265244-36265244 on Build GRCh38
rs777642912 Chr.22: 36265455-36265455 on Build GRCh38
rs368340430 Chr.22: 36265370-36265370 on Build GRCh38
rs554687721 Chr.22: 36265603-36265603 on Build GRCh38
rs762458687 Chr.22: 36265569-36265569 on Build GRCh38
rs1039347372 Chr.22: 36265268-36265268 on Build GRCh38
rs765954217 Chr.22: 36265401-36265401 on Build GRCh38
rs770907296 Chr.22: 36265531-36265531 on Build GRCh38
rs768995250 Chr.22: 36265515-36265515 on Build GRCh38
rs759828094 Chr.22: 36265229-36265229 on Build GRCh38
rs201739609 Chr.22: 36265472-36265472 on Build GRCh38
rs755917494 Chr.22: 36265398-36265398 on Build GRCh38
rs367662496 Chr.22: 36265228-36265228 on Build GRCh38
rs142955744 Chr.22: 36265633-36265633 on Build GRCh38
rs201870882 Chr.22: 36265301-36265301 on Build GRCh38
rs1027205673 Chr.22: 36265272-36265272 on Build GRCh38
rs188476243 Chr.22: 36265623-36265623 on Build GRCh38
rs755406811 Chr.22: 36265619-36265619 on Build GRCh38
rs778419778 Chr.22: 36265406-36265406 on Build GRCh38
rs759396651 Chr.22: 36265553-36265553 on Build GRCh38
rs757844522 Chr.22: 36265262-36265263 on Build GRCh38
rs183714534 Chr.22: 36265413-36265413 on Build GRCh38
rs752042751 Chr.22: 36265325-36265325 on Build GRCh38
rs752250834 Chr.22: 36265477-36265477 on Build GRCh38
rs775056389 Chr.22: 36265618-36265618 on Build GRCh38
rs764109305 Chr.22: 36265255-36265255 on Build GRCh38
rs146925617 Chr.22: 36265527-36265527 on Build GRCh38
rs763070780 Chr.22: 36265359-36265359 on Build GRCh38
rs944382685 Chr.22: 36265652-36265652 on Build GRCh38
rs779334863 Chr.22: 36265519-36265519 on Build GRCh38
rs944512823 Chr.22: 36265418-36265418 on Build GRCh38
rs375244360 Chr.22: 36265260-36265260 on Build GRCh38
rs761786418 Chr.22: 36265366-36265366 on Build GRCh38
rs574173074 Chr.22: 36265304-36265304 on Build GRCh38
rs1012252761 Chr.22: 36265292-36265292 on Build GRCh38
rs201375679 Chr.22: 36265499-36265499 on Build GRCh38
rs148296684 Chr.22: 36265308-36265308 on Build GRCh38
rs2239785 Chr.22: 36265284-36265284 on Build GRCh38

View Full Product Details