Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38081295-38081837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC16A8
Entrez Gene ID
23539
Gene Name
solute carrier family 16 member 8
Gene Aliases
MCT3, REMP
UniGene
Hs.745256
Gene Chromosome Location
Chr.22: 38078137-38083163 on Build GRCh38
Transcripts
NM_013356.2 XM_017028685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5809975 c.740T>G p.V247G Chr.22: 38081298-38081298 on Build GRCh38
6425713 c.694G>T p.A232S Chr.22: 38081344-38081344 on Build GRCh38
3749767 c.703A>T p.R235W Chr.22: 38081335-38081335 on Build GRCh38
6399965 c.682C>T p.Q228* Chr.22: 38081356-38081356 on Build GRCh38
6401083 c.386C>T p.P129L Chr.22: 38081652-38081652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905158040 Chr.22: 38081812-38081812 on Build GRCh38
rs4289289 Chr.22: 38081335-38081335 on Build GRCh38
rs779974924 Chr.22: 38081349-38081349 on Build GRCh38
rs768720682 Chr.22: 38081399-38081399 on Build GRCh38
rs949476388 Chr.22: 38081776-38081776 on Build GRCh38
rs529402942 Chr.22: 38081654-38081654 on Build GRCh38
rs995701368 Chr.22: 38081374-38081374 on Build GRCh38
rs547544700 Chr.22: 38081666-38081666 on Build GRCh38
rs955330351 Chr.22: 38081612-38081612 on Build GRCh38
rs1045158759 Chr.22: 38081786-38081786 on Build GRCh38
rs935482627 Chr.22: 38081685-38081685 on Build GRCh38
rs937816515 Chr.22: 38081491-38081491 on Build GRCh38
rs763430150 Chr.22: 38081645-38081645 on Build GRCh38
rs996183270 Chr.22: 38081560-38081560 on Build GRCh38
rs1043587124 Chr.22: 38081465-38081465 on Build GRCh38
rs773630609 Chr.22: 38081642-38081642 on Build GRCh38
rs758466043 Chr.22: 38081346-38081346 on Build GRCh38
rs1018317060 Chr.22: 38081618-38081618 on Build GRCh38
rs757131116 Chr.22: 38081341-38081341 on Build GRCh38
rs896184521 Chr.22: 38081320-38081320 on Build GRCh38
rs925576588 Chr.22: 38081672-38081672 on Build GRCh38
rs954191766 Chr.22: 38081398-38081398 on Build GRCh38
rs777137024 Chr.22: 38081332-38081332 on Build GRCh38
rs374567790 Chr.22: 38081364-38081364 on Build GRCh38
rs949131063 Chr.22: 38081730-38081730 on Build GRCh38
rs148254406 Chr.22: 38081582-38081582 on Build GRCh38
rs537522041 Chr.22: 38081782-38081782 on Build GRCh38
rs995463527 Chr.22: 38081397-38081408 on Build GRCh38
rs945257713 Chr.22: 38081461-38081461 on Build GRCh38
rs745797557 Chr.22: 38081345-38081345 on Build GRCh38
rs541520159 Chr.22: 38081326-38081326 on Build GRCh38
rs912638292 Chr.22: 38081419-38081419 on Build GRCh38
rs896586306 Chr.22: 38081361-38081361 on Build GRCh38
rs769218097 Chr.22: 38081768-38081768 on Build GRCh38
rs1052469167 Chr.22: 38081360-38081360 on Build GRCh38
rs1026452789 Chr.22: 38081482-38081499 on Build GRCh38
rs778929954 Chr.22: 38081344-38081344 on Build GRCh38
rs753528029 Chr.22: 38081339-38081339 on Build GRCh38
rs986582430 Chr.22: 38081617-38081617 on Build GRCh38
rs913828695 Chr.22: 38081445-38081445 on Build GRCh38
rs781495654 Chr.22: 38081540-38081540 on Build GRCh38
rs1013258931 Chr.22: 38081506-38081506 on Build GRCh38
rs543643447 Chr.22: 38081598-38081598 on Build GRCh38
rs990616533 Chr.22: 38081703-38081703 on Build GRCh38
rs191281209 Chr.22: 38081652-38081652 on Build GRCh38
rs1037074650 Chr.22: 38081507-38081507 on Build GRCh38
rs905212191 Chr.22: 38081525-38081525 on Build GRCh38
rs763842724 Chr.22: 38081337-38081337 on Build GRCh38
rs541109969 Chr.22: 38081756-38081756 on Build GRCh38
rs896195891 Chr.22: 38081498-38081498 on Build GRCh38
rs189345550 Chr.22: 38081551-38081551 on Build GRCh38
rs759946084 Chr.22: 38081409-38081409 on Build GRCh38
rs896841397 Chr.22: 38081789-38081789 on Build GRCh38
rs965400443 Chr.22: 38081599-38081599 on Build GRCh38
rs898324750 Chr.22: 38081576-38081576 on Build GRCh38
rs963752487 Chr.22: 38081624-38081624 on Build GRCh38
rs917684768 Chr.22: 38081714-38081714 on Build GRCh38
rs545795719 Chr.22: 38081368-38081368 on Build GRCh38
rs985538597 Chr.22: 38081417-38081417 on Build GRCh38
rs950868111 Chr.22: 38081586-38081610 on Build GRCh38
rs937828038 Chr.22: 38081702-38081702 on Build GRCh38
rs1015847191 Chr.22: 38081343-38081343 on Build GRCh38
rs761627924 Chr.22: 38081691-38081691 on Build GRCh38
rs753534531 Chr.22: 38081688-38081688 on Build GRCh38
rs1045442183 Chr.22: 38081495-38081495 on Build GRCh38
rs988296236 Chr.22: 38081754-38081754 on Build GRCh38
rs866048461 Chr.22: 38081317-38081317 on Build GRCh38
rs765699451 Chr.22: 38081663-38081664 on Build GRCh38
rs965389027 Chr.22: 38081435-38081435 on Build GRCh38

View Full Product Details