Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41810051-41810485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC134
Entrez Gene ID
79879
Gene Name
coiled-coil domain containing 134
Gene Aliases
-
UniGene
Hs.474991
Gene Chromosome Location
Chr.22: 41800610-41832164 on Build GRCh38
Transcripts
NM_001304797.1 NM_024821.3 XM_005261748.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1416559 c.226-1G>T p.? Chr.22: 41810206-41810206 on Build GRCh38
6621503 c.268C>T p.L90F Chr.22: 41810249-41810249 on Build GRCh38
70036 c.278G>A p.G93E Chr.22: 41810259-41810259 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375620106 Chr.22: 41810275-41810275 on Build GRCh38
rs73885829 Chr.22: 41810389-41810389 on Build GRCh38
rs996400624 Chr.22: 41810073-41810073 on Build GRCh38
rs769416137 Chr.22: 41810235-41810235 on Build GRCh38
rs1023734565 Chr.22: 41810115-41810115 on Build GRCh38
rs374311596 Chr.22: 41810194-41810194 on Build GRCh38
rs760854164 Chr.22: 41810332-41810332 on Build GRCh38
rs1050658487 Chr.22: 41810358-41810358 on Build GRCh38
rs750946184 Chr.22: 41810300-41810300 on Build GRCh38
rs201589404 Chr.22: 41810276-41810276 on Build GRCh38
rs759442562 Chr.22: 41810169-41810169 on Build GRCh38
rs747876754 Chr.22: 41810233-41810233 on Build GRCh38
rs777891835 Chr.22: 41810193-41810193 on Build GRCh38
rs763042168 Chr.22: 41810170-41810170 on Build GRCh38
rs776081928 Chr.22: 41810232-41810232 on Build GRCh38
rs766175068 Chr.22: 41810171-41810171 on Build GRCh38
rs1005874331 Chr.22: 41810385-41810385 on Build GRCh38
rs552144674 Chr.22: 41810108-41810108 on Build GRCh38
rs1050336965 Chr.22: 41810262-41810262 on Build GRCh38
rs1043734421 Chr.22: 41810291-41810291 on Build GRCh38
rs772910917 Chr.22: 41810236-41810236 on Build GRCh38
rs186574723 Chr.22: 41810299-41810299 on Build GRCh38
rs758686825 Chr.22: 41810301-41810301 on Build GRCh38
rs747784748 Chr.22: 41810310-41810310 on Build GRCh38
rs767583993 Chr.22: 41810184-41810184 on Build GRCh38
rs762956676 Chr.22: 41810238-41810238 on Build GRCh38
rs764847131 Chr.22: 41810168-41810168 on Build GRCh38
rs537418025 Chr.22: 41810239-41810239 on Build GRCh38
rs911232984 Chr.22: 41810270-41810270 on Build GRCh38
rs923112699 Chr.22: 41810442-41810442 on Build GRCh38
rs35004678 Chr.22: 41810124-41810124 on Build GRCh38
rs1010669931 Chr.22: 41810400-41810400 on Build GRCh38
rs139931518 Chr.22: 41810237-41810237 on Build GRCh38
rs975926289 Chr.22: 41810223-41810223 on Build GRCh38
rs5996071 Chr.22: 41810437-41810437 on Build GRCh38
rs757356861 Chr.22: 41810294-41810294 on Build GRCh38
rs6002499 Chr.22: 41810410-41810410 on Build GRCh38
rs769614920 Chr.22: 41810311-41810311 on Build GRCh38
rs780525641 Chr.22: 41810305-41810305 on Build GRCh38
rs147483140 Chr.22: 41810222-41810222 on Build GRCh38
rs961289891 Chr.22: 41810100-41810100 on Build GRCh38
rs779304497 Chr.22: 41810226-41810226 on Build GRCh38
rs777602699 Chr.22: 41810320-41810320 on Build GRCh38
rs751352066 Chr.22: 41810174-41810174 on Build GRCh38
rs1009513396 Chr.22: 41810372-41810373 on Build GRCh38
rs573346956 Chr.22: 41810436-41810436 on Build GRCh38
rs373924812 Chr.22: 41810114-41810114 on Build GRCh38
rs374846901 Chr.22: 41810336-41810336 on Build GRCh38
rs890703969 Chr.22: 41810370-41810371 on Build GRCh38
rs558806801 Chr.22: 41810231-41810231 on Build GRCh38
rs1009048303 Chr.22: 41810107-41810107 on Build GRCh38
rs1031176264 Chr.22: 41810374-41810374 on Build GRCh38
rs4512 Chr.22: 41810080-41810080 on Build GRCh38
rs770485477 Chr.22: 41810323-41810323 on Build GRCh38
rs927848186 Chr.22: 41810227-41810227 on Build GRCh38
rs553878280 Chr.22: 41810414-41810414 on Build GRCh38
rs951722391 Chr.22: 41810146-41810146 on Build GRCh38
rs1022040849 Chr.22: 41810431-41810431 on Build GRCh38
rs371419353 Chr.22: 41810272-41810272 on Build GRCh38
rs767346201 Chr.22: 41810274-41810274 on Build GRCh38
rs774370490 Chr.22: 41810263-41810263 on Build GRCh38
rs746176647 Chr.22: 41810228-41810228 on Build GRCh38
rs574129321 Chr.22: 41810322-41810322 on Build GRCh38
rs35172756 Chr.22: 41810447-41810447 on Build GRCh38
rs773495921 Chr.22: 41810261-41810261 on Build GRCh38
rs771746092 Chr.22: 41810331-41810331 on Build GRCh38
rs754835798 Chr.22: 41810175-41810175 on Build GRCh38
rs952381627 Chr.22: 41810360-41810360 on Build GRCh38
rs961769426 Chr.22: 41810421-41810421 on Build GRCh38
rs370344564 Chr.22: 41810185-41810185 on Build GRCh38

View Full Product Details