Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 42587424-42587937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLDIP3
Entrez Gene ID
84271
Gene Name
DNA polymerase delta interacting protein 3
Gene Aliases
PDIP46, SKAR
UniGene
Hs.505802
Gene Chromosome Location
Chr.22: 42583721-42614962 on Build GRCh38
Transcripts
NM_001278657.1 NM_032311.4 NM_178136.2 NR_103820.1 XM_011530457.1 XM_017029010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3708307 c.1088+4A>C p.? Chr.22: 42587502-42587502 on Build GRCh38
1659096 c.1083C>T p.I361I Chr.22: 42587511-42587511 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373068342 Chr.22: 42587462-42587462 on Build GRCh38
rs969388136 Chr.22: 42587874-42587874 on Build GRCh38
rs978182767 Chr.22: 42587485-42587485 on Build GRCh38
rs1022741267 Chr.22: 42587765-42587765 on Build GRCh38
rs112090029 Chr.22: 42587644-42587644 on Build GRCh38
rs202070345 Chr.22: 42587466-42587466 on Build GRCh38
rs757199091 Chr.22: 42587488-42587488 on Build GRCh38
rs141053448 Chr.22: 42587534-42587534 on Build GRCh38
rs200840342 Chr.22: 42587554-42587554 on Build GRCh38
rs992577784 Chr.22: 42587595-42587595 on Build GRCh38
rs112817282 Chr.22: 42587819-42587819 on Build GRCh38
rs1023463544 Chr.22: 42587797-42587797 on Build GRCh38
rs755100807 Chr.22: 42587538-42587538 on Build GRCh38
rs992146572 Chr.22: 42587846-42587846 on Build GRCh38
rs984689767 Chr.22: 42587822-42587822 on Build GRCh38
rs990689907 Chr.22: 42587790-42587790 on Build GRCh38
rs548538239 Chr.22: 42587908-42587908 on Build GRCh38
rs749794600 Chr.22: 42587626-42587626 on Build GRCh38
rs764947480 Chr.22: 42587798-42587798 on Build GRCh38
rs749689945 Chr.22: 42587577-42587577 on Build GRCh38
rs940461821 Chr.22: 42587708-42587708 on Build GRCh38
rs374907862 Chr.22: 42587616-42587616 on Build GRCh38
rs529304922 Chr.22: 42587717-42587717 on Build GRCh38
rs533142607 Chr.22: 42587758-42587758 on Build GRCh38
rs369429756 Chr.22: 42587811-42587811 on Build GRCh38
rs370385252 Chr.22: 42587467-42587467 on Build GRCh38
rs979466530 Chr.22: 42587876-42587876 on Build GRCh38
rs770808307 Chr.22: 42587605-42587606 on Build GRCh38
rs1043844193 Chr.22: 42587654-42587654 on Build GRCh38
rs772155895 Chr.22: 42587478-42587479 on Build GRCh38
rs753825768 Chr.22: 42587481-42587481 on Build GRCh38
rs2071842 Chr.22: 42587776-42587776 on Build GRCh38
rs182810440 Chr.22: 42587461-42587461 on Build GRCh38
rs748547262 Chr.22: 42587458-42587458 on Build GRCh38
rs771381184 Chr.22: 42587586-42587586 on Build GRCh38
rs541297984 Chr.22: 42587752-42587752 on Build GRCh38
rs760292786 Chr.22: 42587469-42587469 on Build GRCh38
rs57377297 Chr.22: 42587871-42587871 on Build GRCh38
rs972629019 Chr.22: 42587491-42587491 on Build GRCh38
rs902488706 Chr.22: 42587770-42587770 on Build GRCh38
rs960529646 Chr.22: 42587781-42587781 on Build GRCh38
rs572593937 Chr.22: 42587546-42587546 on Build GRCh38
rs761614953 Chr.22: 42587779-42587779 on Build GRCh38
rs200868057 Chr.22: 42587523-42587523 on Build GRCh38
rs559435586 Chr.22: 42587753-42587753 on Build GRCh38
rs773242339 Chr.22: 42587570-42587570 on Build GRCh38
rs371667635 Chr.22: 42587735-42587735 on Build GRCh38
rs902127665 Chr.22: 42587711-42587711 on Build GRCh38
rs200986145 Chr.22: 42587614-42587614 on Build GRCh38
rs1052699179 Chr.22: 42587696-42587696 on Build GRCh38
rs961708342 Chr.22: 42587877-42587877 on Build GRCh38
rs368081483 Chr.22: 42587520-42587520 on Build GRCh38
rs916459942 Chr.22: 42587605-42587605 on Build GRCh38
rs775285342 Chr.22: 42587598-42587598 on Build GRCh38
rs1022250512 Chr.22: 42587474-42587474 on Build GRCh38
rs934128109 Chr.22: 42587749-42587749 on Build GRCh38
rs758140257 Chr.22: 42587517-42587517 on Build GRCh38
rs576613634 Chr.22: 42587580-42587580 on Build GRCh38
rs967231037 Chr.22: 42587480-42587480 on Build GRCh38
rs750270061 Chr.22: 42587490-42587490 on Build GRCh38
rs150238622 Chr.22: 42587565-42587565 on Build GRCh38
rs962566391 Chr.22: 42587473-42587473 on Build GRCh38
rs113181776 Chr.22: 42587669-42587669 on Build GRCh38
rs746889272 Chr.22: 42587575-42587576 on Build GRCh38
rs748251792 Chr.22: 42587547-42587547 on Build GRCh38
rs998298244 Chr.22: 42587454-42587454 on Build GRCh38
rs374735813 Chr.22: 42587477-42587477 on Build GRCh38
rs551172158 Chr.22: 42587764-42587764 on Build GRCh38
rs907520540 Chr.22: 42587704-42587704 on Build GRCh38
rs765191833 Chr.22: 42587489-42587489 on Build GRCh38
rs763376656 Chr.22: 42587463-42587463 on Build GRCh38

View Full Product Details