Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 43939907-43940332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNPLA3
Entrez Gene ID
80339
Gene Name
patatin like phospholipase domain containing 3
Gene Aliases
ADPN, C22orf20, iPLA(2)epsilon
UniGene
Hs.654800
Gene Chromosome Location
Chr.22: 43923739-43947568 on Build GRCh38
Transcripts
NM_025225.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6578582 c.995T>C p.M332T Chr.22: 43940008-43940008 on Build GRCh38
6489829 c.1003A>G p.K335E Chr.22: 43940016-43940016 on Build GRCh38
7165188 c.991G>C p.E331Q Chr.22: 43940004-43940004 on Build GRCh38
5755769 c.1031A>G p.N344S Chr.22: 43940044-43940044 on Build GRCh38
5474457 c.1100C>T p.A367V Chr.22: 43940113-43940113 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199525362 Chr.22: 43940082-43940082 on Build GRCh38
rs987672589 Chr.22: 43940250-43940250 on Build GRCh38
rs765574516 Chr.22: 43939965-43939965 on Build GRCh38
rs760916944 Chr.22: 43940054-43940054 on Build GRCh38
rs890013575 Chr.22: 43940168-43940168 on Build GRCh38
rs144233415 Chr.22: 43940005-43940005 on Build GRCh38
rs112092226 Chr.22: 43940143-43940143 on Build GRCh38
rs763431870 Chr.22: 43940174-43940174 on Build GRCh38
rs144872932 Chr.22: 43939978-43939978 on Build GRCh38
rs1006866942 Chr.22: 43939961-43939961 on Build GRCh38
rs201680395 Chr.22: 43940147-43940147 on Build GRCh38
rs373663693 Chr.22: 43940095-43940095 on Build GRCh38
rs55743053 Chr.22: 43940227-43940227 on Build GRCh38
rs1021566754 Chr.22: 43940100-43940100 on Build GRCh38
rs766701461 Chr.22: 43940091-43940091 on Build GRCh38
rs777316809 Chr.22: 43939984-43939984 on Build GRCh38
rs187206372 Chr.22: 43940141-43940141 on Build GRCh38
rs533961551 Chr.22: 43940076-43940076 on Build GRCh38
rs1021891823 Chr.22: 43940279-43940279 on Build GRCh38
rs943034220 Chr.22: 43940261-43940261 on Build GRCh38
rs1031462628 Chr.22: 43939993-43939993 on Build GRCh38
rs750778191 Chr.22: 43939967-43939967 on Build GRCh38
rs138736228 Chr.22: 43940139-43940139 on Build GRCh38
rs574351066 Chr.22: 43940178-43940178 on Build GRCh38
rs745670695 Chr.22: 43940023-43940023 on Build GRCh38
rs868306603 Chr.22: 43940249-43940249 on Build GRCh38
rs990012953 Chr.22: 43940019-43940019 on Build GRCh38
rs757757471 Chr.22: 43939946-43939946 on Build GRCh38
rs769830028 Chr.22: 43940169-43940169 on Build GRCh38
rs367920030 Chr.22: 43939995-43939995 on Build GRCh38
rs781212740 Chr.22: 43939979-43939979 on Build GRCh38
rs754340054 Chr.22: 43939945-43939945 on Build GRCh38
rs755622820 Chr.22: 43939981-43939981 on Build GRCh38
rs1001862478 Chr.22: 43940304-43940304 on Build GRCh38
rs912170847 Chr.22: 43940253-43940253 on Build GRCh38
rs370920329 Chr.22: 43940086-43940086 on Build GRCh38
rs371036359 Chr.22: 43939963-43939963 on Build GRCh38
rs749956077 Chr.22: 43940303-43940303 on Build GRCh38
rs756663646 Chr.22: 43940114-43940114 on Build GRCh38
rs768665435 Chr.22: 43940059-43940059 on Build GRCh38
rs765749635 Chr.22: 43940083-43940083 on Build GRCh38
rs753393489 Chr.22: 43940113-43940113 on Build GRCh38
rs560451808 Chr.22: 43940220-43940220 on Build GRCh38
rs1049055644 Chr.22: 43940167-43940167 on Build GRCh38
rs771943403 Chr.22: 43940031-43940031 on Build GRCh38
rs969968470 Chr.22: 43940288-43940288 on Build GRCh38
rs778223816 Chr.22: 43940211-43940211 on Build GRCh38
rs1041010212 Chr.22: 43940177-43940177 on Build GRCh38
rs898935161 Chr.22: 43940073-43940073 on Build GRCh38
rs911566723 Chr.22: 43939962-43939962 on Build GRCh38
rs369346918 Chr.22: 43940146-43940146 on Build GRCh38
rs773751210 Chr.22: 43940171-43940171 on Build GRCh38
rs1011342735 Chr.22: 43940274-43940274 on Build GRCh38
rs563753915 Chr.22: 43939999-43939999 on Build GRCh38
rs1033755543 Chr.22: 43940151-43940151 on Build GRCh38
rs141163247 Chr.22: 43939950-43939950 on Build GRCh38
rs758707816 Chr.22: 43939976-43939976 on Build GRCh38
rs891613733 Chr.22: 43940266-43940266 on Build GRCh38
rs942952807 Chr.22: 43940170-43940170 on Build GRCh38
rs552354116 Chr.22: 43940273-43940273 on Build GRCh38
rs200563318 Chr.22: 43940150-43940150 on Build GRCh38
rs558775561 Chr.22: 43939944-43939944 on Build GRCh38
rs146059709 Chr.22: 43940035-43940035 on Build GRCh38
rs141538262 Chr.22: 43940162-43940162 on Build GRCh38
rs770898005 Chr.22: 43940000-43940000 on Build GRCh38

View Full Product Details