Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45359716-45360226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMC1B
Entrez Gene ID
27127
Gene Name
structural maintenance of chromosomes 1B
Gene Aliases
SMC1BETA, SMC1L2
UniGene
Hs.334176
Gene Chromosome Location
Chr.22: 45344063-45413671 on Build GRCh38
Transcripts
NM_001291501.1 NM_148674.4 XM_011530144.2 XM_011530145.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4104732 c.2839G>T p.D947Y Chr.22: 45359822-45359822 on Build GRCh38
3063059 c.2735G>A p.S912N Chr.22: 45359926-45359926 on Build GRCh38
4104734 c.2751G>A p.L917L Chr.22: 45359910-45359910 on Build GRCh38
1484285 c.2860G>A p.E954K Chr.22: 45359807-45359807 on Build GRCh38
1484286 c.2854G>A p.E952K Chr.22: 45359807-45359807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989797617 Chr.22: 45359924-45359924 on Build GRCh38
rs932968836 Chr.22: 45360177-45360177 on Build GRCh38
rs759167793 Chr.22: 45359756-45359756 on Build GRCh38
rs1037200993 Chr.22: 45360002-45360002 on Build GRCh38
rs1049101486 Chr.22: 45360033-45360033 on Build GRCh38
rs746619556 Chr.22: 45359971-45359971 on Build GRCh38
rs758562337 Chr.22: 45359909-45359909 on Build GRCh38
rs1012493 Chr.22: 45359786-45359786 on Build GRCh38
rs766577785 Chr.22: 45359910-45359910 on Build GRCh38
rs758324682 Chr.22: 45359976-45359977 on Build GRCh38
rs576346887 Chr.22: 45359911-45359911 on Build GRCh38
rs1012494 Chr.22: 45360140-45360140 on Build GRCh38
rs267606273 Chr.22: 45359846-45359846 on Build GRCh38
rs528783819 Chr.22: 45359753-45359753 on Build GRCh38
rs771600283 Chr.22: 45359956-45359956 on Build GRCh38
rs777136131 Chr.22: 45359993-45359993 on Build GRCh38
rs770352207 Chr.22: 45360084-45360084 on Build GRCh38
rs557392959 Chr.22: 45359898-45359898 on Build GRCh38
rs754330325 Chr.22: 45359778-45359778 on Build GRCh38
rs192489047 Chr.22: 45359976-45359976 on Build GRCh38
rs926097379 Chr.22: 45360074-45360074 on Build GRCh38
rs764612184 Chr.22: 45359773-45359773 on Build GRCh38
rs916279422 Chr.22: 45359926-45359926 on Build GRCh38
rs747915950 Chr.22: 45359820-45359820 on Build GRCh38
rs34743320 Chr.22: 45360046-45360046 on Build GRCh38
rs778828404 Chr.22: 45359945-45359945 on Build GRCh38
rs758684037 Chr.22: 45359815-45359815 on Build GRCh38
rs773874445 Chr.22: 45360000-45360000 on Build GRCh38
rs73442488 Chr.22: 45359968-45359968 on Build GRCh38
rs777108869 Chr.22: 45359933-45359935 on Build GRCh38
rs755577782 Chr.22: 45359930-45359930 on Build GRCh38
rs776382291 Chr.22: 45360121-45360121 on Build GRCh38
rs557636660 Chr.22: 45360073-45360073 on Build GRCh38
rs760144787 Chr.22: 45359929-45359931 on Build GRCh38
rs546593612 Chr.22: 45359835-45359835 on Build GRCh38
rs999611094 Chr.22: 45360141-45360141 on Build GRCh38
rs571820642 Chr.22: 45359839-45359839 on Build GRCh38
rs761058934 Chr.22: 45359872-45359872 on Build GRCh38
rs147786046 Chr.22: 45359934-45359934 on Build GRCh38
rs780450249 Chr.22: 45359772-45359772 on Build GRCh38
rs142507356 Chr.22: 45360144-45360144 on Build GRCh38
rs759116364 Chr.22: 45359852-45359852 on Build GRCh38
rs780426114 Chr.22: 45359818-45359818 on Build GRCh38
rs762268145 Chr.22: 45359992-45359992 on Build GRCh38
rs777102135 Chr.22: 45359888-45359888 on Build GRCh38
rs771647394 Chr.22: 45359857-45359857 on Build GRCh38
rs769688508 Chr.22: 45360152-45360152 on Build GRCh38
rs756090320 Chr.22: 45359786-45359789 on Build GRCh38
rs755272393 Chr.22: 45359923-45359923 on Build GRCh38
rs889811251 Chr.22: 45360062-45360062 on Build GRCh38
rs778999861 Chr.22: 45359907-45359907 on Build GRCh38
rs753103187 Chr.22: 45359767-45359767 on Build GRCh38
rs1051955328 Chr.22: 45360189-45360189 on Build GRCh38
rs148863291 Chr.22: 45360008-45360008 on Build GRCh38
rs767216670 Chr.22: 45359765-45359765 on Build GRCh38
rs749153217 Chr.22: 45359837-45359837 on Build GRCh38
rs750458916 Chr.22: 45359906-45359906 on Build GRCh38
rs765906991 Chr.22: 45359899-45359899 on Build GRCh38
rs537268164 Chr.22: 45359876-45359876 on Build GRCh38
rs745324457 Chr.22: 45359954-45359954 on Build GRCh38
rs374035924 Chr.22: 45360106-45360106 on Build GRCh38
rs114160772 Chr.22: 45360025-45360025 on Build GRCh38
rs779112241 Chr.22: 45359785-45359785 on Build GRCh38
rs777412324 Chr.22: 45359925-45359925 on Build GRCh38
rs201958107 Chr.22: 45359836-45359836 on Build GRCh38
rs367972523 Chr.22: 45360097-45360097 on Build GRCh38
rs757181704 Chr.22: 45359940-45359940 on Build GRCh38
rs757878947 Chr.22: 45359781-45359781 on Build GRCh38
rs200018788 Chr.22: 45359889-45359889 on Build GRCh38
rs1569413 Chr.22: 45360201-45360201 on Build GRCh38
rs773894242 Chr.22: 45359755-45359755 on Build GRCh38
rs770905388 Chr.22: 45359838-45359838 on Build GRCh38
rs775026667 Chr.22: 45359859-45359859 on Build GRCh38

View Full Product Details